Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69447052-69447346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5B
Entrez Gene ID
80777
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M, CYPB5M, OMB5
UniGene
Hs.461131
Gene Chromosome Location
Chr.16: 69424595-69466264 on Build GRCh38
Transcripts
NM_030579.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972892 c.290G>T p.G97V Chr.16: 69447265-69447265 on Build GRCh38
4532707 c.190G>A p.E64K Chr.16: 69447165-69447165 on Build GRCh38
472006 c.258T>A p.D86E Chr.16: 69447233-69447233 on Build GRCh38
472008 c.260C>T p.A87V Chr.16: 69447235-69447235 on Build GRCh38
472007 c.259G>A p.A87T Chr.16: 69447234-69447234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776642532 Chr.16: 69447258-69447258 on Build GRCh38
rs189673648 Chr.16: 69447323-69447323 on Build GRCh38
rs753948929 Chr.16: 69447112-69447112 on Build GRCh38
rs886086637 Chr.16: 69447241-69447241 on Build GRCh38
rs543345769 Chr.16: 69447308-69447308 on Build GRCh38
rs113477024 Chr.16: 69447279-69447279 on Build GRCh38
rs755783941 Chr.16: 69447174-69447174 on Build GRCh38
rs369430739 Chr.16: 69447313-69447313 on Build GRCh38
rs760873085 Chr.16: 69447255-69447255 on Build GRCh38
rs768091984 Chr.16: 69447213-69447213 on Build GRCh38
rs746056755 Chr.16: 69447170-69447170 on Build GRCh38
rs750215844 Chr.16: 69447318-69447318 on Build GRCh38
rs772373417 Chr.16: 69447246-69447246 on Build GRCh38
rs763042234 Chr.16: 69447119-69447119 on Build GRCh38
rs751501732 Chr.16: 69447155-69447155 on Build GRCh38
rs776649212 Chr.16: 69447265-69447265 on Build GRCh38
rs770235130 Chr.16: 69447291-69447292 on Build GRCh38
rs771172165 Chr.16: 69447259-69447259 on Build GRCh38
rs868361051 Chr.16: 69447223-69447227 on Build GRCh38
rs957961870 Chr.16: 69447224-69447224 on Build GRCh38
rs764131629 Chr.16: 69447133-69447133 on Build GRCh38
rs748814259 Chr.16: 69447200-69447200 on Build GRCh38
rs949854784 Chr.16: 69447122-69447122 on Build GRCh38
rs377098912 Chr.16: 69447277-69447277 on Build GRCh38
rs199873890 Chr.16: 69447234-69447234 on Build GRCh38
rs765357837 Chr.16: 69447278-69447278 on Build GRCh38
rs966497115 Chr.16: 69447117-69447117 on Build GRCh38
rs181501333 Chr.16: 69447173-69447173 on Build GRCh38
rs779431166 Chr.16: 69447214-69447214 on Build GRCh38
rs201614686 Chr.16: 69447168-69447168 on Build GRCh38
rs113061257 Chr.16: 69447280-69447280 on Build GRCh38
rs773597307 Chr.16: 69447248-69447248 on Build GRCh38
rs748608087 Chr.16: 69447238-69447238 on Build GRCh38
rs1048695268 Chr.16: 69447208-69447208 on Build GRCh38
rs184627050 Chr.16: 69447197-69447197 on Build GRCh38

View Full Product Details