Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7539855-7540411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA6
Entrez Gene ID
10908
Gene Name
patatin like phospholipase domain containing 6
Gene Aliases
BNHS, LNMS, NTE, NTEMND, OMCS, SPG39, iPLA2delta, sws
UniGene
Hs.631863
Gene Chromosome Location
Chr.19: 7534152-7561767 on Build GRCh38
Transcripts
NM_001166111.1 NM_001166112.1 NM_001166113.1 NM_001166114.1 NM_006702.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307393 c.532G>T p.A178S Chr.19: 7540009-7540009 on Build GRCh38
7196241 c.588G>A p.L196L Chr.19: 7540155-7540155 on Build GRCh38
1663602 c.549C>A p.S183R Chr.19: 7540260-7540260 on Build GRCh38
6307394 c.388G>T p.A130S Chr.19: 7540009-7540009 on Build GRCh38
1590431 c.597C>T p.F199F Chr.19: 7540164-7540164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200521839 Chr.19: 7540215-7540215 on Build GRCh38
rs747209749 Chr.19: 7540251-7540251 on Build GRCh38
rs149630061 Chr.19: 7539900-7539900 on Build GRCh38
rs764125789 Chr.19: 7539925-7539925 on Build GRCh38
rs183762415 Chr.19: 7540129-7540129 on Build GRCh38
rs368637145 Chr.19: 7540314-7540314 on Build GRCh38
rs955026991 Chr.19: 7539881-7539881 on Build GRCh38
rs375424103 Chr.19: 7540278-7540278 on Build GRCh38
rs767086941 Chr.19: 7540341-7540341 on Build GRCh38
rs761091412 Chr.19: 7540106-7540106 on Build GRCh38
rs534775987 Chr.19: 7540246-7540246 on Build GRCh38
rs376396266 Chr.19: 7539956-7539956 on Build GRCh38
rs965642578 Chr.19: 7539922-7539922 on Build GRCh38
rs760557592 Chr.19: 7539918-7539918 on Build GRCh38
rs569943531 Chr.19: 7540194-7540194 on Build GRCh38
rs780995803 Chr.19: 7540223-7540223 on Build GRCh38
rs915372009 Chr.19: 7540032-7540032 on Build GRCh38
rs759699733 Chr.19: 7540130-7540130 on Build GRCh38
rs778359563 Chr.19: 7539996-7539996 on Build GRCh38
rs776624904 Chr.19: 7540083-7540083 on Build GRCh38
rs868697445 Chr.19: 7539931-7539931 on Build GRCh38
rs543360288 Chr.19: 7540367-7540367 on Build GRCh38
rs953147743 Chr.19: 7539904-7539904 on Build GRCh38
rs985812017 Chr.19: 7539908-7539908 on Build GRCh38
rs751202036 Chr.19: 7540213-7540213 on Build GRCh38
rs773675660 Chr.19: 7540305-7540305 on Build GRCh38
rs374388554 Chr.19: 7540329-7540329 on Build GRCh38
rs778438044 Chr.19: 7540053-7540053 on Build GRCh38
rs773099865 Chr.19: 7539926-7539926 on Build GRCh38
rs766433753 Chr.19: 7539948-7539948 on Build GRCh38
rs558004229 Chr.19: 7540266-7540266 on Build GRCh38
rs771735161 Chr.19: 7540065-7540065 on Build GRCh38
rs535890270 Chr.19: 7540211-7540211 on Build GRCh38
rs1018645754 Chr.19: 7540231-7540231 on Build GRCh38
rs771196056 Chr.19: 7539911-7539913 on Build GRCh38
rs762180901 Chr.19: 7540302-7540302 on Build GRCh38
rs754588559 Chr.19: 7540333-7540333 on Build GRCh38
rs775314229 Chr.19: 7539916-7539916 on Build GRCh38
rs10405345 Chr.19: 7540385-7540385 on Build GRCh38
rs370828407 Chr.19: 7540002-7540002 on Build GRCh38
rs374075198 Chr.19: 7540123-7540123 on Build GRCh38
rs768391928 Chr.19: 7540081-7540081 on Build GRCh38
rs757980617 Chr.19: 7540115-7540115 on Build GRCh38
rs753050856 Chr.19: 7540104-7540104 on Build GRCh38
rs780897059 Chr.19: 7540127-7540127 on Build GRCh38
rs202211650 Chr.19: 7540107-7540107 on Build GRCh38
rs756791412 Chr.19: 7540021-7540021 on Build GRCh38
rs566213812 Chr.19: 7540242-7540242 on Build GRCh38
rs373136941 Chr.19: 7540128-7540128 on Build GRCh38
rs770435865 Chr.19: 7539896-7539896 on Build GRCh38
rs772339510 Chr.19: 7540135-7540135 on Build GRCh38
rs201099946 Chr.19: 7540136-7540136 on Build GRCh38
rs767940871 Chr.19: 7539973-7539973 on Build GRCh38
rs367968144 Chr.19: 7540347-7540347 on Build GRCh38
rs888912174 Chr.19: 7540138-7540138 on Build GRCh38
rs983462140 Chr.19: 7539946-7539946 on Build GRCh38
rs749967266 Chr.19: 7540114-7540114 on Build GRCh38
rs757757050 Chr.19: 7540111-7540111 on Build GRCh38
rs765649401 Chr.19: 7540304-7540304 on Build GRCh38
rs983596236 Chr.19: 7540355-7540355 on Build GRCh38
rs766088580 Chr.19: 7540093-7540093 on Build GRCh38
rs577820446 Chr.19: 7540361-7540361 on Build GRCh38
rs774591720 Chr.19: 7540098-7540098 on Build GRCh38
rs559066236 Chr.19: 7539942-7539942 on Build GRCh38
rs749879325 Chr.19: 7540193-7540193 on Build GRCh38
rs746356492 Chr.19: 7540134-7540134 on Build GRCh38
rs150620708 Chr.19: 7540243-7540243 on Build GRCh38
rs775227844 Chr.19: 7540070-7540070 on Build GRCh38
rs779697397 Chr.19: 7540116-7540116 on Build GRCh38
rs761695365 Chr.19: 7540089-7540089 on Build GRCh38
rs375141952 Chr.19: 7540099-7540099 on Build GRCh38
rs754176901 Chr.19: 7540167-7540167 on Build GRCh38
rs527855771 Chr.19: 7540102-7540102 on Build GRCh38
rs746826877 Chr.19: 7540071-7540071 on Build GRCh38
rs999360872 Chr.19: 7540206-7540206 on Build GRCh38
rs111278968 Chr.19: 7540292-7540292 on Build GRCh38
rs771827201 Chr.19: 7539907-7539907 on Build GRCh38
rs1014805676 Chr.19: 7539879-7539879 on Build GRCh38
rs768464955 Chr.19: 7540122-7540122 on Build GRCh38
rs1007265464 Chr.19: 7540164-7540164 on Build GRCh38
rs746705186 Chr.19: 7540117-7540117 on Build GRCh38
rs762342193 Chr.19: 7540173-7540173 on Build GRCh38
rs777235341 Chr.19: 7540069-7540070 on Build GRCh38
rs769793855 Chr.19: 7540238-7540238 on Build GRCh38
rs777837588 Chr.19: 7540241-7540241 on Build GRCh38
rs778346004 Chr.19: 7540113-7540113 on Build GRCh38
rs776987928 Chr.19: 7540293-7540293 on Build GRCh38
rs763499274 Chr.19: 7540307-7540307 on Build GRCh38
rs371188270 Chr.19: 7540315-7540315 on Build GRCh38
rs755300288 Chr.19: 7539972-7539972 on Build GRCh38
rs865966998 Chr.19: 7539941-7539941 on Build GRCh38
rs542345815 Chr.19: 7539901-7539901 on Build GRCh38
rs756508790 Chr.19: 7539979-7539979 on Build GRCh38
rs765739232 Chr.19: 7540176-7540176 on Build GRCh38
rs745499228 Chr.19: 7540064-7540064 on Build GRCh38
rs200634227 Chr.19: 7539895-7539895 on Build GRCh38
rs924207953 Chr.19: 7539955-7539955 on Build GRCh38
rs764501990 Chr.19: 7540146-7540146 on Build GRCh38
rs543377698 Chr.19: 7539891-7539891 on Build GRCh38
rs747896630 Chr.19: 7540230-7540230 on Build GRCh38
rs911565175 Chr.19: 7539914-7539914 on Build GRCh38

View Full Product Details