Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18391024-18391614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC25
Entrez Gene ID
126364
Gene Name
leucine rich repeat containing 25
Gene Aliases
MAPA
UniGene
Hs.332156
Gene Chromosome Location
Chr.19: 18391137-18397642 on Build GRCh38
Transcripts
NM_145256.2 XM_005259739.3

SNP Detail

SNP ID Chromosome Location
rs34401419 Chr.19: 18391171-18391172 on Build GRCh38
rs966811479 Chr.19: 18391134-18391134 on Build GRCh38
rs775725017 Chr.19: 18391336-18391336 on Build GRCh38
rs913894780 Chr.19: 18391141-18391141 on Build GRCh38
rs995108152 Chr.19: 18391560-18391560 on Build GRCh38
rs891339526 Chr.19: 18391202-18391202 on Build GRCh38
rs773717897 Chr.19: 18391181-18391181 on Build GRCh38
rs555022758 Chr.19: 18391493-18391493 on Build GRCh38
rs900753224 Chr.19: 18391125-18391125 on Build GRCh38
rs979873291 Chr.19: 18391411-18391411 on Build GRCh38
rs960466897 Chr.19: 18391196-18391196 on Build GRCh38
rs145536188 Chr.19: 18391472-18391472 on Build GRCh38
rs3848649 Chr.19: 18391527-18391527 on Build GRCh38
rs116399833 Chr.19: 18391564-18391564 on Build GRCh38
rs1011611853 Chr.19: 18391128-18391128 on Build GRCh38
rs570983901 Chr.19: 18391433-18391433 on Build GRCh38
rs951299210 Chr.19: 18391549-18391555 on Build GRCh38
rs1003025695 Chr.19: 18391567-18391567 on Build GRCh38
rs528350130 Chr.19: 18391540-18391543 on Build GRCh38
rs934574113 Chr.19: 18391133-18391133 on Build GRCh38
rs898165708 Chr.19: 18391277-18391277 on Build GRCh38
rs62122433 Chr.19: 18391407-18391407 on Build GRCh38
rs747512691 Chr.19: 18391189-18391189 on Build GRCh38
rs768091320 Chr.19: 18391531-18391532 on Build GRCh38
rs563907762 Chr.19: 18391053-18391053 on Build GRCh38
rs902786999 Chr.19: 18391430-18391430 on Build GRCh38
rs4328570 Chr.19: 18391491-18391491 on Build GRCh38
rs995160253 Chr.19: 18391305-18391305 on Build GRCh38
rs946737998 Chr.19: 18391154-18391154 on Build GRCh38
rs780657362 Chr.19: 18391499-18391499 on Build GRCh38
rs369692302 Chr.19: 18391561-18391561 on Build GRCh38
rs928543096 Chr.19: 18391126-18391126 on Build GRCh38
rs1010100968 Chr.19: 18391580-18391580 on Build GRCh38
rs1021522651 Chr.19: 18391583-18391583 on Build GRCh38
rs12461382 Chr.19: 18391265-18391265 on Build GRCh38
rs999636242 Chr.19: 18391469-18391469 on Build GRCh38
rs766238793 Chr.19: 18391228-18391228 on Build GRCh38
rs912715476 Chr.19: 18391494-18391494 on Build GRCh38
rs748739825 Chr.19: 18391099-18391099 on Build GRCh38
rs1043813940 Chr.19: 18391168-18391168 on Build GRCh38
rs6512261 Chr.19: 18391483-18391483 on Build GRCh38
rs1010910507 Chr.19: 18391406-18391406 on Build GRCh38
rs989130279 Chr.19: 18391397-18391397 on Build GRCh38
rs3848648 Chr.19: 18391309-18391309 on Build GRCh38
rs374032828 Chr.19: 18391554-18391557 on Build GRCh38
rs1008887722 Chr.19: 18391118-18391118 on Build GRCh38
rs386807493 Chr.19: 18391525-18391527 on Build GRCh38
rs1018047528 Chr.19: 18391122-18391122 on Build GRCh38
rs946679619 Chr.19: 18391582-18391582 on Build GRCh38
rs537507634 Chr.19: 18391396-18391396 on Build GRCh38
rs946685934 Chr.19: 18391415-18391415 on Build GRCh38
rs189595546 Chr.19: 18391569-18391569 on Build GRCh38
rs956047825 Chr.19: 18391579-18391579 on Build GRCh38
rs898782045 Chr.19: 18391553-18391553 on Build GRCh38
rs990497698 Chr.19: 18391254-18391254 on Build GRCh38
rs986630473 Chr.19: 18391562-18391562 on Build GRCh38
rs549858882 Chr.19: 18391140-18391140 on Build GRCh38
rs3848647 Chr.19: 18391268-18391268 on Build GRCh38
rs1035938607 Chr.19: 18391389-18391389 on Build GRCh38
rs981256856 Chr.19: 18391347-18391347 on Build GRCh38
rs3848646 Chr.19: 18391176-18391176 on Build GRCh38
rs112738452 Chr.19: 18391525-18391525 on Build GRCh38
rs780730013 Chr.19: 18391564-18391564 on Build GRCh38
rs542512902 Chr.19: 18391492-18391492 on Build GRCh38
rs960435773 Chr.19: 18391474-18391474 on Build GRCh38
rs1050942948 Chr.19: 18391310-18391310 on Build GRCh38
rs1035483375 Chr.19: 18391570-18391570 on Build GRCh38
rs1047817188 Chr.19: 18391050-18391050 on Build GRCh38
rs529232798 Chr.19: 18391291-18391291 on Build GRCh38
rs916266276 Chr.19: 18391258-18391258 on Build GRCh38
rs372041307 Chr.19: 18391360-18391360 on Build GRCh38
rs771323424 Chr.19: 18391236-18391236 on Build GRCh38
rs568471993 Chr.19: 18391378-18391378 on Build GRCh38

View Full Product Details