Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36637124-36637717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF461
Entrez Gene ID
92283
Gene Name
zinc finger protein 461
Gene Aliases
GIOT-1, GIOT1, HZF28
UniGene
Hs.590972
Gene Chromosome Location
Chr.19: 36636621-36666853 on Build GRCh38
Transcripts
NM_001297623.2 NM_001322821.1 NM_001322823.1 NM_001322825.1 NM_001322826.1 NM_001322827.1 NM_001322828.1 NM_153257.4 XM_011527486.2 XM_017027470.1

SNP Detail

SNP ID Chromosome Location
rs889031589 Chr.19: 36637603-36637603 on Build GRCh38
rs183475354 Chr.19: 36637442-36637442 on Build GRCh38
rs539852367 Chr.19: 36637667-36637667 on Build GRCh38
rs1048421369 Chr.19: 36637554-36637554 on Build GRCh38
rs1043687112 Chr.19: 36637599-36637599 on Build GRCh38
rs1047983309 Chr.19: 36637363-36637363 on Build GRCh38
rs949028666 Chr.19: 36637321-36637321 on Build GRCh38
rs1880563 Chr.19: 36637173-36637173 on Build GRCh38
rs909479026 Chr.19: 36637568-36637568 on Build GRCh38
rs900582966 Chr.19: 36637649-36637649 on Build GRCh38
rs916147993 Chr.19: 36637478-36637478 on Build GRCh38
rs964682581 Chr.19: 36637158-36637158 on Build GRCh38
rs530444091 Chr.19: 36637443-36637443 on Build GRCh38
rs528242708 Chr.19: 36637467-36637467 on Build GRCh38
rs966638457 Chr.19: 36637200-36637200 on Build GRCh38
rs942177943 Chr.19: 36637570-36637570 on Build GRCh38
rs972097159 Chr.19: 36637434-36637434 on Build GRCh38
rs187337301 Chr.19: 36637629-36637629 on Build GRCh38
rs546474934 Chr.19: 36637501-36637501 on Build GRCh38
rs3108173 Chr.19: 36637402-36637402 on Build GRCh38
rs113305151 Chr.19: 36637391-36637391 on Build GRCh38
rs977914234 Chr.19: 36637241-36637241 on Build GRCh38
rs990683155 Chr.19: 36637480-36637480 on Build GRCh38
rs79595417 Chr.19: 36637209-36637209 on Build GRCh38
rs78047429 Chr.19: 36637208-36637208 on Build GRCh38
rs1007431965 Chr.19: 36637628-36637628 on Build GRCh38
rs1018075481 Chr.19: 36637416-36637416 on Build GRCh38
rs570662586 Chr.19: 36637452-36637452 on Build GRCh38
rs1030238095 Chr.19: 36637194-36637194 on Build GRCh38
rs1011216530 Chr.19: 36637390-36637390 on Build GRCh38
rs192821747 Chr.19: 36637260-36637260 on Build GRCh38
rs535712052 Chr.19: 36637353-36637359 on Build GRCh38
rs760688513 Chr.19: 36637197-36637201 on Build GRCh38
rs188071422 Chr.19: 36637257-36637257 on Build GRCh38
rs900615616 Chr.19: 36637436-36637436 on Build GRCh38
rs1018753630 Chr.19: 36637674-36637674 on Build GRCh38
rs952344976 Chr.19: 36637449-36637449 on Build GRCh38
rs549722492 Chr.19: 36637354-36637354 on Build GRCh38
rs936528468 Chr.19: 36637258-36637258 on Build GRCh38
rs541560456 Chr.19: 36637437-36637437 on Build GRCh38
rs946390386 Chr.19: 36637564-36637564 on Build GRCh38
rs892096091 Chr.19: 36637561-36637561 on Build GRCh38
rs997633172 Chr.19: 36637650-36637650 on Build GRCh38
rs907446818 Chr.19: 36637328-36637328 on Build GRCh38
rs552016707 Chr.19: 36637446-36637446 on Build GRCh38
rs1032264609 Chr.19: 36637447-36637447 on Build GRCh38
rs114641629 Chr.19: 36637545-36637545 on Build GRCh38
rs377169661 Chr.19: 36637683-36637683 on Build GRCh38
rs939060577 Chr.19: 36637553-36637553 on Build GRCh38
rs966108652 Chr.19: 36637424-36637424 on Build GRCh38
rs953460818 Chr.19: 36637196-36637197 on Build GRCh38
rs573346391 Chr.19: 36637353-36637353 on Build GRCh38
rs557016002 Chr.19: 36637362-36637363 on Build GRCh38
rs779287639 Chr.19: 36637511-36637511 on Build GRCh38
rs138705278 Chr.19: 36637517-36637517 on Build GRCh38
rs970475398 Chr.19: 36637516-36637516 on Build GRCh38
rs768496892 Chr.19: 36637322-36637322 on Build GRCh38
rs546492925 Chr.19: 36637242-36637242 on Build GRCh38
rs990937640 Chr.19: 36637259-36637259 on Build GRCh38
rs368417117 Chr.19: 36637159-36637159 on Build GRCh38
rs1039226737 Chr.19: 36637625-36637625 on Build GRCh38
rs1003196846 Chr.19: 36637371-36637372 on Build GRCh38
rs568814779 Chr.19: 36637412-36637412 on Build GRCh38
rs887903772 Chr.19: 36637378-36637378 on Build GRCh38
rs892384956 Chr.19: 36637372-36637372 on Build GRCh38
rs966057076 Chr.19: 36637201-36637201 on Build GRCh38
rs927254757 Chr.19: 36637431-36637431 on Build GRCh38
rs557514139 Chr.19: 36637552-36637552 on Build GRCh38
rs375002840 Chr.19: 36637342-36637342 on Build GRCh38
rs986184275 Chr.19: 36637197-36637197 on Build GRCh38
rs972468494 Chr.19: 36637210-36637214 on Build GRCh38
rs919549995 Chr.19: 36637236-36637236 on Build GRCh38
rs916352981 Chr.19: 36637281-36637281 on Build GRCh38

View Full Product Details