Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40205857-40206288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K10
Entrez Gene ID
4294
Gene Name
mitogen-activated protein kinase kinase kinase 10
Gene Aliases
MEKK10, MLK2, MST
UniGene
Hs.466743
Gene Chromosome Location
Chr.19: 40191657-40215575 on Build GRCh38
Transcripts
NM_002446.3 XM_006723222.2 XM_006723223.3 XM_011526981.2 XM_011526982.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6988071 c.1267G>T p.E423* Chr.19: 40205989-40205989 on Build GRCh38
996611 c.1264C>T p.R422W Chr.19: 40205986-40205986 on Build GRCh38
5605867 c.1299C>T p.I433I Chr.19: 40206021-40206021 on Build GRCh38
996609 c.1219C>T p.R407W Chr.19: 40205941-40205941 on Build GRCh38
48521 c.1388G>C p.R463P Chr.19: 40206110-40206110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761641528 Chr.19: 40205994-40205994 on Build GRCh38
rs915531870 Chr.19: 40206207-40206207 on Build GRCh38
rs895664482 Chr.19: 40205951-40205951 on Build GRCh38
rs750419573 Chr.19: 40205968-40205980 on Build GRCh38
rs750945146 Chr.19: 40206181-40206181 on Build GRCh38
rs777624809 Chr.19: 40206196-40206196 on Build GRCh38
rs765220429 Chr.19: 40205916-40205916 on Build GRCh38
rs749106630 Chr.19: 40206125-40206125 on Build GRCh38
rs770974520 Chr.19: 40206129-40206129 on Build GRCh38
rs376827274 Chr.19: 40205922-40205922 on Build GRCh38
rs758760290 Chr.19: 40205940-40205940 on Build GRCh38
rs751627089 Chr.19: 40206055-40206055 on Build GRCh38
rs954750202 Chr.19: 40206150-40206150 on Build GRCh38
rs769604843 Chr.19: 40206117-40206117 on Build GRCh38
rs767775266 Chr.19: 40206079-40206079 on Build GRCh38
rs779474272 Chr.19: 40205975-40205975 on Build GRCh38
rs371667509 Chr.19: 40206086-40206086 on Build GRCh38
rs768199803 Chr.19: 40205879-40205879 on Build GRCh38
rs764392464 Chr.19: 40206088-40206088 on Build GRCh38
rs1007992552 Chr.19: 40206155-40206155 on Build GRCh38
rs747695251 Chr.19: 40206186-40206186 on Build GRCh38
rs367847300 Chr.19: 40205923-40205923 on Build GRCh38
rs769530363 Chr.19: 40206004-40206004 on Build GRCh38
rs778221515 Chr.19: 40205949-40205949 on Build GRCh38
rs534097924 Chr.19: 40206103-40206103 on Build GRCh38
rs774321395 Chr.19: 40206209-40206209 on Build GRCh38
rs992591551 Chr.19: 40206212-40206212 on Build GRCh38
rs776214953 Chr.19: 40205986-40205986 on Build GRCh38
rs765702617 Chr.19: 40206180-40206180 on Build GRCh38
rs555573641 Chr.19: 40206110-40206110 on Build GRCh38
rs755766456 Chr.19: 40206190-40206190 on Build GRCh38
rs747567246 Chr.19: 40206184-40206184 on Build GRCh38
rs749841833 Chr.19: 40205953-40205953 on Build GRCh38
rs757686251 Chr.19: 40206177-40206177 on Build GRCh38
rs768342095 Chr.19: 40205982-40205982 on Build GRCh38
rs772925511 Chr.19: 40206181-40206182 on Build GRCh38
rs892680139 Chr.19: 40206124-40206124 on Build GRCh38
rs771449066 Chr.19: 40205960-40205960 on Build GRCh38
rs781279884 Chr.19: 40205942-40205942 on Build GRCh38
rs758971847 Chr.19: 40206182-40206182 on Build GRCh38
rs778225660 Chr.19: 40206102-40206102 on Build GRCh38
rs750430245 Chr.19: 40205918-40205918 on Build GRCh38
rs192728745 Chr.19: 40206085-40206085 on Build GRCh38
rs376427920 Chr.19: 40206200-40206200 on Build GRCh38
rs746537447 Chr.19: 40205981-40205981 on Build GRCh38
rs756502610 Chr.19: 40205946-40205946 on Build GRCh38
rs553818989 Chr.19: 40206018-40206018 on Build GRCh38
rs775660074 Chr.19: 40206161-40206161 on Build GRCh38
rs952735749 Chr.19: 40205984-40205984 on Build GRCh38
rs746524729 Chr.19: 40206109-40206109 on Build GRCh38
rs985625892 Chr.19: 40206204-40206204 on Build GRCh38
rs761572394 Chr.19: 40205896-40205896 on Build GRCh38
rs760766805 Chr.19: 40206167-40206167 on Build GRCh38
rs916960455 Chr.19: 40206028-40206028 on Build GRCh38
rs904167606 Chr.19: 40206080-40206080 on Build GRCh38
rs202229714 Chr.19: 40206134-40206134 on Build GRCh38
rs757704923 Chr.19: 40206104-40206104 on Build GRCh38
rs145321643 Chr.19: 40206123-40206123 on Build GRCh38
rs747708612 Chr.19: 40206111-40206111 on Build GRCh38
rs755159756 Chr.19: 40205941-40205941 on Build GRCh38
rs766421744 Chr.19: 40206048-40206048 on Build GRCh38
rs557424009 Chr.19: 40206206-40206206 on Build GRCh38
rs762940502 Chr.19: 40206037-40206037 on Build GRCh38
rs568902554 Chr.19: 40205903-40205903 on Build GRCh38
rs946947439 Chr.19: 40206218-40206218 on Build GRCh38
rs1012667400 Chr.19: 40205973-40205973 on Build GRCh38
rs759588424 Chr.19: 40206074-40206074 on Build GRCh38
rs764315161 Chr.19: 40206168-40206168 on Build GRCh38
rs753088988 Chr.19: 40205943-40205943 on Build GRCh38
rs948401170 Chr.19: 40206036-40206036 on Build GRCh38
rs759628815 Chr.19: 40206133-40206133 on Build GRCh38
rs776429838 Chr.19: 40205895-40205895 on Build GRCh38
rs760679322 Chr.19: 40206207-40206207 on Build GRCh38
rs539418221 Chr.19: 40205928-40205928 on Build GRCh38
rs139191765 Chr.19: 40206132-40206132 on Build GRCh38

View Full Product Details