Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 82760868-82761450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCLO
Entrez Gene ID
27445
Gene Name
piccolo presynaptic cytomatrix protein
Gene Aliases
ACZ, PCH3
UniGene
Hs.12376
Gene Chromosome Location
Chr.7: 82754005-83164162 on Build GRCh38
Transcripts
NM_033026.5 XM_017012006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
86594 c.15125C>T p.S5042F Chr.7: 82761376-82761376 on Build GRCh38
3431787 c.15108T>C p.I5036I Chr.7: 82761393-82761393 on Build GRCh38
1092149 c.15113A>G p.Y5038C Chr.7: 82761388-82761388 on Build GRCh38
1092151 c.15091C>A p.L5031I Chr.7: 82761410-82761410 on Build GRCh38
369603 c.15101G>T p.R5034I Chr.7: 82761400-82761400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80229639 Chr.7: 82760892-82760892 on Build GRCh38
rs933084628 Chr.7: 82760933-82760933 on Build GRCh38
rs768271097 Chr.7: 82761337-82761337 on Build GRCh38
rs1056868193 Chr.7: 82761098-82761098 on Build GRCh38
rs760089997 Chr.7: 82761418-82761418 on Build GRCh38
rs1021177864 Chr.7: 82761118-82761118 on Build GRCh38
rs866397394 Chr.7: 82761374-82761374 on Build GRCh38
rs749413796 Chr.7: 82761322-82761322 on Build GRCh38
rs747790550 Chr.7: 82761368-82761368 on Build GRCh38
rs540609087 Chr.7: 82760904-82760904 on Build GRCh38
rs762328780 Chr.7: 82761391-82761391 on Build GRCh38
rs35172911 Chr.7: 82761150-82761150 on Build GRCh38
rs73708923 Chr.7: 82761043-82761043 on Build GRCh38
rs565170513 Chr.7: 82761224-82761224 on Build GRCh38
rs964303583 Chr.7: 82761310-82761310 on Build GRCh38
rs748483835 Chr.7: 82760924-82760924 on Build GRCh38
rs753113921 Chr.7: 82761422-82761422 on Build GRCh38
rs1000017813 Chr.7: 82761149-82761149 on Build GRCh38
rs757392914 Chr.7: 82761330-82761330 on Build GRCh38
rs908382237 Chr.7: 82760950-82760950 on Build GRCh38
rs955036718 Chr.7: 82761268-82761268 on Build GRCh38
rs756708677 Chr.7: 82761083-82761083 on Build GRCh38
rs758132529 Chr.7: 82761175-82761175 on Build GRCh38
rs941148079 Chr.7: 82761004-82761004 on Build GRCh38
rs941201717 Chr.7: 82761349-82761349 on Build GRCh38
rs143051243 Chr.7: 82761166-82761166 on Build GRCh38
rs1018526554 Chr.7: 82761228-82761228 on Build GRCh38
rs767860682 Chr.7: 82761419-82761419 on Build GRCh38
rs192183370 Chr.7: 82761111-82761111 on Build GRCh38
rs1038485744 Chr.7: 82761027-82761027 on Build GRCh38
rs546430532 Chr.7: 82761237-82761237 on Build GRCh38
rs922268794 Chr.7: 82761245-82761245 on Build GRCh38
rs769619257 Chr.7: 82761378-82761378 on Build GRCh38
rs768191701 Chr.7: 82761328-82761328 on Build GRCh38
rs111787752 Chr.7: 82760948-82760948 on Build GRCh38
rs764868320 Chr.7: 82761119-82761119 on Build GRCh38
rs770495441 Chr.7: 82761392-82761392 on Build GRCh38
rs1011105633 Chr.7: 82761180-82761180 on Build GRCh38
rs866731030 Chr.7: 82761231-82761231 on Build GRCh38
rs371116944 Chr.7: 82760943-82760943 on Build GRCh38
rs774660642 Chr.7: 82761405-82761405 on Build GRCh38
rs866623269 Chr.7: 82761371-82761371 on Build GRCh38
rs552896681 Chr.7: 82760894-82760894 on Build GRCh38
rs975150615 Chr.7: 82761195-82761195 on Build GRCh38
rs906592771 Chr.7: 82761132-82761132 on Build GRCh38
rs1012188789 Chr.7: 82761116-82761116 on Build GRCh38
rs1032757165 Chr.7: 82761151-82761151 on Build GRCh38
rs767222339 Chr.7: 82760944-82760954 on Build GRCh38
rs941901733 Chr.7: 82761091-82761091 on Build GRCh38
rs560598695 Chr.7: 82760979-82760979 on Build GRCh38
rs933903849 Chr.7: 82760936-82760936 on Build GRCh38
rs371927465 Chr.7: 82761346-82761346 on Build GRCh38
rs886279775 Chr.7: 82761082-82761082 on Build GRCh38
rs71093398 Chr.7: 82760970-82760971 on Build GRCh38
rs138354320 Chr.7: 82761097-82761097 on Build GRCh38
rs772816654 Chr.7: 82761390-82761390 on Build GRCh38
rs983120740 Chr.7: 82761288-82761288 on Build GRCh38
rs779809611 Chr.7: 82761332-82761332 on Build GRCh38
rs1046939454 Chr.7: 82760937-82760937 on Build GRCh38
rs896449872 Chr.7: 82761100-82761100 on Build GRCh38
rs958191511 Chr.7: 82761161-82761161 on Build GRCh38

View Full Product Details