Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 8798702-8799245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9A
Entrez Gene ID
171482
Gene Name
family with sequence similarity 9 member A
Gene Aliases
TEX39A
UniGene
Hs.382062
Gene Chromosome Location
Chr.X: 8790795-8801383 on Build GRCh38
Transcripts
NM_001171186.1 NM_174951.3

SNP Detail

SNP ID Chromosome Location
rs751592118 Chr.X: 8799048-8799048 on Build GRCh38
rs868526097 Chr.X: 8798791-8798791 on Build GRCh38
rs772809973 Chr.X: 8799099-8799099 on Build GRCh38
rs984730950 Chr.X: 8798768-8798770 on Build GRCh38
rs41311477 Chr.X: 8798925-8798925 on Build GRCh38
rs759890136 Chr.X: 8799107-8799108 on Build GRCh38
rs866499738 Chr.X: 8799060-8799060 on Build GRCh38
rs370041103 Chr.X: 8799001-8799001 on Build GRCh38
rs753770002 Chr.X: 8799175-8799175 on Build GRCh38
rs747588079 Chr.X: 8799097-8799097 on Build GRCh38
rs58250037 Chr.X: 8798902-8798902 on Build GRCh38
rs766883749 Chr.X: 8798919-8798919 on Build GRCh38
rs910131521 Chr.X: 8798780-8798780 on Build GRCh38
rs984286118 Chr.X: 8799215-8799215 on Build GRCh38
rs202152195 Chr.X: 8798928-8798928 on Build GRCh38
rs138951356 Chr.X: 8798977-8798977 on Build GRCh38
rs778945670 Chr.X: 8799070-8799070 on Build GRCh38
rs147452548 Chr.X: 8798987-8798987 on Build GRCh38
rs192671574 Chr.X: 8799186-8799186 on Build GRCh38
rs746088694 Chr.X: 8799072-8799072 on Build GRCh38
rs932989155 Chr.X: 8798897-8798897 on Build GRCh38
rs148243105 Chr.X: 8798900-8798900 on Build GRCh38
rs200757954 Chr.X: 8798991-8798991 on Build GRCh38
rs765782877 Chr.X: 8798979-8798979 on Build GRCh38
rs139267531 Chr.X: 8799012-8799012 on Build GRCh38
rs754948649 Chr.X: 8799050-8799050 on Build GRCh38
rs918107861 Chr.X: 8799220-8799220 on Build GRCh38
rs35340970 Chr.X: 8799052-8799053 on Build GRCh38
rs182946669 Chr.X: 8798873-8798873 on Build GRCh38
rs766195253 Chr.X: 8799028-8799028 on Build GRCh38
rs777089543 Chr.X: 8799106-8799115 on Build GRCh38
rs199558560 Chr.X: 8798992-8798992 on Build GRCh38
rs373467378 Chr.X: 8799088-8799088 on Build GRCh38
rs747563355 Chr.X: 8799077-8799077 on Build GRCh38
rs73631413 Chr.X: 8799180-8799180 on Build GRCh38
rs780435446 Chr.X: 8799075-8799075 on Build GRCh38
rs138064338 Chr.X: 8798983-8798983 on Build GRCh38
rs762640291 Chr.X: 8799117-8799117 on Build GRCh38
rs751532280 Chr.X: 8798917-8798917 on Build GRCh38
rs111647877 Chr.X: 8798837-8798837 on Build GRCh38
rs778022650 Chr.X: 8799055-8799055 on Build GRCh38
rs779785214 Chr.X: 8798916-8798916 on Build GRCh38
rs376530494 Chr.X: 8798982-8798982 on Build GRCh38
rs779368127 Chr.X: 8798944-8798944 on Build GRCh38
rs867887029 Chr.X: 8799162-8799162 on Build GRCh38
rs1036016308 Chr.X: 8798842-8798842 on Build GRCh38
rs200288519 Chr.X: 8798920-8798920 on Build GRCh38
rs758754370 Chr.X: 8799073-8799073 on Build GRCh38
rs769099799 Chr.X: 8799078-8799078 on Build GRCh38
rs202040210 Chr.X: 8799124-8799124 on Build GRCh38
rs757507611 Chr.X: 8799069-8799069 on Build GRCh38
rs753929157 Chr.X: 8799059-8799059 on Build GRCh38

View Full Product Details