Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116342047-116342610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_011539771.2 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246568 c.352G>A p.D118N Chr.10: 116342105-116342105 on Build GRCh38
1675593 c.376G>A p.E126K Chr.10: 116342129-116342129 on Build GRCh38
6030521 c.402G>T p.K134N Chr.10: 116342155-116342155 on Build GRCh38
7078564 c.387G>T p.M129I Chr.10: 116342140-116342140 on Build GRCh38
1474382 c.353delA p.D118fs*4 Chr.10: 116342106-116342106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78334481 Chr.10: 116342206-116342206 on Build GRCh38
rs904180613 Chr.10: 116342582-116342582 on Build GRCh38
rs377464128 Chr.10: 116342345-116342345 on Build GRCh38
rs760263051 Chr.10: 116342238-116342238 on Build GRCh38
rs778200621 Chr.10: 116342166-116342166 on Build GRCh38
rs892695600 Chr.10: 116342570-116342570 on Build GRCh38
rs754378767 Chr.10: 116342131-116342131 on Build GRCh38
rs748213847 Chr.10: 116342139-116342139 on Build GRCh38
rs939240373 Chr.10: 116342280-116342280 on Build GRCh38
rs939499430 Chr.10: 116342499-116342499 on Build GRCh38
rs953519650 Chr.10: 116342553-116342553 on Build GRCh38
rs767008059 Chr.10: 116342231-116342231 on Build GRCh38
rs534403525 Chr.10: 116342452-116342452 on Build GRCh38
rs770056121 Chr.10: 116342505-116342505 on Build GRCh38
rs776507814 Chr.10: 116342456-116342456 on Build GRCh38
rs763255969 Chr.10: 116342127-116342127 on Build GRCh38
rs191620202 Chr.10: 116342437-116342437 on Build GRCh38
rs765328815 Chr.10: 116342112-116342112 on Build GRCh38
rs894963011 Chr.10: 116342341-116342341 on Build GRCh38
rs769720190 Chr.10: 116342190-116342190 on Build GRCh38
rs182338368 Chr.10: 116342251-116342251 on Build GRCh38
rs987713121 Chr.10: 116342296-116342296 on Build GRCh38
rs1057045066 Chr.10: 116342549-116342549 on Build GRCh38
rs35617064 Chr.10: 116342416-116342417 on Build GRCh38
rs755387650 Chr.10: 116342132-116342132 on Build GRCh38
rs777847656 Chr.10: 116342395-116342395 on Build GRCh38
rs201698300 Chr.10: 116342107-116342107 on Build GRCh38
rs370356140 Chr.10: 116342446-116342446 on Build GRCh38
rs1056334691 Chr.10: 116342310-116342310 on Build GRCh38
rs756538004 Chr.10: 116342097-116342097 on Build GRCh38
rs187448259 Chr.10: 116342358-116342358 on Build GRCh38
rs968397006 Chr.10: 116342229-116342229 on Build GRCh38
rs953236780 Chr.10: 116342289-116342289 on Build GRCh38
rs1056677266 Chr.10: 116342141-116342141 on Build GRCh38
rs750061944 Chr.10: 116342095-116342095 on Build GRCh38
rs774315344 Chr.10: 116342211-116342211 on Build GRCh38
rs569527828 Chr.10: 116342278-116342278 on Build GRCh38
rs1005060410 Chr.10: 116342417-116342417 on Build GRCh38
rs759582063 Chr.10: 116342073-116342074 on Build GRCh38
rs868571860 Chr.10: 116342111-116342111 on Build GRCh38
rs997750449 Chr.10: 116342497-116342497 on Build GRCh38
rs111749880 Chr.10: 116342382-116342382 on Build GRCh38
rs532636333 Chr.10: 116342102-116342102 on Build GRCh38
rs892715952 Chr.10: 116342170-116342170 on Build GRCh38
rs779078810 Chr.10: 116342135-116342135 on Build GRCh38
rs767204397 Chr.10: 116342085-116342085 on Build GRCh38
rs78019944 Chr.10: 116342159-116342159 on Build GRCh38
rs750080200 Chr.10: 116342233-116342233 on Build GRCh38
rs772259916 Chr.10: 116342165-116342165 on Build GRCh38
rs1048891197 Chr.10: 116342363-116342363 on Build GRCh38
rs144199930 Chr.10: 116342579-116342579 on Build GRCh38
rs764161004 Chr.10: 116342144-116342145 on Build GRCh38
rs759633238 Chr.10: 116342486-116342486 on Build GRCh38
rs752679290 Chr.10: 116342116-116342118 on Build GRCh38
rs775090202 Chr.10: 116342191-116342191 on Build GRCh38
rs201128510 Chr.10: 116342565-116342566 on Build GRCh38
rs907214964 Chr.10: 116342255-116342255 on Build GRCh38
rs141489077 Chr.10: 116342188-116342188 on Build GRCh38
rs751401701 Chr.10: 116342248-116342249 on Build GRCh38
rs972016942 Chr.10: 116342384-116342384 on Build GRCh38
rs17094567 Chr.10: 116342574-116342574 on Build GRCh38
rs762970200 Chr.10: 116342200-116342200 on Build GRCh38
rs569901035 Chr.10: 116342362-116342362 on Build GRCh38
rs17094564 Chr.10: 116342399-116342399 on Build GRCh38
rs570498292 Chr.10: 116342444-116342444 on Build GRCh38
rs1028782146 Chr.10: 116342283-116342283 on Build GRCh38
rs552607892 Chr.10: 116342466-116342466 on Build GRCh38
rs761369245 Chr.10: 116342224-116342224 on Build GRCh38

View Full Product Details