Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41841736-41842234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL10
Entrez Gene ID
317719
Gene Name
kelch like family member 10
Gene Aliases
SPGF11
UniGene
Hs.127510
Gene Chromosome Location
Chr.17: 41835685-41848347 on Build GRCh38
Transcripts
NM_152467.3 XM_011524706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223876 c.604G>A p.A202T Chr.17: 41842232-41842232 on Build GRCh38
4066459 c.319G>A p.D107N Chr.17: 41841947-41841947 on Build GRCh38
4066458 c.318G>A p.P106P Chr.17: 41841946-41841946 on Build GRCh38
5852522 c.355delT p.F119fs*38 Chr.17: 41841983-41841983 on Build GRCh38
148273 c.571G>A p.E191K Chr.17: 41842199-41842199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782284363 Chr.17: 41842155-41842155 on Build GRCh38
rs782026183 Chr.17: 41842048-41842048 on Build GRCh38
rs781834674 Chr.17: 41841933-41841933 on Build GRCh38
rs1529933 Chr.17: 41841889-41841889 on Build GRCh38
rs782395311 Chr.17: 41842124-41842124 on Build GRCh38
rs782469970 Chr.17: 41842082-41842082 on Build GRCh38
rs1013257036 Chr.17: 41841853-41841853 on Build GRCh38
rs782750953 Chr.17: 41841809-41841809 on Build GRCh38
rs768950250 Chr.17: 41841879-41841879 on Build GRCh38
rs536242171 Chr.17: 41841919-41841919 on Build GRCh38
rs781868754 Chr.17: 41842071-41842071 on Build GRCh38
rs201335504 Chr.17: 41842016-41842016 on Build GRCh38
rs202162199 Chr.17: 41841876-41841876 on Build GRCh38
rs36065902 Chr.17: 41841870-41841870 on Build GRCh38
rs782455157 Chr.17: 41841802-41841802 on Build GRCh38
rs782327717 Chr.17: 41841883-41841883 on Build GRCh38
rs973869604 Chr.17: 41842114-41842114 on Build GRCh38
rs782132199 Chr.17: 41842001-41842001 on Build GRCh38
rs781932658 Chr.17: 41841763-41841763 on Build GRCh38
rs369099688 Chr.17: 41842075-41842075 on Build GRCh38
rs377742054 Chr.17: 41841865-41841865 on Build GRCh38
rs782336501 Chr.17: 41841864-41841864 on Build GRCh38
rs537646033 Chr.17: 41841840-41841840 on Build GRCh38
rs202110308 Chr.17: 41842111-41842111 on Build GRCh38
rs782586458 Chr.17: 41841995-41841995 on Build GRCh38
rs782254201 Chr.17: 41842107-41842107 on Build GRCh38
rs375880722 Chr.17: 41841907-41841907 on Build GRCh38
rs374071801 Chr.17: 41841890-41841890 on Build GRCh38
rs781808591 Chr.17: 41842080-41842080 on Build GRCh38
rs781866908 Chr.17: 41841911-41841911 on Build GRCh38
rs782292158 Chr.17: 41841951-41841951 on Build GRCh38
rs781792062 Chr.17: 41841801-41841801 on Build GRCh38
rs782759956 Chr.17: 41841908-41841908 on Build GRCh38
rs1021250962 Chr.17: 41841945-41841945 on Build GRCh38
rs782221918 Chr.17: 41841998-41841998 on Build GRCh38
rs782751925 Chr.17: 41842066-41842066 on Build GRCh38
rs544035963 Chr.17: 41842000-41842000 on Build GRCh38
rs574533496 Chr.17: 41842174-41842174 on Build GRCh38
rs929428278 Chr.17: 41842153-41842153 on Build GRCh38
rs782707612 Chr.17: 41842055-41842055 on Build GRCh38
rs782045864 Chr.17: 41842054-41842054 on Build GRCh38
rs34340836 Chr.17: 41841791-41841792 on Build GRCh38
rs541818300 Chr.17: 41842193-41842193 on Build GRCh38
rs782311644 Chr.17: 41841954-41841954 on Build GRCh38
rs1048016944 Chr.17: 41842168-41842168 on Build GRCh38
rs541192046 Chr.17: 41842072-41842072 on Build GRCh38
rs782646029 Chr.17: 41841847-41841847 on Build GRCh38
rs200286521 Chr.17: 41842137-41842137 on Build GRCh38
rs549517893 Chr.17: 41841862-41841862 on Build GRCh38
rs782038542 Chr.17: 41841881-41841881 on Build GRCh38
rs782099047 Chr.17: 41842062-41842062 on Build GRCh38
rs782138226 Chr.17: 41841916-41841916 on Build GRCh38
rs376803007 Chr.17: 41841851-41841851 on Build GRCh38
rs200355741 Chr.17: 41842144-41842144 on Build GRCh38
rs71373458 Chr.17: 41841793-41841793 on Build GRCh38
rs782517337 Chr.17: 41841845-41841845 on Build GRCh38
rs943436389 Chr.17: 41842086-41842086 on Build GRCh38
rs782194151 Chr.17: 41841863-41841863 on Build GRCh38
rs782820025 Chr.17: 41842197-41842197 on Build GRCh38
rs782600399 Chr.17: 41841932-41841932 on Build GRCh38
rs782079064 Chr.17: 41841771-41841771 on Build GRCh38
rs370786117 Chr.17: 41841946-41841946 on Build GRCh38
rs781822818 Chr.17: 41842061-41842061 on Build GRCh38
rs781926688 Chr.17: 41842184-41842184 on Build GRCh38
rs782626896 Chr.17: 41842106-41842106 on Build GRCh38
rs782307083 Chr.17: 41841762-41841762 on Build GRCh38
rs782065666 Chr.17: 41841808-41841809 on Build GRCh38
rs782215386 Chr.17: 41842188-41842188 on Build GRCh38
rs115928775 Chr.17: 41841882-41841882 on Build GRCh38
rs781862065 Chr.17: 41841794-41841794 on Build GRCh38
rs192564496 Chr.17: 41842003-41842003 on Build GRCh38
rs369676198 Chr.17: 41841858-41841858 on Build GRCh38

View Full Product Details