Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32038474-32039000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP21A2
Entrez Gene ID
1589
Gene Name
cytochrome P450 family 21 subfamily A member 2
Gene Aliases
CA21H, CAH1, CPS1, CYP21, CYP21B, P450c21B
UniGene
Hs.654479
Gene Chromosome Location
Chr.6: 32038316-32041670 on Build GRCh38
Transcripts
NM_000500.7 NM_001128590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117644 c.84C>T p.L28L Chr.6: 32038506-32038506 on Build GRCh38
6639055 c.83T>G p.L28R Chr.6: 32038505-32038505 on Build GRCh38
483890 c.230C>A p.T77N Chr.6: 32038749-32038749 on Build GRCh38
4160892 c.138C>A p.P46P Chr.6: 32038560-32038560 on Build GRCh38
6503017 c.60G>A p.W20* Chr.6: 32038482-32038482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6468 Chr.6: 32038540-32038540 on Build GRCh38
rs566306310 Chr.6: 32038565-32038565 on Build GRCh38
rs746666047 Chr.6: 32038848-32038848 on Build GRCh38
rs1005488948 Chr.6: 32038856-32038856 on Build GRCh38
rs6464 Chr.6: 32038560-32038560 on Build GRCh38
rs755085518 Chr.6: 32038811-32038811 on Build GRCh38
rs762507423 Chr.6: 32038552-32038552 on Build GRCh38
rs1038709622 Chr.6: 32038800-32038800 on Build GRCh38
rs754089005 Chr.6: 32038862-32038862 on Build GRCh38
rs745712520 Chr.6: 32038821-32038821 on Build GRCh38
rs966552373 Chr.6: 32038970-32038970 on Build GRCh38
rs780650087 Chr.6: 32038831-32038834 on Build GRCh38
rs530546690 Chr.6: 32038911-32038911 on Build GRCh38
rs533057482 Chr.6: 32038887-32038887 on Build GRCh38
rs6449 Chr.6: 32038878-32038878 on Build GRCh38
rs746857236 Chr.6: 32038503-32038503 on Build GRCh38
rs762440952 Chr.6: 32038717-32038717 on Build GRCh38
rs771942449 Chr.6: 32038499-32038499 on Build GRCh38
rs755070239 Chr.6: 32038640-32038640 on Build GRCh38
rs777229786 Chr.6: 32038843-32038844 on Build GRCh38
rs185054992 Chr.6: 32038971-32038971 on Build GRCh38
rs770843305 Chr.6: 32038507-32038507 on Build GRCh38
rs917718695 Chr.6: 32038684-32038684 on Build GRCh38
rs779144910 Chr.6: 32038812-32038812 on Build GRCh38
rs886038207 Chr.6: 32038580-32038581 on Build GRCh38
rs774906217 Chr.6: 32038551-32038551 on Build GRCh38
rs757696541 Chr.6: 32038563-32038563 on Build GRCh38
rs2075561 Chr.6: 32038859-32038859 on Build GRCh38
rs578064226 Chr.6: 32038677-32038677 on Build GRCh38
rs773754525 Chr.6: 32038855-32038855 on Build GRCh38
rs555746722 Chr.6: 32038648-32038648 on Build GRCh38
rs534097844 Chr.6: 32038641-32038641 on Build GRCh38
rs567558109 Chr.6: 32038650-32038650 on Build GRCh38
rs755425808 Chr.6: 32038840-32038841 on Build GRCh38
rs745647843 Chr.6: 32038694-32038694 on Build GRCh38
rs749608963 Chr.6: 32038835-32038835 on Build GRCh38
rs548494893 Chr.6: 32038895-32038895 on Build GRCh38
rs773259857 Chr.6: 32038501-32038501 on Build GRCh38
rs746945052 Chr.6: 32038664-32038664 on Build GRCh38
rs184177366 Chr.6: 32038848-32038848 on Build GRCh38
rs756665358 Chr.6: 32038828-32038828 on Build GRCh38
rs749346869 Chr.6: 32038498-32038498 on Build GRCh38
rs769629885 Chr.6: 32038837-32038837 on Build GRCh38
rs775197696 Chr.6: 32038701-32038701 on Build GRCh38
rs551374557 Chr.6: 32038530-32038530 on Build GRCh38
rs538722250 Chr.6: 32038695-32038695 on Build GRCh38
rs202079601 Chr.6: 32038527-32038527 on Build GRCh38
rs767973196 Chr.6: 32038781-32038781 on Build GRCh38
rs756320878 Chr.6: 32038590-32038590 on Build GRCh38
rs567593813 Chr.6: 32038968-32038968 on Build GRCh38
rs757288233 Chr.6: 32038816-32038816 on Build GRCh38
rs750793252 Chr.6: 32038782-32038782 on Build GRCh38
rs6448 Chr.6: 32038826-32038826 on Build GRCh38
rs761010614 Chr.6: 32038857-32038857 on Build GRCh38
rs6462 Chr.6: 32038820-32038820 on Build GRCh38
rs983698823 Chr.6: 32038569-32038569 on Build GRCh38
rs538071914 Chr.6: 32038668-32038668 on Build GRCh38
rs770382355 Chr.6: 32038847-32038848 on Build GRCh38
rs879281716 Chr.6: 32038518-32038518 on Build GRCh38
rs6463 Chr.6: 32038844-32038844 on Build GRCh38
rs750737459 Chr.6: 32038578-32038578 on Build GRCh38
rs779650227 Chr.6: 32038838-32038838 on Build GRCh38
rs192217993 Chr.6: 32038938-32038938 on Build GRCh38
rs576031977 Chr.6: 32038661-32038662 on Build GRCh38
rs752771213 Chr.6: 32038814-32038814 on Build GRCh38
rs761079139 Chr.6: 32038806-32038806 on Build GRCh38
rs938813750 Chr.6: 32038663-32038663 on Build GRCh38
rs9378251 Chr.6: 32038514-32038514 on Build GRCh38
rs563409717 Chr.6: 32038903-32038903 on Build GRCh38
rs543633765 Chr.6: 32038867-32038867 on Build GRCh38
rs368330593 Chr.6: 32038746-32038746 on Build GRCh38
rs750910646 Chr.6: 32038779-32038780 on Build GRCh38
rs767736158 Chr.6: 32038662-32038662 on Build GRCh38
rs11757034 Chr.6: 32038945-32038945 on Build GRCh38
rs766566786 Chr.6: 32038810-32038810 on Build GRCh38
rs749374064 Chr.6: 32038621-32038621 on Build GRCh38
rs6452 Chr.6: 32038676-32038676 on Build GRCh38
rs766767764 Chr.6: 32038860-32038860 on Build GRCh38
rs188515168 Chr.6: 32038927-32038927 on Build GRCh38
rs764569922 Chr.6: 32038553-32038553 on Build GRCh38
rs200648381 Chr.6: 32038526-32038526 on Build GRCh38
rs560190536 Chr.6: 32038939-32038939 on Build GRCh38
rs548984508 Chr.6: 32038959-32038959 on Build GRCh38
rs763599355 Chr.6: 32038727-32038727 on Build GRCh38
rs9378252 Chr.6: 32038610-32038610 on Build GRCh38
rs1030467767 Chr.6: 32038538-32038538 on Build GRCh38
rs113752787 Chr.6: 32038829-32038829 on Build GRCh38
rs561284668 Chr.6: 32038847-32038847 on Build GRCh38

View Full Product Details