Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149011754-149012249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDIA4
Entrez Gene ID
9601
Gene Name
protein disulfide isomerase family A member 4
Gene Aliases
ERP70, ERP72, ERp-72
UniGene
Hs.93659
Gene Chromosome Location
Chr.7: 149003054-149028690 on Build GRCh38
Transcripts
NM_004911.4 XM_006716185.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3785032 c.775C>T p.R259C Chr.7: 149012200-149012200 on Build GRCh38
1235210 c.844C>T p.Q282* Chr.7: 149011981-149011981 on Build GRCh38
6109189 c.884A>T p.Q295L Chr.7: 149011941-149011941 on Build GRCh38
4388677 c.802G>A p.G268S Chr.7: 149012173-149012173 on Build GRCh38
3259734 c.825C>T p.I275I Chr.7: 149012000-149012000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143550335 Chr.7: 149011885-149011885 on Build GRCh38
rs748473253 Chr.7: 149011897-149011897 on Build GRCh38
rs781689302 Chr.7: 149012149-149012149 on Build GRCh38
rs766921558 Chr.7: 149011815-149011815 on Build GRCh38
rs533334551 Chr.7: 149011901-149011901 on Build GRCh38
rs140131927 Chr.7: 149011846-149011846 on Build GRCh38
rs370147870 Chr.7: 149011824-149011824 on Build GRCh38
rs534122693 Chr.7: 149011976-149011976 on Build GRCh38
rs762583788 Chr.7: 149012166-149012166 on Build GRCh38
rs145092459 Chr.7: 149011898-149011898 on Build GRCh38
rs201131273 Chr.7: 149011810-149011810 on Build GRCh38
rs754657819 Chr.7: 149011978-149011978 on Build GRCh38
rs752950039 Chr.7: 149012196-149012196 on Build GRCh38
rs984268183 Chr.7: 149012226-149012226 on Build GRCh38
rs1054297876 Chr.7: 149012137-149012137 on Build GRCh38
rs756102159 Chr.7: 149012079-149012080 on Build GRCh38
rs555842203 Chr.7: 149012014-149012014 on Build GRCh38
rs372415891 Chr.7: 149011928-149011928 on Build GRCh38
rs138974987 Chr.7: 149011949-149011949 on Build GRCh38
rs1000685174 Chr.7: 149012134-149012134 on Build GRCh38
rs761711328 Chr.7: 149012044-149012044 on Build GRCh38
rs769339244 Chr.7: 149012156-149012156 on Build GRCh38
rs764047250 Chr.7: 149011960-149011960 on Build GRCh38
rs545639058 Chr.7: 149011989-149011989 on Build GRCh38
rs201276356 Chr.7: 149011971-149011971 on Build GRCh38
rs375265274 Chr.7: 149012136-149012136 on Build GRCh38
rs773962719 Chr.7: 149012170-149012170 on Build GRCh38
rs61739279 Chr.7: 149011985-149011985 on Build GRCh38
rs141864886 Chr.7: 149012157-149012157 on Build GRCh38
rs745923795 Chr.7: 149011877-149011877 on Build GRCh38
rs866765414 Chr.7: 149011834-149011834 on Build GRCh38
rs961825633 Chr.7: 149012218-149012218 on Build GRCh38
rs180887981 Chr.7: 149011839-149011839 on Build GRCh38
rs758147565 Chr.7: 149012130-149012130 on Build GRCh38
rs769187636 Chr.7: 149011800-149011800 on Build GRCh38
rs112085881 Chr.7: 149011843-149011843 on Build GRCh38
rs991707916 Chr.7: 149011886-149011886 on Build GRCh38
rs200863165 Chr.7: 149012145-149012145 on Build GRCh38
rs774963960 Chr.7: 149011809-149011809 on Build GRCh38
rs920451617 Chr.7: 149012225-149012225 on Build GRCh38
rs971425159 Chr.7: 149012152-149012152 on Build GRCh38
rs769134342 Chr.7: 149011888-149011888 on Build GRCh38
rs374768570 Chr.7: 149012194-149012194 on Build GRCh38
rs755138821 Chr.7: 149012105-149012105 on Build GRCh38
rs1008947480 Chr.7: 149011792-149011792 on Build GRCh38
rs370665535 Chr.7: 149012037-149012037 on Build GRCh38
rs764379611 Chr.7: 149012222-149012222 on Build GRCh38
rs150481375 Chr.7: 149011847-149011847 on Build GRCh38
rs749625017 Chr.7: 149011799-149011799 on Build GRCh38
rs971628686 Chr.7: 149012189-149012189 on Build GRCh38
rs759245452 Chr.7: 149011920-149011920 on Build GRCh38
rs745330359 Chr.7: 149012150-149012150 on Build GRCh38
rs762690248 Chr.7: 149011932-149011932 on Build GRCh38
rs150793763 Chr.7: 149012174-149012174 on Build GRCh38
rs1041344635 Chr.7: 149011811-149011811 on Build GRCh38
rs770414968 Chr.7: 149012000-149012000 on Build GRCh38
rs201201125 Chr.7: 149011987-149011987 on Build GRCh38
rs541393681 Chr.7: 149011915-149011915 on Build GRCh38
rs747414354 Chr.7: 149012146-149012146 on Build GRCh38
rs779279777 Chr.7: 149011984-149011984 on Build GRCh38
rs760182178 Chr.7: 149011904-149011904 on Build GRCh38
rs945929299 Chr.7: 149012075-149012075 on Build GRCh38
rs111407630 Chr.7: 149011845-149011845 on Build GRCh38
rs1020299613 Chr.7: 149011793-149011793 on Build GRCh38
rs754736434 Chr.7: 149011826-149011826 on Build GRCh38
rs371382515 Chr.7: 149012167-149012167 on Build GRCh38
rs973527006 Chr.7: 149012224-149012224 on Build GRCh38
rs367839477 Chr.7: 149012199-149012199 on Build GRCh38
rs867534782 Chr.7: 149011786-149011786 on Build GRCh38
rs773306098 Chr.7: 149012179-149012179 on Build GRCh38
rs149365261 Chr.7: 149011975-149011975 on Build GRCh38
rs762789882 Chr.7: 149012022-149012022 on Build GRCh38
rs931924104 Chr.7: 149012013-149012013 on Build GRCh38
rs779226690 Chr.7: 149012127-149012127 on Build GRCh38
rs79600968 Chr.7: 149012026-149012026 on Build GRCh38
rs775030714 Chr.7: 149012021-149012021 on Build GRCh38
rs767572743 Chr.7: 149012048-149012048 on Build GRCh38
rs867465730 Chr.7: 149011954-149011954 on Build GRCh38
rs142943511 Chr.7: 149011889-149011889 on Build GRCh38
rs570306170 Chr.7: 149012051-149012051 on Build GRCh38
rs1020227826 Chr.7: 149012058-149012058 on Build GRCh38
rs112506294 Chr.7: 149012006-149012006 on Build GRCh38
rs199605420 Chr.7: 149011913-149011913 on Build GRCh38
rs148084821 Chr.7: 149011999-149011999 on Build GRCh38
rs751505200 Chr.7: 149011962-149011962 on Build GRCh38
rs753510843 Chr.7: 149011825-149011825 on Build GRCh38
rs375710204 Chr.7: 149011798-149011798 on Build GRCh38
rs772718911 Chr.7: 149011823-149011823 on Build GRCh38
rs978981845 Chr.7: 149011835-149011835 on Build GRCh38
rs771363149 Chr.7: 149012148-149012148 on Build GRCh38
rs371332924 Chr.7: 149012001-149012001 on Build GRCh38
rs372962795 Chr.7: 149012010-149012010 on Build GRCh38
rs759666998 Chr.7: 149012190-149012190 on Build GRCh38
rs369248381 Chr.7: 149012178-149012178 on Build GRCh38
rs567343075 Chr.7: 149011873-149011873 on Build GRCh38
rs370450689 Chr.7: 149011973-149011973 on Build GRCh38

View Full Product Details