Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31031938-31032445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PURG
Entrez Gene ID
29942
Gene Name
purine rich element binding protein G
Gene Aliases
PURG-A, PURG-B
UniGene
Hs.373778
Gene Chromosome Location
Chr.8: 30995802-31033715 on Build GRCh38
Transcripts
NM_001015508.2 NM_001323311.1 NM_001323312.1 NM_013357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874581 c.787G>A p.E263K Chr.8: 31031996-31031996 on Build GRCh38
1456661 c.628G>T p.G210C Chr.8: 31032155-31032155 on Build GRCh38
5559110 c.782T>G p.L261R Chr.8: 31032001-31032001 on Build GRCh38
3899633 c.341A>G p.E114G Chr.8: 31032442-31032442 on Build GRCh38
5987287 c.808G>C p.D270H Chr.8: 31031975-31031975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778050810 Chr.8: 31032154-31032154 on Build GRCh38
rs897817458 Chr.8: 31032312-31032312 on Build GRCh38
rs781213690 Chr.8: 31032329-31032329 on Build GRCh38
rs756150951 Chr.8: 31032303-31032303 on Build GRCh38
rs202052368 Chr.8: 31032328-31032328 on Build GRCh38
rs768016013 Chr.8: 31032387-31032387 on Build GRCh38
rs768782840 Chr.8: 31032327-31032327 on Build GRCh38
rs913565647 Chr.8: 31032051-31032051 on Build GRCh38
rs758157265 Chr.8: 31032324-31032324 on Build GRCh38
rs917800079 Chr.8: 31032252-31032252 on Build GRCh38
rs746789993 Chr.8: 31032325-31032325 on Build GRCh38
rs765564331 Chr.8: 31031965-31031965 on Build GRCh38
rs753140688 Chr.8: 31032388-31032388 on Build GRCh38
rs11574152 Chr.8: 31032255-31032255 on Build GRCh38
rs772864161 Chr.8: 31032249-31032249 on Build GRCh38
rs898861287 Chr.8: 31032371-31032371 on Build GRCh38
rs774753611 Chr.8: 31032378-31032378 on Build GRCh38
rs770025349 Chr.8: 31032014-31032014 on Build GRCh38
rs139474524 Chr.8: 31032130-31032130 on Build GRCh38
rs745662043 Chr.8: 31032025-31032025 on Build GRCh38
rs760083519 Chr.8: 31032276-31032276 on Build GRCh38
rs750780650 Chr.8: 31031971-31031971 on Build GRCh38
rs374671334 Chr.8: 31032179-31032179 on Build GRCh38
rs767997655 Chr.8: 31032289-31032289 on Build GRCh38
rs7464560 Chr.8: 31032356-31032356 on Build GRCh38
rs11574151 Chr.8: 31032061-31032061 on Build GRCh38
rs201816816 Chr.8: 31032123-31032123 on Build GRCh38
rs1019113783 Chr.8: 31032379-31032379 on Build GRCh38
rs978738630 Chr.8: 31032030-31032030 on Build GRCh38
rs745651220 Chr.8: 31032170-31032170 on Build GRCh38
rs776451796 Chr.8: 31032075-31032075 on Build GRCh38
rs761386018 Chr.8: 31032242-31032242 on Build GRCh38
rs113193871 Chr.8: 31032043-31032043 on Build GRCh38
rs746900091 Chr.8: 31032216-31032216 on Build GRCh38
rs200922325 Chr.8: 31032078-31032078 on Build GRCh38
rs766728816 Chr.8: 31032271-31032271 on Build GRCh38
rs926463720 Chr.8: 31032169-31032169 on Build GRCh38
rs754039229 Chr.8: 31032316-31032316 on Build GRCh38
rs146203905 Chr.8: 31032330-31032330 on Build GRCh38
rs937597846 Chr.8: 31032036-31032036 on Build GRCh38
rs77814337 Chr.8: 31032225-31032225 on Build GRCh38
rs145712973 Chr.8: 31032293-31032293 on Build GRCh38
rs565416842 Chr.8: 31032318-31032318 on Build GRCh38
rs750824000 Chr.8: 31032119-31032119 on Build GRCh38
rs954660855 Chr.8: 31032007-31032007 on Build GRCh38
rs764804801 Chr.8: 31032247-31032247 on Build GRCh38
rs756349318 Chr.8: 31032148-31032148 on Build GRCh38
rs780609493 Chr.8: 31031994-31031994 on Build GRCh38
rs752841792 Chr.8: 31032297-31032297 on Build GRCh38
rs138893308 Chr.8: 31032345-31032345 on Build GRCh38
rs559569366 Chr.8: 31032195-31032195 on Build GRCh38
rs371756390 Chr.8: 31032270-31032270 on Build GRCh38
rs768694963 Chr.8: 31032233-31032233 on Build GRCh38
rs201025652 Chr.8: 31032313-31032313 on Build GRCh38
rs773670793 Chr.8: 31032009-31032009 on Build GRCh38
rs532874218 Chr.8: 31032340-31032340 on Build GRCh38
rs752014345 Chr.8: 31032121-31032121 on Build GRCh38
rs781288340 Chr.8: 31032124-31032124 on Build GRCh38
rs762707981 Chr.8: 31032366-31032366 on Build GRCh38
rs369001501 Chr.8: 31032373-31032373 on Build GRCh38
rs369086930 Chr.8: 31032010-31032010 on Build GRCh38
rs763646588 Chr.8: 31032063-31032063 on Build GRCh38
rs751891314 Chr.8: 31032273-31032273 on Build GRCh38
rs375267177 Chr.8: 31032008-31032008 on Build GRCh38
rs771968873 Chr.8: 31032031-31032031 on Build GRCh38
rs772773032 Chr.8: 31032355-31032355 on Build GRCh38
rs758723248 Chr.8: 31032120-31032120 on Build GRCh38
rs886167426 Chr.8: 31032280-31032280 on Build GRCh38
rs776340615 Chr.8: 31032237-31032237 on Build GRCh38
rs1049120230 Chr.8: 31032339-31032339 on Build GRCh38
rs761609882 Chr.8: 31032116-31032116 on Build GRCh38
rs774060872 Chr.8: 31032017-31032017 on Build GRCh38
rs940468985 Chr.8: 31032306-31032306 on Build GRCh38
rs758890151 Chr.8: 31031974-31031974 on Build GRCh38
rs11574150 Chr.8: 31032012-31032012 on Build GRCh38

View Full Product Details