Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72198033-72198544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIN4
Entrez Gene ID
5303
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
EPVH, PAR14, PAR17
UniGene
Hs.655623
Gene Chromosome Location
Chr.X: 72181676-72263964 on Build GRCh38
Transcripts
NM_001170747.1 NM_006223.3 NR_033187.1

SNP Detail

SNP ID Chromosome Location
rs928352166 Chr.X: 72198398-72198398 on Build GRCh38
rs190749180 Chr.X: 72198080-72198080 on Build GRCh38
rs769207941 Chr.X: 72198470-72198470 on Build GRCh38
rs200890062 Chr.X: 72198457-72198457 on Build GRCh38
rs929186001 Chr.X: 72198235-72198235 on Build GRCh38
rs938463164 Chr.X: 72198413-72198413 on Build GRCh38
rs979124188 Chr.X: 72198083-72198083 on Build GRCh38
rs766845671 Chr.X: 72198447-72198448 on Build GRCh38
rs145531112 Chr.X: 72198344-72198344 on Build GRCh38
rs937926043 Chr.X: 72198177-72198177 on Build GRCh38
rs990813025 Chr.X: 72198271-72198271 on Build GRCh38
rs1056288083 Chr.X: 72198196-72198196 on Build GRCh38
rs767086808 Chr.X: 72198127-72198127 on Build GRCh38
rs752296762 Chr.X: 72198137-72198137 on Build GRCh38
rs754254826 Chr.X: 72198286-72198286 on Build GRCh38
rs917740392 Chr.X: 72198226-72198227 on Build GRCh38
rs977146066 Chr.X: 72198377-72198377 on Build GRCh38
rs750960486 Chr.X: 72198507-72198507 on Build GRCh38
rs997036324 Chr.X: 72198482-72198482 on Build GRCh38
rs991334309 Chr.X: 72198459-72198459 on Build GRCh38
rs775259083 Chr.X: 72198376-72198376 on Build GRCh38
rs1025700115 Chr.X: 72198129-72198129 on Build GRCh38
rs745343572 Chr.X: 72198207-72198207 on Build GRCh38
rs955477282 Chr.X: 72198155-72198155 on Build GRCh38
rs1008381055 Chr.X: 72198205-72198205 on Build GRCh38
rs182101403 Chr.X: 72198448-72198448 on Build GRCh38
rs760382674 Chr.X: 72198260-72198270 on Build GRCh38
rs769356424 Chr.X: 72198236-72198236 on Build GRCh38
rs763902424 Chr.X: 72198270-72198270 on Build GRCh38
rs762480222 Chr.X: 72198417-72198417 on Build GRCh38
rs926492764 Chr.X: 72198123-72198123 on Build GRCh38
rs1038384633 Chr.X: 72198433-72198433 on Build GRCh38

View Full Product Details