Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72206955-72207503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L
Entrez Gene ID
54821
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
Gene Aliases
PICH, RAD26L
UniGene
Hs.47558
Gene Chromosome Location
Chr.X: 72204657-72239008 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

Gene Symbol
PIN4
Entrez Gene ID
5303
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
EPVH, PAR14, PAR17
UniGene
Hs.655623
Gene Chromosome Location
Chr.X: 72181676-72263964 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

SNP Detail

SNP ID Chromosome Location
rs776347272 Chr.X: 72207270-72207270 on Build GRCh38
rs777793094 Chr.X: 72207056-72207056 on Build GRCh38
rs933735259 Chr.X: 72207292-72207292 on Build GRCh38
rs976090150 Chr.X: 72207354-72207354 on Build GRCh38
rs373424116 Chr.X: 72207192-72207192 on Build GRCh38
rs749715424 Chr.X: 72207196-72207196 on Build GRCh38
rs772789771 Chr.X: 72207222-72207222 on Build GRCh38
rs139531286 Chr.X: 72207273-72207273 on Build GRCh38
rs780243104 Chr.X: 72207459-72207459 on Build GRCh38
rs367844894 Chr.X: 72207406-72207406 on Build GRCh38
rs756526817 Chr.X: 72207438-72207438 on Build GRCh38
rs923238793 Chr.X: 72207382-72207382 on Build GRCh38
rs752769595 Chr.X: 72207033-72207033 on Build GRCh38
rs865790063 Chr.X: 72207463-72207463 on Build GRCh38
rs770702617 Chr.X: 72207256-72207256 on Build GRCh38
rs765128241 Chr.X: 72207287-72207287 on Build GRCh38
rs758443175 Chr.X: 72207038-72207038 on Build GRCh38
rs928138626 Chr.X: 72207102-72207102 on Build GRCh38
rs146254781 Chr.X: 72207378-72207378 on Build GRCh38
rs1037136790 Chr.X: 72207249-72207249 on Build GRCh38
rs757416133 Chr.X: 72207109-72207109 on Build GRCh38
rs752700494 Chr.X: 72207312-72207312 on Build GRCh38
rs989027346 Chr.X: 72207429-72207429 on Build GRCh38
rs751805250 Chr.X: 72207087-72207087 on Build GRCh38
rs750805759 Chr.X: 72207422-72207422 on Build GRCh38
rs150532018 Chr.X: 72207206-72207206 on Build GRCh38
rs746226184 Chr.X: 72207146-72207146 on Build GRCh38
rs376450208 Chr.X: 72207177-72207177 on Build GRCh38
rs370873689 Chr.X: 72207070-72207070 on Build GRCh38
rs1057227288 Chr.X: 72207054-72207054 on Build GRCh38
rs781419143 Chr.X: 72207144-72207144 on Build GRCh38
rs771728537 Chr.X: 72207204-72207204 on Build GRCh38
rs762834270 Chr.X: 72207313-72207313 on Build GRCh38
rs187128885 Chr.X: 72207230-72207230 on Build GRCh38
rs202048178 Chr.X: 72207368-72207368 on Build GRCh38
rs759554474 Chr.X: 72207315-72207315 on Build GRCh38
rs373407969 Chr.X: 72207075-72207075 on Build GRCh38

View Full Product Details