Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74736312-74736820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA2022
Entrez Gene ID
340533
Gene Name
KIAA2022
Gene Aliases
MRX98, XPN
UniGene
Hs.124128
Gene Chromosome Location
Chr.X: 74732856-74925452 on Build GRCh38
Transcripts
NM_001008537.2

SNP Detail

SNP ID Chromosome Location
rs747862960 Chr.X: 74736688-74736688 on Build GRCh38
rs983980970 Chr.X: 74736458-74736458 on Build GRCh38
rs183369239 Chr.X: 74736630-74736630 on Build GRCh38
rs201381161 Chr.X: 74736774-74736774 on Build GRCh38
rs909804565 Chr.X: 74736462-74736462 on Build GRCh38
rs970877137 Chr.X: 74736789-74736789 on Build GRCh38
rs779640184 Chr.X: 74736434-74736434 on Build GRCh38
rs1028924529 Chr.X: 74736562-74736562 on Build GRCh38
rs959267466 Chr.X: 74736456-74736456 on Build GRCh38
rs937009807 Chr.X: 74736354-74736354 on Build GRCh38
rs981905840 Chr.X: 74736380-74736380 on Build GRCh38
rs926842043 Chr.X: 74736350-74736350 on Build GRCh38
rs754663527 Chr.X: 74736596-74736596 on Build GRCh38
rs1009639304 Chr.X: 74736654-74736654 on Build GRCh38
rs563179382 Chr.X: 74736534-74736534 on Build GRCh38
rs962333479 Chr.X: 74736709-74736709 on Build GRCh38
rs972211980 Chr.X: 74736747-74736747 on Build GRCh38
rs997343452 Chr.X: 74736531-74736531 on Build GRCh38
rs1003274244 Chr.X: 74736766-74736766 on Build GRCh38
rs190903029 Chr.X: 74736519-74736519 on Build GRCh38
rs770646923 Chr.X: 74736505-74736505 on Build GRCh38
rs939984077 Chr.X: 74736478-74736478 on Build GRCh38
rs970863311 Chr.X: 74736739-74736739 on Build GRCh38
rs959161559 Chr.X: 74736796-74736796 on Build GRCh38
rs769676863 Chr.X: 74736697-74736697 on Build GRCh38
rs1020655759 Chr.X: 74736672-74736672 on Build GRCh38
rs922675913 Chr.X: 74736510-74736510 on Build GRCh38
rs1054673130 Chr.X: 74736635-74736635 on Build GRCh38
rs892055289 Chr.X: 74736639-74736639 on Build GRCh38

View Full Product Details