Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100827957-100828457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF2
Entrez Gene ID
1478
Gene Name
cleavage stimulation factor subunit 2
Gene Aliases
CstF-64
UniGene
Hs.132370
Gene Chromosome Location
Chr.X: 100820359-100841520 on Build GRCh38
Transcripts
NM_001306206.1 NM_001306209.1 NM_001325.2

SNP Detail

SNP ID Chromosome Location
rs759614429 Chr.X: 100828113-100828113 on Build GRCh38
rs112381744 Chr.X: 100828159-100828159 on Build GRCh38
rs1032018252 Chr.X: 100828165-100828165 on Build GRCh38
rs377525954 Chr.X: 100828324-100828324 on Build GRCh38
rs906049778 Chr.X: 100828253-100828253 on Build GRCh38
rs186961263 Chr.X: 100828073-100828073 on Build GRCh38
rs751759272 Chr.X: 100828404-100828404 on Build GRCh38
rs942310149 Chr.X: 100828169-100828169 on Build GRCh38
rs150355616 Chr.X: 100828386-100828386 on Build GRCh38
rs761824199 Chr.X: 100828264-100828264 on Build GRCh38
rs748011738 Chr.X: 100828048-100828048 on Build GRCh38
rs373434300 Chr.X: 100827994-100827994 on Build GRCh38
rs954330490 Chr.X: 100828058-100828058 on Build GRCh38
rs904416577 Chr.X: 100828394-100828394 on Build GRCh38
rs913541988 Chr.X: 100828293-100828293 on Build GRCh38
rs755231782 Chr.X: 100828427-100828427 on Build GRCh38
rs1037811390 Chr.X: 100828207-100828207 on Build GRCh38
rs751916747 Chr.X: 100828147-100828147 on Build GRCh38
rs1048249314 Chr.X: 100828409-100828409 on Build GRCh38
rs748739572 Chr.X: 100828024-100828024 on Build GRCh38
rs987808137 Chr.X: 100828239-100828239 on Build GRCh38
rs758827409 Chr.X: 100828131-100828131 on Build GRCh38
rs1010641877 Chr.X: 100828382-100828382 on Build GRCh38
rs368898975 Chr.X: 100828128-100828128 on Build GRCh38
rs73636621 Chr.X: 100828304-100828304 on Build GRCh38
rs779726529 Chr.X: 100827995-100827995 on Build GRCh38
rs766527740 Chr.X: 100828080-100828080 on Build GRCh38
rs1000118488 Chr.X: 100828410-100828410 on Build GRCh38
rs182379967 Chr.X: 100828241-100828241 on Build GRCh38
rs372679275 Chr.X: 100828135-100828135 on Build GRCh38
rs543076361 Chr.X: 100828342-100828342 on Build GRCh38
rs771938537 Chr.X: 100828059-100828059 on Build GRCh38
rs140971340 Chr.X: 100828350-100828350 on Build GRCh38
rs772982259 Chr.X: 100828068-100828068 on Build GRCh38
rs754990201 Chr.X: 100828129-100828129 on Build GRCh38
rs187093073 Chr.X: 100828360-100828360 on Build GRCh38
rs191919621 Chr.X: 100828224-100828224 on Build GRCh38
rs908106697 Chr.X: 100828121-100828121 on Build GRCh38
rs771691143 Chr.X: 100827983-100827983 on Build GRCh38
rs765213897 Chr.X: 100828127-100828127 on Build GRCh38
rs957776697 Chr.X: 100828233-100828233 on Build GRCh38
rs776726269 Chr.X: 100828109-100828109 on Build GRCh38
rs926477530 Chr.X: 100828373-100828373 on Build GRCh38
rs141539257 Chr.X: 100828026-100828026 on Build GRCh38
rs750446774 Chr.X: 100828336-100828336 on Build GRCh38
rs937791853 Chr.X: 100828391-100828391 on Build GRCh38

View Full Product Details