Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93184273-93184801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOAP1
Entrez Gene ID
64112
Gene Name
modulator of apoptosis 1
Gene Aliases
MAP-1, PNMA4
UniGene
Hs.24719
Gene Chromosome Location
Chr.14: 93182196-93184904 on Build GRCh38
Transcripts
NM_022151.4

SNP Detail

SNP ID Chromosome Location
rs965623069 Chr.14: 93184495-93184495 on Build GRCh38
rs572174480 Chr.14: 93184710-93184710 on Build GRCh38
rs536625619 Chr.14: 93184641-93184641 on Build GRCh38
rs551674717 Chr.14: 93184450-93184450 on Build GRCh38
rs571772886 Chr.14: 93184457-93184457 on Build GRCh38
rs879051957 Chr.14: 93184300-93184300 on Build GRCh38
rs1033874553 Chr.14: 93184385-93184385 on Build GRCh38
rs114198532 Chr.14: 93184540-93184540 on Build GRCh38
rs1019839995 Chr.14: 93184494-93184494 on Build GRCh38
rs538480174 Chr.14: 93184371-93184371 on Build GRCh38
rs11627341 Chr.14: 93184503-93184503 on Build GRCh38
rs944086074 Chr.14: 93184716-93184716 on Build GRCh38
rs1048086305 Chr.14: 93184768-93184768 on Build GRCh38
rs57461133 Chr.14: 93184511-93184511 on Build GRCh38
rs545246102 Chr.14: 93184700-93184700 on Build GRCh38
rs940640155 Chr.14: 93184634-93184634 on Build GRCh38
rs929265986 Chr.14: 93184760-93184760 on Build GRCh38
rs11545119 Chr.14: 93184689-93184689 on Build GRCh38
rs574099622 Chr.14: 93184541-93184541 on Build GRCh38
rs927335002 Chr.14: 93184582-93184582 on Build GRCh38
rs370830576 Chr.14: 93184698-93184698 on Build GRCh38
rs959508358 Chr.14: 93184407-93184407 on Build GRCh38
rs748977744 Chr.14: 93184433-93184433 on Build GRCh38
rs570011739 Chr.14: 93184351-93184351 on Build GRCh38
rs540782517 Chr.14: 93184754-93184754 on Build GRCh38
rs180750547 Chr.14: 93184352-93184352 on Build GRCh38
rs8017818 Chr.14: 93184709-93184709 on Build GRCh38
rs925337394 Chr.14: 93184706-93184706 on Build GRCh38
rs1033446122 Chr.14: 93184664-93184664 on Build GRCh38
rs887675420 Chr.14: 93184777-93184777 on Build GRCh38
rs1057180041 Chr.14: 93184603-93184603 on Build GRCh38
rs1008582673 Chr.14: 93184721-93184721 on Build GRCh38
rs911313189 Chr.14: 93184712-93184712 on Build GRCh38
rs937343898 Chr.14: 93184587-93184587 on Build GRCh38
rs918187652 Chr.14: 93184428-93184428 on Build GRCh38
rs116450723 Chr.14: 93184308-93184308 on Build GRCh38
rs992302654 Chr.14: 93184417-93184417 on Build GRCh38
rs992337121 Chr.14: 93184686-93184686 on Build GRCh38
rs112065765 Chr.14: 93184486-93184486 on Build GRCh38
rs1008486268 Chr.14: 93184468-93184468 on Build GRCh38
rs953054859 Chr.14: 93184465-93184465 on Build GRCh38
rs868248914 Chr.14: 93184456-93184456 on Build GRCh38
rs1004788104 Chr.14: 93184405-93184405 on Build GRCh38
rs979264572 Chr.14: 93184546-93184546 on Build GRCh38
rs1026011782 Chr.14: 93184626-93184626 on Build GRCh38
rs757903490 Chr.14: 93184291-93184291 on Build GRCh38
rs917243381 Chr.14: 93184630-93184630 on Build GRCh38
rs370381575 Chr.14: 93184350-93184350 on Build GRCh38
rs556542967 Chr.14: 93184645-93184645 on Build GRCh38
rs898995469 Chr.14: 93184670-93184670 on Build GRCh38
rs1025111015 Chr.14: 93184687-93184687 on Build GRCh38
rs901676525 Chr.14: 93184755-93184755 on Build GRCh38

View Full Product Details