Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75657260-75657748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL19
Entrez Gene ID
9801
Gene Name
mitochondrial ribosomal protein L19
Gene Aliases
L19mt, MRP-L15, MRP-L19, MRPL15, RLX1, RPML15
UniGene
Hs.44024
Gene Chromosome Location
Chr.2: 75646783-75662208 on Build GRCh38
Transcripts
NM_014763.3

SNP Detail

SNP ID Chromosome Location
rs551882837 Chr.2: 75657436-75657436 on Build GRCh38
rs923479175 Chr.2: 75657666-75657666 on Build GRCh38
rs933325372 Chr.2: 75657360-75657360 on Build GRCh38
rs1031242913 Chr.2: 75657680-75657680 on Build GRCh38
rs935520301 Chr.2: 75657376-75657376 on Build GRCh38
rs528354268 Chr.2: 75657457-75657457 on Build GRCh38
rs1018261289 Chr.2: 75657620-75657620 on Build GRCh38
rs184807308 Chr.2: 75657348-75657348 on Build GRCh38
rs147211744 Chr.2: 75657626-75657626 on Build GRCh38
rs1001065632 Chr.2: 75657495-75657495 on Build GRCh38
rs988348242 Chr.2: 75657351-75657351 on Build GRCh38
rs968318725 Chr.2: 75657400-75657400 on Build GRCh38
rs764024072 Chr.2: 75657710-75657711 on Build GRCh38
rs917000994 Chr.2: 75657422-75657422 on Build GRCh38
rs1038327568 Chr.2: 75657429-75657429 on Build GRCh38
rs183561889 Chr.2: 75657475-75657475 on Build GRCh38
rs563370452 Chr.2: 75657353-75657354 on Build GRCh38
rs577120500 Chr.2: 75657704-75657704 on Build GRCh38
rs969035659 Chr.2: 75657493-75657493 on Build GRCh38
rs896195123 Chr.2: 75657455-75657455 on Build GRCh38
rs954917224 Chr.2: 75657668-75657668 on Build GRCh38
rs1046797697 Chr.2: 75657452-75657452 on Build GRCh38
rs770761842 Chr.2: 75657614-75657614 on Build GRCh38
rs745329853 Chr.2: 75657403-75657403 on Build GRCh38
rs1032597499 Chr.2: 75657541-75657541 on Build GRCh38
rs541873855 Chr.2: 75657355-75657355 on Build GRCh38
rs137865984 Chr.2: 75657333-75657333 on Build GRCh38
rs1029949793 Chr.2: 75657563-75657563 on Build GRCh38
rs112655619 Chr.2: 75657584-75657584 on Build GRCh38
rs773363352 Chr.2: 75657310-75657311 on Build GRCh38
rs536365289 Chr.2: 75657546-75657546 on Build GRCh38
rs925422503 Chr.2: 75657301-75657301 on Build GRCh38
rs555082699 Chr.2: 75657551-75657551 on Build GRCh38
rs566692713 Chr.2: 75657449-75657450 on Build GRCh38
rs186882563 Chr.2: 75657391-75657391 on Build GRCh38
rs570032980 Chr.2: 75657622-75657622 on Build GRCh38
rs558479235 Chr.2: 75657682-75657682 on Build GRCh38
rs1022547249 Chr.2: 75657372-75657372 on Build GRCh38
rs896947955 Chr.2: 75657501-75657501 on Build GRCh38
rs574174422 Chr.2: 75657589-75657592 on Build GRCh38
rs1013356651 Chr.2: 75657466-75657466 on Build GRCh38
rs568234960 Chr.2: 75657487-75657487 on Build GRCh38
rs193016209 Chr.2: 75657309-75657309 on Build GRCh38
rs191368345 Chr.2: 75657450-75657450 on Build GRCh38
rs780816707 Chr.2: 75657354-75657354 on Build GRCh38
rs769867969 Chr.2: 75657712-75657712 on Build GRCh38
rs143422178 Chr.2: 75657399-75657399 on Build GRCh38

View Full Product Details