Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97142355-97142913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
UniGene
Hs.541894
Gene Chromosome Location
Chr.2: 97113186-97264521 on Build GRCh38
Transcripts
NM_001164315.1 XM_006712514.3 XM_011511130.1 XM_011511131.1 XM_017004009.1 XM_017004010.1 XM_017004011.1 XM_017004012.1 XM_017004013.1 XM_017004014.1 XM_017004015.1 XM_017004016.1 XM_017004017.1 XM_017004018.1 XM_017004019.1 XM_017004020.1 XM_017004021.1 XM_017004022.1 XM_017004023.1 XM_017004024.1 XM_017004025.1 XM_017004026.1 XM_017004027.1 XM_017004028.1 XM_017004029.1 XM_017004030.1 XM_017004031.1 XM_017004032.1 XM_017004033.1 XM_017004034.1 XM_017004035.1 XM_017004036.1 XM_017004037.1 XM_017004038.1 XM_017004039.1 XM_017004040.1 XM_017004041.1 XM_017004042.1 XM_017004043.1 XM_017004044.1 XM_017004045.1 XM_017004046.1 XM_017004047.1 XM_017004048.1 XM_017004049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6335376 c.828+7A>T p.? Chr.2: 97142675-97142675 on Build GRCh38
6600701 c.815A>C p.Q272P Chr.2: 97142655-97142655 on Build GRCh38
5030609 c.901+4_901+5insA p.? Chr.2: 97142839-97142840 on Build GRCh38
127071 c.901+2_901+3insA p.? Chr.2: 97142837-97142838 on Build GRCh38
3933723 c.891A>C p.K297N Chr.2: 97142825-97142825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750336673 Chr.2: 97142839-97142839 on Build GRCh38
rs777692262 Chr.2: 97142780-97142780 on Build GRCh38
rs748048983 Chr.2: 97142617-97142617 on Build GRCh38
rs760325783 Chr.2: 97142632-97142632 on Build GRCh38
rs62153044 Chr.2: 97142657-97142657 on Build GRCh38
rs781227646 Chr.2: 97142593-97142593 on Build GRCh38
rs760929257 Chr.2: 97142632-97142633 on Build GRCh38
rs74969160 Chr.2: 97142492-97142492 on Build GRCh38
rs775068333 Chr.2: 97142770-97142770 on Build GRCh38
rs752007991 Chr.2: 97142653-97142653 on Build GRCh38
rs747469072 Chr.2: 97142791-97142791 on Build GRCh38
rs556502593 Chr.2: 97142562-97142562 on Build GRCh38
rs112299457 Chr.2: 97142692-97142692 on Build GRCh38
rs759716334 Chr.2: 97142776-97142776 on Build GRCh38
rs878945517 Chr.2: 97142505-97142505 on Build GRCh38
rs759962475 Chr.2: 97142799-97142799 on Build GRCh38
rs757138047 Chr.2: 97142781-97142781 on Build GRCh38
rs1054821603 Chr.2: 97142408-97142408 on Build GRCh38
rs751298876 Chr.2: 97142829-97142829 on Build GRCh38
rs201265245 Chr.2: 97142778-97142778 on Build GRCh38
rs765361964 Chr.2: 97142815-97142815 on Build GRCh38
rs376259802 Chr.2: 97142837-97142838 on Build GRCh38
rs756131132 Chr.2: 97142661-97142661 on Build GRCh38
rs755250109 Chr.2: 97142670-97142670 on Build GRCh38
rs759266891 Chr.2: 97142649-97142649 on Build GRCh38
rs747993542 Chr.2: 97142680-97142680 on Build GRCh38
rs766321031 Chr.2: 97142749-97142749 on Build GRCh38
rs919140694 Chr.2: 97142738-97142738 on Build GRCh38
rs202223019 Chr.2: 97142787-97142787 on Build GRCh38
rs752523921 Chr.2: 97142760-97142760 on Build GRCh38
rs774694336 Chr.2: 97142713-97142713 on Build GRCh38
rs1021178681 Chr.2: 97142471-97142471 on Build GRCh38
rs184098361 Chr.2: 97142777-97142777 on Build GRCh38
rs771325097 Chr.2: 97142792-97142792 on Build GRCh38
rs768079503 Chr.2: 97142656-97142656 on Build GRCh38
rs749429130 Chr.2: 97142664-97142664 on Build GRCh38
rs757731221 Chr.2: 97142654-97142654 on Build GRCh38
rs764320743 Chr.2: 97142651-97142651 on Build GRCh38
rs191673442 Chr.2: 97142443-97142443 on Build GRCh38
rs188855382 Chr.2: 97142871-97142871 on Build GRCh38
rs780541116 Chr.2: 97142767-97142767 on Build GRCh38
rs776083963 Chr.2: 97142646-97142646 on Build GRCh38
rs188603624 Chr.2: 97142859-97142859 on Build GRCh38
rs202056714 Chr.2: 97142844-97142844 on Build GRCh38
rs745451855 Chr.2: 97142769-97142769 on Build GRCh38
rs772182251 Chr.2: 97142619-97142619 on Build GRCh38
rs774233380 Chr.2: 97142614-97142614 on Build GRCh38
rs62153043 Chr.2: 97142612-97142612 on Build GRCh38
rs758206122 Chr.2: 97142764-97142764 on Build GRCh38
rs954180911 Chr.2: 97142583-97142583 on Build GRCh38
rs761199537 Chr.2: 97142740-97142740 on Build GRCh38
rs773587565 Chr.2: 97142772-97142772 on Build GRCh38
rs62153042 Chr.2: 97142609-97142609 on Build GRCh38
rs770309288 Chr.2: 97142697-97142697 on Build GRCh38
rs770953660 Chr.2: 97142766-97142766 on Build GRCh38
rs775777240 Chr.2: 97142615-97142616 on Build GRCh38
rs769282495 Chr.2: 97142783-97142783 on Build GRCh38
rs745987203 Chr.2: 97142606-97142606 on Build GRCh38
rs192915513 Chr.2: 97142875-97142875 on Build GRCh38
rs766114111 Chr.2: 97142639-97142639 on Build GRCh38
rs769527612 Chr.2: 97142712-97142712 on Build GRCh38
rs902807554 Chr.2: 97142885-97142885 on Build GRCh38
rs762225822 Chr.2: 97142715-97142715 on Build GRCh38
rs569751784 Chr.2: 97142831-97142831 on Build GRCh38
rs778549119 Chr.2: 97142678-97142678 on Build GRCh38
rs377477830 Chr.2: 97142581-97142581 on Build GRCh38
rs763239572 Chr.2: 97142823-97142823 on Build GRCh38
rs767222137 Chr.2: 97142836-97142836 on Build GRCh38
rs779515367 Chr.2: 97142847-97142847 on Build GRCh38
rs375469632 Chr.2: 97142735-97142735 on Build GRCh38
rs79070930 Chr.2: 97142481-97142481 on Build GRCh38
rs75359815 Chr.2: 97142825-97142825 on Build GRCh38
rs772127177 Chr.2: 97142687-97142687 on Build GRCh38
rs201677961 Chr.2: 97142445-97142445 on Build GRCh38
rs62153045 Chr.2: 97142758-97142758 on Build GRCh38
rs748889829 Chr.2: 97142785-97142785 on Build GRCh38
rs777135061 Chr.2: 97142797-97142797 on Build GRCh38
rs879069986 Chr.2: 97142542-97142542 on Build GRCh38
rs749537041 Chr.2: 97142610-97142610 on Build GRCh38
rs71429319 Chr.2: 97142425-97142425 on Build GRCh38
rs759092881 Chr.2: 97142710-97142710 on Build GRCh38
rs896140413 Chr.2: 97142409-97142409 on Build GRCh38
rs755120173 Chr.2: 97142752-97142752 on Build GRCh38
rs779570197 Chr.2: 97142784-97142784 on Build GRCh38
rs767883329 Chr.2: 97142729-97142729 on Build GRCh38
rs771257830 Chr.2: 97142883-97142883 on Build GRCh38
rs192610966 Chr.2: 97142877-97142877 on Build GRCh38
rs56786098 Chr.2: 97142461-97142461 on Build GRCh38
rs745808582 Chr.2: 97142782-97142782 on Build GRCh38
rs754620059 Chr.2: 97142861-97142861 on Build GRCh38
rs772814301 Chr.2: 97142625-97142625 on Build GRCh38
rs200495869 Chr.2: 97142577-97142577 on Build GRCh38
rs199920970 Chr.2: 97142773-97142773 on Build GRCh38
rs777745471 Chr.2: 97142765-97142765 on Build GRCh38
rs779899993 Chr.2: 97142865-97142865 on Build GRCh38
rs776230376 Chr.2: 97142698-97142698 on Build GRCh38
rs896156798 Chr.2: 97142832-97142832 on Build GRCh38
rs757020376 Chr.2: 97142833-97142833 on Build GRCh38
rs753998370 Chr.2: 97142751-97142751 on Build GRCh38
rs551610867 Chr.2: 97142819-97142819 on Build GRCh38
rs937755197 Chr.2: 97142393-97142393 on Build GRCh38
rs199536652 Chr.2: 97142412-97142412 on Build GRCh38
rs200040989 Chr.2: 97142423-97142423 on Build GRCh38
rs1001068565 Chr.2: 97142487-97142487 on Build GRCh38
rs762599299 Chr.2: 97142771-97142771 on Build GRCh38
rs902691251 Chr.2: 97142479-97142479 on Build GRCh38
rs773338633 Chr.2: 97142690-97142690 on Build GRCh38

View Full Product Details