Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101001498-101002053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC12
Entrez Gene ID
10071
Gene Name
mucin 12, cell surface associated
Gene Aliases
MUC-11, MUC-12, MUC11
UniGene
Hs.489355
Gene Chromosome Location
Chr.7: 100969623-101018949 on Build GRCh38
Transcripts
NM_001164462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4414996 c.11149C>T p.R3717C Chr.7: 101001712-101001712 on Build GRCh38
3684856 c.11260A>C p.T3754P Chr.7: 101001823-101001823 on Build GRCh38
3684852 c.11248T>C p.S3750P Chr.7: 101001811-101001811 on Build GRCh38
6409942 c.11476A>G p.S3826G Chr.7: 101001610-101001610 on Build GRCh38
4414997 c.11578C>T p.R3860C Chr.7: 101001712-101001712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773656841 Chr.7: 101002019-101002019 on Build GRCh38
rs202085425 Chr.7: 101001680-101001680 on Build GRCh38
rs201110833 Chr.7: 101001674-101001674 on Build GRCh38
rs201665808 Chr.7: 101001599-101001599 on Build GRCh38
rs200339904 Chr.7: 101001881-101001881 on Build GRCh38
rs147541791 Chr.7: 101001840-101001840 on Build GRCh38
rs745433915 Chr.7: 101001956-101001956 on Build GRCh38
rs749136607 Chr.7: 101001878-101001878 on Build GRCh38
rs770006229 Chr.7: 101001988-101001988 on Build GRCh38
rs771849115 Chr.7: 101001960-101001960 on Build GRCh38
rs757255908 Chr.7: 101001932-101001932 on Build GRCh38
rs200120045 Chr.7: 101001647-101001647 on Build GRCh38
rs201148522 Chr.7: 101001890-101001890 on Build GRCh38
rs200935425 Chr.7: 101001713-101001713 on Build GRCh38
rs778268742 Chr.7: 101001935-101001935 on Build GRCh38
rs202003798 Chr.7: 101001961-101001961 on Build GRCh38
rs200394041 Chr.7: 101001712-101001712 on Build GRCh38
rs199931267 Chr.7: 101002030-101002030 on Build GRCh38
rs776080811 Chr.7: 101002026-101002028 on Build GRCh38
rs548804581 Chr.7: 101002028-101002028 on Build GRCh38
rs746675102 Chr.7: 101001977-101001977 on Build GRCh38

View Full Product Details