Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15459607-15459761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIR
Entrez Gene ID
8544
Gene Name
pirin
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15384799-15493588 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

Gene Symbol
PIR-FIGF
Entrez Gene ID
100532742
Gene Name
PIR-FIGF readthrough
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15345591-15491309 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

SNP Detail

SNP ID Chromosome Location
rs780078643 Chr.X: 15459721-15459721 on Build GRCh38
rs750390136 Chr.X: 15459699-15459699 on Build GRCh38
rs778749678 Chr.X: 15459690-15459690 on Build GRCh38
rs757045955 Chr.X: 15459688-15459688 on Build GRCh38
rs763677923 Chr.X: 15459675-15459675 on Build GRCh38
rs753491411 Chr.X: 15459684-15459684 on Build GRCh38
rs748257098 Chr.X: 15459722-15459722 on Build GRCh38
rs1042818236 Chr.X: 15459698-15459698 on Build GRCh38
rs760120323 Chr.X: 15459631-15459631 on Build GRCh38
rs866898423 Chr.X: 15459660-15459660 on Build GRCh38
rs758408940 Chr.X: 15459705-15459705 on Build GRCh38

View Full Product Details