Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24313227-24313771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT20HL2
Entrez Gene ID
170067
Gene Name
SPT20 homolog, SAGA complex component-like 2
Gene Aliases
FAM48B1, FAM48B2
UniGene
Hs.716893
Gene Chromosome Location
Chr.X: 24310862-24313315 on Build GRCh38
Transcripts
NM_001136233.1

SNP Detail

SNP ID Chromosome Location
rs753368559 Chr.X: 24313733-24313733 on Build GRCh38
rs776353199 Chr.X: 24313442-24313442 on Build GRCh38
rs757618135 Chr.X: 24313537-24313538 on Build GRCh38
rs757450324 Chr.X: 24313663-24313663 on Build GRCh38
rs1020512627 Chr.X: 24313743-24313743 on Build GRCh38
rs773717904 Chr.X: 24313349-24313349 on Build GRCh38
rs532648872 Chr.X: 24313630-24313630 on Build GRCh38
rs1055435323 Chr.X: 24313332-24313332 on Build GRCh38
rs370831048 Chr.X: 24313420-24313420 on Build GRCh38
rs752234360 Chr.X: 24313271-24313271 on Build GRCh38
rs375991307 Chr.X: 24313269-24313269 on Build GRCh38
rs200885459 Chr.X: 24313538-24313539 on Build GRCh38
rs1018258719 Chr.X: 24313513-24313513 on Build GRCh38
rs770092521 Chr.X: 24313703-24313703 on Build GRCh38
rs756015899 Chr.X: 24313280-24313280 on Build GRCh38
rs372582351 Chr.X: 24313295-24313295 on Build GRCh38
rs771126843 Chr.X: 24313375-24313375 on Build GRCh38
rs779033553 Chr.X: 24313678-24313678 on Build GRCh38
rs187810490 Chr.X: 24313544-24313544 on Build GRCh38
rs759315497 Chr.X: 24313364-24313364 on Build GRCh38
rs764849491 Chr.X: 24313463-24313463 on Build GRCh38
rs747276547 Chr.X: 24313308-24313308 on Build GRCh38
rs138774825 Chr.X: 24313449-24313449 on Build GRCh38
rs763962638 Chr.X: 24313377-24313377 on Build GRCh38
rs1727961 Chr.X: 24313416-24313416 on Build GRCh38
rs749662017 Chr.X: 24313587-24313587 on Build GRCh38
rs759231747 Chr.X: 24313261-24313261 on Build GRCh38
rs771461026 Chr.X: 24313359-24313359 on Build GRCh38
rs998428747 Chr.X: 24313620-24313620 on Build GRCh38
rs745881605 Chr.X: 24313705-24313705 on Build GRCh38
rs770208861 Chr.X: 24313338-24313338 on Build GRCh38
rs369122902 Chr.X: 24313282-24313282 on Build GRCh38
rs774194699 Chr.X: 24313259-24313259 on Build GRCh38
rs766149204 Chr.X: 24313257-24313257 on Build GRCh38
rs753609875 Chr.X: 24313284-24313284 on Build GRCh38
rs182774993 Chr.X: 24313382-24313382 on Build GRCh38
rs781187767 Chr.X: 24313328-24313328 on Build GRCh38
rs748202696 Chr.X: 24313336-24313336 on Build GRCh38
rs371393621 Chr.X: 24313644-24313644 on Build GRCh38
rs374546337 Chr.X: 24313512-24313512 on Build GRCh38
rs774864892 Chr.X: 24313372-24313372 on Build GRCh38
rs774851354 Chr.X: 24313361-24313361 on Build GRCh38
rs994591050 Chr.X: 24313410-24313410 on Build GRCh38
rs1007416827 Chr.X: 24313431-24313431 on Build GRCh38
rs374125552 Chr.X: 24313753-24313753 on Build GRCh38
rs368948962 Chr.X: 24313643-24313643 on Build GRCh38
rs190821449 Chr.X: 24313567-24313567 on Build GRCh38
rs760368337 Chr.X: 24313695-24313695 on Build GRCh38
rs749788440 Chr.X: 24313353-24313353 on Build GRCh38
rs767135917 Chr.X: 24313369-24313369 on Build GRCh38
rs965797796 Chr.X: 24313525-24313525 on Build GRCh38
rs759633653 Chr.X: 24313415-24313415 on Build GRCh38

View Full Product Details