Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55077255-55077802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAGE2B
Entrez Gene ID
389860
Gene Name
PAGE family member 2B
Gene Aliases
CT16.5, GAGEE3, PAGE-2B
UniGene
Hs.293317
Gene Chromosome Location
Chr.X: 55027927-55078903 on Build GRCh38
Transcripts
NM_001015038.2 XM_011530785.2 XM_011530786.2 XM_011530787.2 XM_017029513.1

SNP Detail

SNP ID Chromosome Location
rs762951892 Chr.X: 55077412-55077412 on Build GRCh38
rs770941394 Chr.X: 55077558-55077558 on Build GRCh38
rs188237400 Chr.X: 55077389-55077389 on Build GRCh38
rs753233574 Chr.X: 55077478-55077478 on Build GRCh38
rs191228717 Chr.X: 55077778-55077778 on Build GRCh38
rs780381830 Chr.X: 55077485-55077485 on Build GRCh38
rs2270533 Chr.X: 55077300-55077300 on Build GRCh38
rs752959813 Chr.X: 55077524-55077524 on Build GRCh38
rs766178237 Chr.X: 55077422-55077422 on Build GRCh38
rs149758580 Chr.X: 55077498-55077498 on Build GRCh38
rs749287217 Chr.X: 55077552-55077552 on Build GRCh38
rs1053004748 Chr.X: 55077587-55077587 on Build GRCh38
rs776260356 Chr.X: 55077324-55077324 on Build GRCh38
rs758768563 Chr.X: 55077364-55077364 on Build GRCh38
rs760596339 Chr.X: 55077281-55077281 on Build GRCh38
rs759296702 Chr.X: 55077454-55077454 on Build GRCh38
rs773354245 Chr.X: 55077541-55077541 on Build GRCh38
rs1965321 Chr.X: 55077375-55077375 on Build GRCh38
rs771203770 Chr.X: 55077738-55077738 on Build GRCh38
rs773089488 Chr.X: 55077398-55077398 on Build GRCh38
rs1046921553 Chr.X: 55077739-55077739 on Build GRCh38
rs750103545 Chr.X: 55077353-55077353 on Build GRCh38
rs752335336 Chr.X: 55077463-55077463 on Build GRCh38
rs763668231 Chr.X: 55077471-55077471 on Build GRCh38
rs891661083 Chr.X: 55077731-55077731 on Build GRCh38
rs1052367 Chr.X: 55077484-55077484 on Build GRCh38
rs747415188 Chr.X: 55077489-55077489 on Build GRCh38
rs764749209 Chr.X: 55077349-55077349 on Build GRCh38
rs145809926 Chr.X: 55077425-55077425 on Build GRCh38
rs765982162 Chr.X: 55077381-55077381 on Build GRCh38
rs4520303 Chr.X: 55077334-55077334 on Build GRCh38
rs376448393 Chr.X: 55077743-55077743 on Build GRCh38
rs756518576 Chr.X: 55077641-55077641 on Build GRCh38
rs370035288 Chr.X: 55077772-55077772 on Build GRCh38
rs755601281 Chr.X: 55077357-55077357 on Build GRCh38
rs1965320 Chr.X: 55077346-55077346 on Build GRCh38
rs373279664 Chr.X: 55077715-55077715 on Build GRCh38
rs748285951 Chr.X: 55077383-55077383 on Build GRCh38
rs868722407 Chr.X: 55077771-55077771 on Build GRCh38
rs769840003 Chr.X: 55077533-55077533 on Build GRCh38
rs781659082 Chr.X: 55077526-55077527 on Build GRCh38
rs781620826 Chr.X: 55077503-55077503 on Build GRCh38
rs146503411 Chr.X: 55077456-55077456 on Build GRCh38
rs746385422 Chr.X: 55077358-55077358 on Build GRCh38
rs755691367 Chr.X: 55077468-55077468 on Build GRCh38
rs777795620 Chr.X: 55077648-55077648 on Build GRCh38
rs780476493 Chr.X: 55077370-55077370 on Build GRCh38
rs776960529 Chr.X: 55077382-55077382 on Build GRCh38

View Full Product Details