Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32154319-32154826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507547
Entrez Gene ID
100507547
Gene Name
uncharacterized LOC100507547
Gene Aliases
-
UniGene
Hs.549204
Gene Chromosome Location
Chr.6: 32152802-32154365 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

Gene Symbol
PPT2
Entrez Gene ID
9374
Gene Name
palmitoyl-protein thioesterase 2
Gene Aliases
C6orf8, G14, PPT-2
UniGene
Hs.332138
Gene Chromosome Location
Chr.6: 32153452-32163681 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

Gene Symbol
PPT2-EGFL8
Entrez Gene ID
100532746
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32153999-32168285 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4607256 c.17G>A p.S6N Chr.6: 32154593-32154593 on Build GRCh38
7087995 c.15C>G p.C5W Chr.6: 32154609-32154609 on Build GRCh38
596133 c.157G>T p.D53Y Chr.6: 32154733-32154733 on Build GRCh38
7087994 c.33C>G p.C11W Chr.6: 32154609-32154609 on Build GRCh38
3348378 c.90G>A p.L30L Chr.6: 32154666-32154666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547841904 Chr.6: 32154660-32154660 on Build GRCh38
rs766038131 Chr.6: 32154363-32154363 on Build GRCh38
rs1015439217 Chr.6: 32154733-32154733 on Build GRCh38
rs767666652 Chr.6: 32154591-32154591 on Build GRCh38
rs775330102 Chr.6: 32154777-32154777 on Build GRCh38
rs890257143 Chr.6: 32154362-32154362 on Build GRCh38
rs758416989 Chr.6: 32154590-32154590 on Build GRCh38
rs765182387 Chr.6: 32154575-32154575 on Build GRCh38
rs1042474476 Chr.6: 32154670-32154670 on Build GRCh38
rs759526196 Chr.6: 32154729-32154729 on Build GRCh38
rs749590210 Chr.6: 32154610-32154610 on Build GRCh38
rs779206910 Chr.6: 32154697-32154697 on Build GRCh38
rs772711131 Chr.6: 32154564-32154564 on Build GRCh38
rs746472495 Chr.6: 32154657-32154657 on Build GRCh38
rs774142318 Chr.6: 32154799-32154799 on Build GRCh38
rs568267465 Chr.6: 32154794-32154794 on Build GRCh38
rs369997504 Chr.6: 32154444-32154444 on Build GRCh38
rs199569262 Chr.6: 32154654-32154654 on Build GRCh38
rs775458858 Chr.6: 32154681-32154681 on Build GRCh38
rs143386727 Chr.6: 32154773-32154773 on Build GRCh38
rs770544843 Chr.6: 32154658-32154658 on Build GRCh38
rs762808191 Chr.6: 32154789-32154789 on Build GRCh38
rs770290541 Chr.6: 32154726-32154726 on Build GRCh38
rs937644020 Chr.6: 32154419-32154420 on Build GRCh38
rs1027535206 Chr.6: 32154752-32154752 on Build GRCh38
rs938275417 Chr.6: 32154619-32154619 on Build GRCh38
rs991824805 Chr.6: 32154420-32154420 on Build GRCh38
rs3134604 Chr.6: 32154609-32154609 on Build GRCh38
rs772596610 Chr.6: 32154653-32154653 on Build GRCh38
rs994756703 Chr.6: 32154737-32154737 on Build GRCh38
rs752623987 Chr.6: 32154587-32154587 on Build GRCh38
rs768988905 Chr.6: 32154710-32154712 on Build GRCh38
rs369636289 Chr.6: 32154805-32154805 on Build GRCh38
rs776138788 Chr.6: 32154667-32154667 on Build GRCh38
rs747613803 Chr.6: 32154719-32154719 on Build GRCh38
rs753945236 Chr.6: 32154690-32154690 on Build GRCh38
rs945364426 Chr.6: 32154662-32154662 on Build GRCh38
rs764099068 Chr.6: 32154684-32154684 on Build GRCh38
rs369769958 Chr.6: 32154544-32154544 on Build GRCh38
rs774730573 Chr.6: 32154720-32154725 on Build GRCh38
rs190824876 Chr.6: 32154531-32154531 on Build GRCh38
rs759240503 Chr.6: 32154669-32154669 on Build GRCh38
rs756143220 Chr.6: 32154687-32154687 on Build GRCh38
rs759249544 Chr.6: 32154573-32154573 on Build GRCh38
rs137893908 Chr.6: 32154571-32154571 on Build GRCh38
rs149978286 Chr.6: 32154668-32154668 on Build GRCh38
rs755281509 Chr.6: 32154620-32154620 on Build GRCh38
rs983128373 Chr.6: 32154543-32154543 on Build GRCh38
rs774833571 Chr.6: 32154549-32154549 on Build GRCh38
rs772961576 Chr.6: 32154532-32154532 on Build GRCh38
rs779258367 Chr.6: 32154633-32154633 on Build GRCh38
rs760329247 Chr.6: 32154566-32154566 on Build GRCh38
rs762361101 Chr.6: 32154552-32154552 on Build GRCh38
rs758805739 Chr.6: 32154702-32154702 on Build GRCh38
rs375515057 Chr.6: 32154572-32154572 on Build GRCh38
rs534337753 Chr.6: 32154743-32154743 on Build GRCh38
rs766537037 Chr.6: 32154689-32154689 on Build GRCh38
rs372923109 Chr.6: 32154560-32154560 on Build GRCh38
rs750552624 Chr.6: 32154598-32154598 on Build GRCh38
rs748585480 Chr.6: 32154634-32154634 on Build GRCh38
rs748532154 Chr.6: 32154700-32154700 on Build GRCh38
rs3096696 Chr.6: 32154695-32154695 on Build GRCh38
rs995185235 Chr.6: 32154522-32154522 on Build GRCh38
rs867433139 Chr.6: 32154693-32154693 on Build GRCh38
rs765057669 Chr.6: 32154675-32154675 on Build GRCh38
rs964642529 Chr.6: 32154498-32154498 on Build GRCh38
rs146938399 Chr.6: 32154398-32154398 on Build GRCh38
rs552013847 Chr.6: 32154686-32154686 on Build GRCh38
rs778252168 Chr.6: 32154708-32154708 on Build GRCh38
rs12523706 Chr.6: 32154699-32154699 on Build GRCh38

View Full Product Details