Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32826036-32826542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAP2
Entrez Gene ID
6891
Gene Name
transporter 2, ATP binding cassette subfamily B member
Gene Aliases
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
UniGene
Hs.502
Gene Chromosome Location
Chr.6: 32821833-32838823 on Build GRCh38
Transcripts
NM_000544.3 NM_001290043.1 NM_018833.2

SNP Detail

SNP ID Chromosome Location
rs1036040108 Chr.6: 32826424-32826424 on Build GRCh38
rs926572769 Chr.6: 32826485-32826485 on Build GRCh38
rs962161273 Chr.6: 32826162-32826162 on Build GRCh38
rs1044042 Chr.6: 32826479-32826479 on Build GRCh38
rs905208663 Chr.6: 32826075-32826075 on Build GRCh38
rs113575716 Chr.6: 32826289-32826289 on Build GRCh38
rs893610790 Chr.6: 32826121-32826121 on Build GRCh38
rs11330427 Chr.6: 32826449-32826449 on Build GRCh38
rs1044043 Chr.6: 32826204-32826204 on Build GRCh38
rs6905503 Chr.6: 32826342-32826342 on Build GRCh38
rs1054424973 Chr.6: 32826067-32826067 on Build GRCh38
rs997015117 Chr.6: 32826294-32826294 on Build GRCh38
rs970749242 Chr.6: 32826263-32826263 on Build GRCh38
rs1008535329 Chr.6: 32826407-32826407 on Build GRCh38
rs758647960 Chr.6: 32826401-32826401 on Build GRCh38
rs915440586 Chr.6: 32826362-32826362 on Build GRCh38
rs6925336 Chr.6: 32826193-32826193 on Build GRCh38
rs143496175 Chr.6: 32826404-32826404 on Build GRCh38
rs952873707 Chr.6: 32826350-32826350 on Build GRCh38
rs754179497 Chr.6: 32826222-32826222 on Build GRCh38
rs559572541 Chr.6: 32826303-32826303 on Build GRCh38
rs1046422285 Chr.6: 32826070-32826070 on Build GRCh38
rs6905122 Chr.6: 32826180-32826180 on Build GRCh38
rs757521213 Chr.6: 32826275-32826275 on Build GRCh38
rs935470636 Chr.6: 32826311-32826311 on Build GRCh38
rs796834094 Chr.6: 32826081-32826081 on Build GRCh38
rs747461573 Chr.6: 32826470-32826470 on Build GRCh38
rs6925520 Chr.6: 32826181-32826181 on Build GRCh38
rs369706835 Chr.6: 32826454-32826454 on Build GRCh38
rs1014649435 Chr.6: 32826139-32826139 on Build GRCh38
rs1017240661 Chr.6: 32826284-32826284 on Build GRCh38
rs767894009 Chr.6: 32826443-32826443 on Build GRCh38
rs759811150 Chr.6: 32826380-32826380 on Build GRCh38
rs386699794 Chr.6: 32826180-32826181 on Build GRCh38
rs1008588948 Chr.6: 32826133-32826133 on Build GRCh38
rs780613165 Chr.6: 32826460-32826460 on Build GRCh38
rs1027485334 Chr.6: 32826451-32826451 on Build GRCh38
rs541794572 Chr.6: 32826373-32826373 on Build GRCh38
rs530518569 Chr.6: 32826417-32826417 on Build GRCh38
rs916574048 Chr.6: 32826254-32826254 on Build GRCh38
rs1053707924 Chr.6: 32826089-32826089 on Build GRCh38
rs1028619466 Chr.6: 32826112-32826112 on Build GRCh38

View Full Product Details