Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33068543-33069075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPA1
Entrez Gene ID
3113
Gene Name
major histocompatibility complex, class II, DP alpha 1
Gene Aliases
DP(W3), DP(W4), HLA-DP1A, HLADP, HLASB, PLT1
UniGene
Hs.347270
Gene Chromosome Location
Chr.6: 33064569-33080778 on Build GRCh38
Transcripts
NM_001242524.1 NM_001242525.1 NM_033554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
483921 c.746G>A p.R249H Chr.6: 33068687-33068687 on Build GRCh38
244920 c.731T>C p.I244T Chr.6: 33068702-33068702 on Build GRCh38
5870345 c.680C>T p.A227V Chr.6: 33068753-33068753 on Build GRCh38
7171746 c.616C>G p.L206V Chr.6: 33069031-33069031 on Build GRCh38
6106435 c.763C>T p.R255W Chr.6: 33068670-33068670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144280705 Chr.6: 33068866-33068866 on Build GRCh38
rs996389870 Chr.6: 33068619-33068619 on Build GRCh38
rs375593288 Chr.6: 33068624-33068624 on Build GRCh38
rs775560319 Chr.6: 33069000-33069000 on Build GRCh38
rs770665831 Chr.6: 33068976-33068976 on Build GRCh38
rs532563735 Chr.6: 33068873-33068873 on Build GRCh38
rs571128277 Chr.6: 33069047-33069047 on Build GRCh38
rs746175208 Chr.6: 33068636-33068636 on Build GRCh38
rs777540597 Chr.6: 33068766-33068766 on Build GRCh38
rs765581858 Chr.6: 33069042-33069042 on Build GRCh38
rs1042434 Chr.6: 33068728-33068728 on Build GRCh38
rs35947638 Chr.6: 33068940-33068940 on Build GRCh38
rs773014134 Chr.6: 33068608-33068608 on Build GRCh38
rs144045930 Chr.6: 33068660-33068660 on Build GRCh38
rs370530697 Chr.6: 33069035-33069035 on Build GRCh38
rs1001154141 Chr.6: 33068570-33068570 on Build GRCh38
rs34702621 Chr.6: 33068931-33068932 on Build GRCh38
rs73739663 Chr.6: 33068854-33068854 on Build GRCh38
rs751649425 Chr.6: 33068709-33068709 on Build GRCh38
rs56119512 Chr.6: 33068803-33068803 on Build GRCh38
rs916692464 Chr.6: 33068735-33068735 on Build GRCh38
rs377649060 Chr.6: 33069033-33069033 on Build GRCh38
rs755432204 Chr.6: 33068734-33068734 on Build GRCh38
rs909073134 Chr.6: 33068658-33068659 on Build GRCh38
rs756588838 Chr.6: 33068656-33068656 on Build GRCh38
rs768136825 Chr.6: 33068600-33068600 on Build GRCh38
rs752142772 Chr.6: 33068875-33068875 on Build GRCh38
rs752840315 Chr.6: 33068613-33068613 on Build GRCh38
rs747632169 Chr.6: 33068752-33068752 on Build GRCh38
rs77981473 Chr.6: 33068932-33068932 on Build GRCh38
rs756565886 Chr.6: 33068719-33068719 on Build GRCh38
rs753289265 Chr.6: 33069044-33069044 on Build GRCh38
rs551534043 Chr.6: 33068642-33068642 on Build GRCh38
rs746827411 Chr.6: 33068621-33068622 on Build GRCh38
rs68071176 Chr.6: 33068611-33068611 on Build GRCh38
rs758608352 Chr.6: 33068614-33068614 on Build GRCh38
rs946783212 Chr.6: 33068739-33068739 on Build GRCh38
rs17509489 Chr.6: 33068772-33068772 on Build GRCh38
rs144535712 Chr.6: 33068807-33068807 on Build GRCh38
rs138654414 Chr.6: 33068607-33068607 on Build GRCh38
rs760782956 Chr.6: 33068822-33068822 on Build GRCh38
rs73739662 Chr.6: 33068853-33068853 on Build GRCh38
rs778429978 Chr.6: 33068741-33068741 on Build GRCh38
rs769698699 Chr.6: 33068989-33068989 on Build GRCh38
rs767720204 Chr.6: 33068716-33068716 on Build GRCh38
rs774053198 Chr.6: 33068602-33068602 on Build GRCh38
rs775250428 Chr.6: 33068913-33068913 on Build GRCh38
rs73400048 Chr.6: 33068617-33068617 on Build GRCh38
rs772240925 Chr.6: 33069015-33069015 on Build GRCh38
rs77746323 Chr.6: 33068624-33068624 on Build GRCh38
rs551415787 Chr.6: 33068718-33068718 on Build GRCh38
rs56046206 Chr.6: 33068771-33068771 on Build GRCh38
rs143120448 Chr.6: 33068669-33068669 on Build GRCh38
rs770307009 Chr.6: 33068687-33068687 on Build GRCh38
rs757423272 Chr.6: 33068710-33068710 on Build GRCh38
rs560435082 Chr.6: 33068814-33068814 on Build GRCh38
rs759515868 Chr.6: 33068699-33068699 on Build GRCh38
rs781137504 Chr.6: 33068980-33068980 on Build GRCh38
rs776776442 Chr.6: 33068695-33068695 on Build GRCh38
rs768191828 Chr.6: 33068662-33068662 on Build GRCh38
rs896485007 Chr.6: 33068706-33068706 on Build GRCh38
rs766747418 Chr.6: 33068782-33068782 on Build GRCh38
rs773120072 Chr.6: 33069018-33069018 on Build GRCh38
rs755485189 Chr.6: 33068826-33068826 on Build GRCh38
rs190707736 Chr.6: 33068670-33068670 on Build GRCh38
rs746123315 Chr.6: 33068625-33068626 on Build GRCh38
rs757994269 Chr.6: 33068851-33068851 on Build GRCh38
rs148463918 Chr.6: 33068844-33068844 on Build GRCh38
rs556101955 Chr.6: 33069013-33069013 on Build GRCh38
rs1042495317 Chr.6: 33068958-33068958 on Build GRCh38
rs774585626 Chr.6: 33068793-33068793 on Build GRCh38
rs386699853 Chr.6: 33068853-33068854 on Build GRCh38
rs77350016 Chr.6: 33068931-33068931 on Build GRCh38
rs369210530 Chr.6: 33068825-33068825 on Build GRCh38
rs537578968 Chr.6: 33068987-33068987 on Build GRCh38
rs759952653 Chr.6: 33069036-33069036 on Build GRCh38
rs950506577 Chr.6: 33068643-33068643 on Build GRCh38
rs68101439 Chr.6: 33068624-33068625 on Build GRCh38
rs760991360 Chr.6: 33069022-33069022 on Build GRCh38
rs34370305 Chr.6: 33068840-33068840 on Build GRCh38
rs150578166 Chr.6: 33068591-33068591 on Build GRCh38
rs7768345 Chr.6: 33068912-33068912 on Build GRCh38
rs945330027 Chr.6: 33068952-33068952 on Build GRCh38
rs533994936 Chr.6: 33068667-33068667 on Build GRCh38
rs917951615 Chr.6: 33068941-33068941 on Build GRCh38
rs764266255 Chr.6: 33068616-33068616 on Build GRCh38
rs111559071 Chr.6: 33068622-33068623 on Build GRCh38
rs766076005 Chr.6: 33068688-33068688 on Build GRCh38
rs773764963 Chr.6: 33068770-33068770 on Build GRCh38
rs764290968 Chr.6: 33068708-33068708 on Build GRCh38
rs766754740 Chr.6: 33068824-33068824 on Build GRCh38
rs761427780 Chr.6: 33068604-33068604 on Build GRCh38
rs543788602 Chr.6: 33068860-33068860 on Build GRCh38
rs760563442 Chr.6: 33068609-33068609 on Build GRCh38
rs199532186 Chr.6: 33068758-33068758 on Build GRCh38
rs200236128 Chr.6: 33068653-33068653 on Build GRCh38
rs1126769 Chr.6: 33068658-33068658 on Build GRCh38

View Full Product Details