Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33278187-33278697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT4
Entrez Gene ID
8705
Gene Name
beta-1,3-galactosyltransferase 4
Gene Aliases
BETA3GALT4, GALT2, GALT4
UniGene
Hs.534375
Gene Chromosome Location
Chr.6: 33277140-33278825 on Build GRCh38
Transcripts
NM_003782.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609231 c.1051delG p.E352fs*22 Chr.6: 33278470-33278470 on Build GRCh38
6440516 c.953C>T p.P318L Chr.6: 33278372-33278372 on Build GRCh38
3873133 c.1064C>T p.A355V Chr.6: 33278483-33278483 on Build GRCh38
3921518 c.861C>T p.L287L Chr.6: 33278280-33278280 on Build GRCh38
305329 c.1136_1137delGA p.*379fs? Chr.6: 33278555-33278556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771293570 Chr.6: 33278352-33278352 on Build GRCh38
rs767249550 Chr.6: 33278531-33278531 on Build GRCh38
rs377384565 Chr.6: 33278586-33278586 on Build GRCh38
rs559571289 Chr.6: 33278520-33278520 on Build GRCh38
rs768899668 Chr.6: 33278293-33278294 on Build GRCh38
rs765740368 Chr.6: 33278372-33278372 on Build GRCh38
rs2231261 Chr.6: 33278415-33278415 on Build GRCh38
rs753821848 Chr.6: 33278553-33278553 on Build GRCh38
rs199514813 Chr.6: 33278568-33278568 on Build GRCh38
rs1018901534 Chr.6: 33278413-33278413 on Build GRCh38
rs756963911 Chr.6: 33278405-33278405 on Build GRCh38
rs1011131495 Chr.6: 33278315-33278315 on Build GRCh38
rs146942820 Chr.6: 33278417-33278417 on Build GRCh38
rs910030558 Chr.6: 33278340-33278341 on Build GRCh38
rs906298019 Chr.6: 33278438-33278438 on Build GRCh38
rs769759557 Chr.6: 33278294-33278294 on Build GRCh38
rs149120716 Chr.6: 33278483-33278483 on Build GRCh38
rs772005642 Chr.6: 33278340-33278340 on Build GRCh38
rs763454153 Chr.6: 33278381-33278381 on Build GRCh38
rs188267327 Chr.6: 33278606-33278606 on Build GRCh38
rs976830774 Chr.6: 33278662-33278662 on Build GRCh38
rs72654493 Chr.6: 33278495-33278495 on Build GRCh38
rs981724603 Chr.6: 33278615-33278615 on Build GRCh38
rs374570461 Chr.6: 33278265-33278265 on Build GRCh38
rs781066162 Chr.6: 33278284-33278284 on Build GRCh38
rs773509608 Chr.6: 33278344-33278344 on Build GRCh38
rs1008790390 Chr.6: 33278396-33278396 on Build GRCh38
rs748237478 Chr.6: 33278411-33278411 on Build GRCh38
rs1050284635 Chr.6: 33278334-33278334 on Build GRCh38
rs758022759 Chr.6: 33278237-33278237 on Build GRCh38
rs749134945 Chr.6: 33278463-33278463 on Build GRCh38
rs779147778 Chr.6: 33278296-33278297 on Build GRCh38
rs776978175 Chr.6: 33278367-33278367 on Build GRCh38
rs776077315 Chr.6: 33278490-33278490 on Build GRCh38
rs1011232859 Chr.6: 33278558-33278558 on Build GRCh38
rs752226336 Chr.6: 33278226-33278226 on Build GRCh38
rs754869913 Chr.6: 33278562-33278562 on Build GRCh38
rs530128430 Chr.6: 33278584-33278584 on Build GRCh38
rs755960143 Chr.6: 33278537-33278537 on Build GRCh38
rs993522064 Chr.6: 33278465-33278465 on Build GRCh38
rs1018286998 Chr.6: 33278651-33278651 on Build GRCh38
rs751207543 Chr.6: 33278262-33278262 on Build GRCh38
rs760833363 Chr.6: 33278346-33278346 on Build GRCh38
rs930204927 Chr.6: 33278328-33278328 on Build GRCh38
rs777889789 Chr.6: 33278575-33278575 on Build GRCh38
rs755413867 Chr.6: 33278217-33278217 on Build GRCh38
rs567561157 Chr.6: 33278618-33278618 on Build GRCh38
rs781412360 Chr.6: 33278588-33278588 on Build GRCh38
rs763301300 Chr.6: 33278502-33278502 on Build GRCh38
rs770000033 Chr.6: 33278487-33278487 on Build GRCh38
rs749452536 Chr.6: 33278271-33278272 on Build GRCh38
rs778910553 Chr.6: 33278303-33278303 on Build GRCh38
rs369629453 Chr.6: 33278470-33278470 on Build GRCh38
rs771117789 Chr.6: 33278473-33278473 on Build GRCh38
rs916111650 Chr.6: 33278595-33278595 on Build GRCh38
rs777508227 Chr.6: 33278252-33278252 on Build GRCh38
rs377704128 Chr.6: 33278380-33278380 on Build GRCh38
rs772101657 Chr.6: 33278461-33278471 on Build GRCh38
rs918587432 Chr.6: 33278664-33278665 on Build GRCh38
rs374919836 Chr.6: 33278548-33278548 on Build GRCh38
rs763725350 Chr.6: 33278506-33278506 on Build GRCh38
rs760083232 Chr.6: 33278369-33278369 on Build GRCh38
rs745864964 Chr.6: 33278285-33278285 on Build GRCh38
rs143166422 Chr.6: 33278484-33278484 on Build GRCh38
rs780072568 Chr.6: 33278458-33278458 on Build GRCh38
rs766034930 Chr.6: 33278549-33278549 on Build GRCh38
rs748229218 Chr.6: 33278339-33278339 on Build GRCh38
rs4713616 Chr.6: 33278469-33278469 on Build GRCh38
rs761609610 Chr.6: 33278524-33278524 on Build GRCh38
rs924864463 Chr.6: 33278665-33278665 on Build GRCh38
rs756046614 Chr.6: 33278453-33278453 on Build GRCh38
rs779673748 Chr.6: 33278613-33278613 on Build GRCh38
rs751260597 Chr.6: 33278390-33278390 on Build GRCh38
rs750057937 Chr.6: 33278533-33278533 on Build GRCh38
rs965421810 Chr.6: 33278661-33278661 on Build GRCh38

View Full Product Details