Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172533357-172533858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237000 c.62+8G>A p.? Chr.1: 172533505-172533505 on Build GRCh38
6317071 c.550G>T p.D184Y Chr.1: 172533397-172533397 on Build GRCh38
6237001 c.650+8G>A p.? Chr.1: 172533505-172533505 on Build GRCh38
6572272 c.516C>G p.L172L Chr.1: 172533363-172533363 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560250046 Chr.1: 172533527-172533527 on Build GRCh38
rs931206302 Chr.1: 172533515-172533515 on Build GRCh38
rs987796516 Chr.1: 172533666-172533666 on Build GRCh38
rs369353070 Chr.1: 172533506-172533506 on Build GRCh38
rs779443744 Chr.1: 172533412-172533412 on Build GRCh38
rs761819273 Chr.1: 172533507-172533507 on Build GRCh38
rs573575388 Chr.1: 172533569-172533569 on Build GRCh38
rs899904824 Chr.1: 172533597-172533597 on Build GRCh38
rs142696673 Chr.1: 172533650-172533650 on Build GRCh38
rs937566721 Chr.1: 172533782-172533782 on Build GRCh38
rs372947243 Chr.1: 172533422-172533422 on Build GRCh38
rs1020025329 Chr.1: 172533594-172533594 on Build GRCh38
rs906093283 Chr.1: 172533532-172533532 on Build GRCh38
rs996960603 Chr.1: 172533600-172533600 on Build GRCh38
rs912562250 Chr.1: 172533452-172533452 on Build GRCh38
rs3216843 Chr.1: 172533587-172533588 on Build GRCh38
rs917603528 Chr.1: 172533751-172533751 on Build GRCh38
rs775383180 Chr.1: 172533449-172533449 on Build GRCh38
rs746767207 Chr.1: 172533657-172533657 on Build GRCh38
rs55696309 Chr.1: 172533588-172533588 on Build GRCh38
rs867821271 Chr.1: 172533720-172533720 on Build GRCh38
rs895478091 Chr.1: 172533815-172533815 on Build GRCh38
rs866140737 Chr.1: 172533421-172533421 on Build GRCh38
rs375476480 Chr.1: 172533679-172533679 on Build GRCh38
rs769505483 Chr.1: 172533442-172533442 on Build GRCh38
rs1009939070 Chr.1: 172533583-172533583 on Build GRCh38
rs927460199 Chr.1: 172533745-172533745 on Build GRCh38
rs868022885 Chr.1: 172533481-172533481 on Build GRCh38
rs191673711 Chr.1: 172533555-172533555 on Build GRCh38
rs139101361 Chr.1: 172533518-172533518 on Build GRCh38
rs377462064 Chr.1: 172533384-172533385 on Build GRCh38
rs45505403 Chr.1: 172533477-172533477 on Build GRCh38
rs184136865 Chr.1: 172533633-172533633 on Build GRCh38
rs760770569 Chr.1: 172533530-172533530 on Build GRCh38
rs562162216 Chr.1: 172533611-172533611 on Build GRCh38
rs780348355 Chr.1: 172533391-172533391 on Build GRCh38
rs187803389 Chr.1: 172533553-172533553 on Build GRCh38
rs376204309 Chr.1: 172533471-172533471 on Build GRCh38
rs545968714 Chr.1: 172533795-172533795 on Build GRCh38
rs186609945 Chr.1: 172533701-172533701 on Build GRCh38
rs960607706 Chr.1: 172533733-172533733 on Build GRCh38
rs879190876 Chr.1: 172533395-172533395 on Build GRCh38
rs745506396 Chr.1: 172533424-172533424 on Build GRCh38
rs887073993 Chr.1: 172533534-172533534 on Build GRCh38
rs770415148 Chr.1: 172533420-172533420 on Build GRCh38
rs984275939 Chr.1: 172533742-172533742 on Build GRCh38
rs945744657 Chr.1: 172533817-172533817 on Build GRCh38
rs931405305 Chr.1: 172533730-172533730 on Build GRCh38
rs769027617 Chr.1: 172533420-172533422 on Build GRCh38
rs544075894 Chr.1: 172533596-172533596 on Build GRCh38
rs904256591 Chr.1: 172533839-172533839 on Build GRCh38
rs539931607 Chr.1: 172533383-172533383 on Build GRCh38
rs1057514062 Chr.1: 172533790-172533790 on Build GRCh38
rs778660682 Chr.1: 172533456-172533456 on Build GRCh38
rs528725796 Chr.1: 172533819-172533819 on Build GRCh38
rs935022433 Chr.1: 172533379-172533379 on Build GRCh38
rs765518027 Chr.1: 172533509-172533511 on Build GRCh38
rs944065510 Chr.1: 172533466-172533466 on Build GRCh38
rs754234840 Chr.1: 172533392-172533392 on Build GRCh38
rs1019610039 Chr.1: 172533678-172533678 on Build GRCh38
rs746508595 Chr.1: 172533413-172533413 on Build GRCh38
rs79939531 Chr.1: 172533557-172533557 on Build GRCh38
rs937601378 Chr.1: 172533541-172533541 on Build GRCh38
rs532753957 Chr.1: 172533628-172533628 on Build GRCh38
rs1044864888 Chr.1: 172533439-172533439 on Build GRCh38
rs946129595 Chr.1: 172533398-172533398 on Build GRCh38
rs972802482 Chr.1: 172533719-172533719 on Build GRCh38
rs755444644 Chr.1: 172533402-172533402 on Build GRCh38
rs113565021 Chr.1: 172533641-172533641 on Build GRCh38
rs766629995 Chr.1: 172533536-172533536 on Build GRCh38
rs947810608 Chr.1: 172533771-172533771 on Build GRCh38
rs918700028 Chr.1: 172533725-172533725 on Build GRCh38
rs757896020 Chr.1: 172533451-172533451 on Build GRCh38

View Full Product Details