Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88910783-88911320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAMBPL1
Entrez Gene ID
57559
Gene Name
STAM binding protein like 1
Gene Aliases
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2
UniGene
Hs.16229
Gene Chromosome Location
Chr.10: 88879368-88923502 on Build GRCh38
Transcripts
NM_020799.3 XM_006717928.2 XM_006717930.3 XM_011539983.1 XM_011539985.1 XM_017016453.1 XM_017016454.1 XM_017016455.1 XM_017016456.1 XM_017016457.1 XM_017016458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5089714 c.420+8C>T p.? Chr.10: 88911019-88911019 on Build GRCh38
4730396 c.370G>A p.D124N Chr.10: 88910961-88910961 on Build GRCh38
1288232 c.369C>T p.N123N Chr.10: 88910960-88910960 on Build GRCh38
6037350 c.390C>T p.N130N Chr.10: 88910981-88910981 on Build GRCh38
4543273 c.334G>A p.E112K Chr.10: 88910925-88910925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003816289 Chr.10: 88910896-88910896 on Build GRCh38
rs766109105 Chr.10: 88910977-88910977 on Build GRCh38
rs912575880 Chr.10: 88910858-88910858 on Build GRCh38
rs939725738 Chr.10: 88911284-88911284 on Build GRCh38
rs748590885 Chr.10: 88910897-88910897 on Build GRCh38
rs746102243 Chr.10: 88910923-88910923 on Build GRCh38
rs774576528 Chr.10: 88910921-88910921 on Build GRCh38
rs372569176 Chr.10: 88911059-88911059 on Build GRCh38
rs776712911 Chr.10: 88910943-88910943 on Build GRCh38
rs572533424 Chr.10: 88910838-88910838 on Build GRCh38
rs966738163 Chr.10: 88911095-88911095 on Build GRCh38
rs562480820 Chr.10: 88911135-88911135 on Build GRCh38
rs765945438 Chr.10: 88911139-88911139 on Build GRCh38
rs577401014 Chr.10: 88911177-88911177 on Build GRCh38
rs540071822 Chr.10: 88910833-88910833 on Build GRCh38
rs780324109 Chr.10: 88910873-88910892 on Build GRCh38
rs190915081 Chr.10: 88911190-88911190 on Build GRCh38
rs778307127 Chr.10: 88910907-88910907 on Build GRCh38
rs746000713 Chr.10: 88911058-88911058 on Build GRCh38
rs146371206 Chr.10: 88911243-88911243 on Build GRCh38
rs771005495 Chr.10: 88910914-88910914 on Build GRCh38
rs746092399 Chr.10: 88911197-88911198 on Build GRCh38
rs544489086 Chr.10: 88911082-88911082 on Build GRCh38
rs995387364 Chr.10: 88910939-88910939 on Build GRCh38
rs17114116 Chr.10: 88911183-88911183 on Build GRCh38
rs955691897 Chr.10: 88911112-88911112 on Build GRCh38
rs756226028 Chr.10: 88910866-88910867 on Build GRCh38
rs911860386 Chr.10: 88911162-88911162 on Build GRCh38
rs978077891 Chr.10: 88911113-88911113 on Build GRCh38
rs575743817 Chr.10: 88911088-88911088 on Build GRCh38
rs57630520 Chr.10: 88910857-88910857 on Build GRCh38
rs756587207 Chr.10: 88910903-88910903 on Build GRCh38
rs752994307 Chr.10: 88910978-88910978 on Build GRCh38
rs757279692 Chr.10: 88911051-88911051 on Build GRCh38
rs527348512 Chr.10: 88911231-88911231 on Build GRCh38
rs1036953391 Chr.10: 88910885-88910885 on Build GRCh38
rs370544295 Chr.10: 88910895-88910895 on Build GRCh38
rs968608874 Chr.10: 88911186-88911186 on Build GRCh38
rs760576374 Chr.10: 88910941-88910941 on Build GRCh38
rs1035319562 Chr.10: 88911098-88911098 on Build GRCh38
rs979647456 Chr.10: 88911188-88911188 on Build GRCh38
rs1048270635 Chr.10: 88910860-88910860 on Build GRCh38
rs148417190 Chr.10: 88911259-88911259 on Build GRCh38
rs888361652 Chr.10: 88911261-88911261 on Build GRCh38
rs749368734 Chr.10: 88910913-88910913 on Build GRCh38
rs201314718 Chr.10: 88910942-88910942 on Build GRCh38
rs997185470 Chr.10: 88910859-88910859 on Build GRCh38
rs750145963 Chr.10: 88910981-88910981 on Build GRCh38
rs764796438 Chr.10: 88910970-88910970 on Build GRCh38
rs377420595 Chr.10: 88910871-88910871 on Build GRCh38
rs750048530 Chr.10: 88910967-88910967 on Build GRCh38
rs1036665719 Chr.10: 88911292-88911292 on Build GRCh38
rs560894196 Chr.10: 88911258-88911258 on Build GRCh38
rs771860979 Chr.10: 88910925-88910925 on Build GRCh38
rs780390305 Chr.10: 88911061-88911061 on Build GRCh38
rs148947892 Chr.10: 88910982-88910982 on Build GRCh38
rs755049 Chr.10: 88911053-88911053 on Build GRCh38
rs1010229652 Chr.10: 88911164-88911164 on Build GRCh38
rs754093370 Chr.10: 88911061-88911068 on Build GRCh38
rs754357845 Chr.10: 88910989-88910989 on Build GRCh38
rs111381530 Chr.10: 88911197-88911197 on Build GRCh38
rs142109544 Chr.10: 88910845-88910845 on Build GRCh38
rs990165747 Chr.10: 88911068-88911068 on Build GRCh38
rs551287791 Chr.10: 88911287-88911287 on Build GRCh38
rs555689928 Chr.10: 88910935-88910935 on Build GRCh38
rs906162182 Chr.10: 88911297-88911297 on Build GRCh38
rs756519248 Chr.10: 88911219-88911219 on Build GRCh38
rs764859212 Chr.10: 88910960-88910960 on Build GRCh38
rs757952991 Chr.10: 88910975-88910975 on Build GRCh38
rs761346323 Chr.10: 88910957-88910957 on Build GRCh38

View Full Product Details