Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101785841-101786411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGEA5
Entrez Gene ID
10724
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
Gene Aliases
MEA5, NCOAT, OGA
UniGene
Hs.500842
Gene Chromosome Location
Chr.10: 101784443-101818709 on Build GRCh38
Transcripts
NM_001142434.1 NM_012215.3 XM_017015585.1 XM_017015586.1

SNP Detail

SNP ID Chromosome Location
rs151060751 Chr.10: 101786055-101786055 on Build GRCh38
rs894876533 Chr.10: 101785999-101785999 on Build GRCh38
rs976409801 Chr.10: 101786015-101786016 on Build GRCh38
rs978756055 Chr.10: 101786281-101786281 on Build GRCh38
rs1042555875 Chr.10: 101786154-101786154 on Build GRCh38
rs1043250 Chr.10: 101785902-101785902 on Build GRCh38
rs142846079 Chr.10: 101785932-101785932 on Build GRCh38
rs112909241 Chr.10: 101786234-101786234 on Build GRCh38
rs1048804378 Chr.10: 101786359-101786359 on Build GRCh38
rs1015503000 Chr.10: 101786003-101786003 on Build GRCh38
rs965329387 Chr.10: 101786013-101786016 on Build GRCh38
rs367586498 Chr.10: 101786224-101786225 on Build GRCh38
rs545306249 Chr.10: 101786368-101786368 on Build GRCh38
rs745717140 Chr.10: 101785942-101785942 on Build GRCh38
rs1006704035 Chr.10: 101786137-101786137 on Build GRCh38
rs953847573 Chr.10: 101786053-101786053 on Build GRCh38
rs1030743653 Chr.10: 101786221-101786221 on Build GRCh38
rs964776797 Chr.10: 101786180-101786180 on Build GRCh38
rs958220312 Chr.10: 101786315-101786315 on Build GRCh38
rs780827489 Chr.10: 101785899-101785899 on Build GRCh38
rs998008913 Chr.10: 101786384-101786384 on Build GRCh38
rs895212956 Chr.10: 101785961-101785961 on Build GRCh38
rs530554255 Chr.10: 101785892-101785892 on Build GRCh38
rs1054813125 Chr.10: 101785994-101785994 on Build GRCh38
rs936805746 Chr.10: 101785991-101785991 on Build GRCh38
rs555413772 Chr.10: 101786179-101786179 on Build GRCh38
rs925442907 Chr.10: 101785972-101785972 on Build GRCh38
rs1046104883 Chr.10: 101786335-101786335 on Build GRCh38
rs1042590275 Chr.10: 101785912-101785912 on Build GRCh38
rs966694386 Chr.10: 101786274-101786274 on Build GRCh38
rs537064858 Chr.10: 101786175-101786175 on Build GRCh38
rs199556668 Chr.10: 101785958-101785958 on Build GRCh38
rs563587543 Chr.10: 101786381-101786381 on Build GRCh38
rs769531298 Chr.10: 101786014-101786015 on Build GRCh38
rs574087195 Chr.10: 101786183-101786183 on Build GRCh38
rs900627110 Chr.10: 101786377-101786377 on Build GRCh38
rs1055087526 Chr.10: 101785960-101785960 on Build GRCh38
rs916362490 Chr.10: 101786322-101786322 on Build GRCh38
rs769331758 Chr.10: 101785965-101785965 on Build GRCh38
rs929625815 Chr.10: 101786343-101786343 on Build GRCh38
rs1049113737 Chr.10: 101786231-101786231 on Build GRCh38
rs1026864664 Chr.10: 101786095-101786095 on Build GRCh38
rs549062242 Chr.10: 101785900-101785900 on Build GRCh38
rs775344629 Chr.10: 101786292-101786292 on Build GRCh38
rs989222201 Chr.10: 101786059-101786059 on Build GRCh38
rs550342520 Chr.10: 101786163-101786163 on Build GRCh38
rs915373205 Chr.10: 101785885-101785885 on Build GRCh38
rs577789273 Chr.10: 101786336-101786336 on Build GRCh38
rs11553298 Chr.10: 101786334-101786334 on Build GRCh38
rs945553554 Chr.10: 101786113-101786113 on Build GRCh38
rs77030441 Chr.10: 101786325-101786325 on Build GRCh38
rs990931335 Chr.10: 101786320-101786320 on Build GRCh38
rs776977467 Chr.10: 101785880-101785881 on Build GRCh38
rs994488646 Chr.10: 101786088-101786088 on Build GRCh38
rs762711005 Chr.10: 101786385-101786385 on Build GRCh38
rs1006756315 Chr.10: 101786369-101786369 on Build GRCh38
rs888288916 Chr.10: 101786129-101786129 on Build GRCh38
rs997560113 Chr.10: 101786196-101786196 on Build GRCh38
rs771620933 Chr.10: 101786307-101786307 on Build GRCh38
rs535630453 Chr.10: 101786018-101786018 on Build GRCh38
rs1004047678 Chr.10: 101786236-101786236 on Build GRCh38
rs1015471887 Chr.10: 101786291-101786291 on Build GRCh38
rs184831697 Chr.10: 101786052-101786052 on Build GRCh38
rs139721047 Chr.10: 101786065-101786065 on Build GRCh38
rs895105973 Chr.10: 101786203-101786203 on Build GRCh38
rs1053956863 Chr.10: 101785874-101785874 on Build GRCh38
rs187050831 Chr.10: 101786161-101786161 on Build GRCh38
rs541206168 Chr.10: 101786186-101786186 on Build GRCh38
rs1028154062 Chr.10: 101785983-101785983 on Build GRCh38
rs867571683 Chr.10: 101786370-101786370 on Build GRCh38
rs558058517 Chr.10: 101786120-101786120 on Build GRCh38

View Full Product Details