Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 790180-790760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A22
Entrez Gene ID
79751
Gene Name
solute carrier family 25 member 22
Gene Aliases
EIEE3, GC-1, GC1, NET44
UniGene
Hs.99486
Gene Chromosome Location
Chr.11: 790475-798269 on Build GRCh38
Transcripts
NM_001191060.1 NM_001191061.1 NM_024698.5 XM_011520369.1 XM_011520370.1 XM_011520371.1

SNP Detail

SNP ID Chromosome Location
rs946331878 Chr.11: 790247-790247 on Build GRCh38
rs34592943 Chr.11: 790684-790685 on Build GRCh38
rs1018266411 Chr.11: 790210-790210 on Build GRCh38
rs550630361 Chr.11: 790353-790353 on Build GRCh38
rs1048897855 Chr.11: 790501-790505 on Build GRCh38
rs4963154 Chr.11: 790253-790253 on Build GRCh38
rs879908582 Chr.11: 790378-790378 on Build GRCh38
rs548785686 Chr.11: 790517-790517 on Build GRCh38
rs971421784 Chr.11: 790525-790527 on Build GRCh38
rs1025914465 Chr.11: 790590-790590 on Build GRCh38
rs547695882 Chr.11: 790670-790670 on Build GRCh38
rs893255607 Chr.11: 790512-790512 on Build GRCh38
rs1014861898 Chr.11: 790362-790362 on Build GRCh38
rs367812123 Chr.11: 790576-790576 on Build GRCh38
rs1016422308 Chr.11: 790629-790629 on Build GRCh38
rs984968160 Chr.11: 790328-790328 on Build GRCh38
rs893297185 Chr.11: 790206-790206 on Build GRCh38
rs998952221 Chr.11: 790200-790200 on Build GRCh38
rs1008137803 Chr.11: 790513-790513 on Build GRCh38
rs535428182 Chr.11: 790298-790298 on Build GRCh38
rs577916880 Chr.11: 790376-790376 on Build GRCh38
rs887605460 Chr.11: 790720-790720 on Build GRCh38
rs923621537 Chr.11: 790463-790463 on Build GRCh38
rs146349187 Chr.11: 790372-790372 on Build GRCh38
rs1019804049 Chr.11: 790442-790443 on Build GRCh38
rs966306800 Chr.11: 790239-790239 on Build GRCh38
rs917820557 Chr.11: 790417-790417 on Build GRCh38
rs559349713 Chr.11: 790536-790536 on Build GRCh38
rs539795191 Chr.11: 790234-790253 on Build GRCh38
rs35813865 Chr.11: 790477-790478 on Build GRCh38
rs575292705 Chr.11: 790614-790614 on Build GRCh38
rs970608597 Chr.11: 790731-790731 on Build GRCh38
rs1033157741 Chr.11: 790203-790203 on Build GRCh38
rs893396010 Chr.11: 790240-790240 on Build GRCh38
rs975703387 Chr.11: 790272-790272 on Build GRCh38
rs887633301 Chr.11: 790502-790502 on Build GRCh38
rs755572915 Chr.11: 790486-790489 on Build GRCh38
rs934693721 Chr.11: 790442-790442 on Build GRCh38
rs541377254 Chr.11: 790320-790320 on Build GRCh38
rs983391760 Chr.11: 790352-790352 on Build GRCh38
rs903201815 Chr.11: 790437-790437 on Build GRCh38
rs529018067 Chr.11: 790484-790484 on Build GRCh38
rs185516425 Chr.11: 790384-790384 on Build GRCh38
rs944409908 Chr.11: 790483-790483 on Build GRCh38
rs937542565 Chr.11: 790231-790231 on Build GRCh38
rs74573437 Chr.11: 790418-790418 on Build GRCh38
rs1040455145 Chr.11: 790640-790640 on Build GRCh38
rs1042020297 Chr.11: 790459-790462 on Build GRCh38
rs927411213 Chr.11: 790391-790391 on Build GRCh38
rs755953694 Chr.11: 790428-790428 on Build GRCh38
rs909009450 Chr.11: 790335-790335 on Build GRCh38
rs1025966713 Chr.11: 790293-790293 on Build GRCh38
rs912808376 Chr.11: 790637-790637 on Build GRCh38
rs570780598 Chr.11: 790671-790671 on Build GRCh38
rs1003290288 Chr.11: 790558-790558 on Build GRCh38
rs900254990 Chr.11: 790485-790488 on Build GRCh38
rs191455128 Chr.11: 790638-790638 on Build GRCh38
rs572345972 Chr.11: 790242-790242 on Build GRCh38
rs990705887 Chr.11: 790415-790415 on Build GRCh38
rs990294259 Chr.11: 790608-790608 on Build GRCh38
rs950392937 Chr.11: 790603-790603 on Build GRCh38
rs1048825159 Chr.11: 790678-790678 on Build GRCh38
rs1042089687 Chr.11: 790258-790258 on Build GRCh38
rs779716143 Chr.11: 790538-790538 on Build GRCh38
rs1007777189 Chr.11: 790207-790207 on Build GRCh38
rs1008634391 Chr.11: 790288-790288 on Build GRCh38
rs977666014 Chr.11: 790632-790632 on Build GRCh38
rs951975222 Chr.11: 790350-790350 on Build GRCh38
rs753154899 Chr.11: 790333-790334 on Build GRCh38
rs927472334 Chr.11: 790201-790201 on Build GRCh38
rs887098012 Chr.11: 790334-790348 on Build GRCh38
rs991045447 Chr.11: 790657-790657 on Build GRCh38
rs1004459287 Chr.11: 790348-790362 on Build GRCh38
rs902092008 Chr.11: 790578-790578 on Build GRCh38
rs997738405 Chr.11: 790586-790586 on Build GRCh38

View Full Product Details