Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67633920-67634487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX10
Entrez Gene ID
347853
Gene Name
T-box 10
Gene Aliases
TBX13, TBX7
UniGene
Hs.454480
Gene Chromosome Location
Chr.11: 67631303-67639560 on Build GRCh38
Transcripts
NM_005995.4 XM_011544992.2 XM_011544993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130974 c.549+1G>A p.? Chr.11: 67634188-67634188 on Build GRCh38
6346293 c.525C>A p.N175K Chr.11: 67634213-67634213 on Build GRCh38
931148 c.457G>A p.D153N Chr.11: 67634281-67634281 on Build GRCh38
1704301 c.421G>A p.D141N Chr.11: 67634317-67634317 on Build GRCh38
931147 c.538G>A p.D180N Chr.11: 67634200-67634200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048415860 Chr.11: 67634020-67634020 on Build GRCh38
rs151337371 Chr.11: 67634250-67634250 on Build GRCh38
rs528938618 Chr.11: 67633956-67633956 on Build GRCh38
rs530787158 Chr.11: 67634452-67634452 on Build GRCh38
rs143361816 Chr.11: 67634063-67634063 on Build GRCh38
rs754069888 Chr.11: 67634227-67634227 on Build GRCh38
rs772669171 Chr.11: 67634171-67634171 on Build GRCh38
rs925727900 Chr.11: 67633942-67633942 on Build GRCh38
rs910129756 Chr.11: 67634459-67634459 on Build GRCh38
rs756186418 Chr.11: 67634205-67634205 on Build GRCh38
rs754563145 Chr.11: 67633963-67633963 on Build GRCh38
rs1012407473 Chr.11: 67634124-67634124 on Build GRCh38
rs535076569 Chr.11: 67634121-67634121 on Build GRCh38
rs528719595 Chr.11: 67634450-67634450 on Build GRCh38
rs369297491 Chr.11: 67634354-67634354 on Build GRCh38
rs372763899 Chr.11: 67634298-67634298 on Build GRCh38
rs1002230878 Chr.11: 67633986-67633986 on Build GRCh38
rs573671280 Chr.11: 67634303-67634303 on Build GRCh38
rs774083949 Chr.11: 67634363-67634363 on Build GRCh38
rs955694281 Chr.11: 67634135-67634135 on Build GRCh38
rs914753077 Chr.11: 67634285-67634285 on Build GRCh38
rs781736938 Chr.11: 67634342-67634342 on Build GRCh38
rs745402649 Chr.11: 67634279-67634279 on Build GRCh38
rs745305858 Chr.11: 67634343-67634343 on Build GRCh38
rs762726021 Chr.11: 67634283-67634283 on Build GRCh38
rs879372181 Chr.11: 67633993-67633993 on Build GRCh38
rs887637769 Chr.11: 67634378-67634378 on Build GRCh38
rs117709265 Chr.11: 67634189-67634189 on Build GRCh38
rs544794599 Chr.11: 67634443-67634443 on Build GRCh38
rs1047121575 Chr.11: 67633966-67633966 on Build GRCh38
rs373969860 Chr.11: 67634173-67634173 on Build GRCh38
rs142569144 Chr.11: 67634235-67634235 on Build GRCh38
rs940056070 Chr.11: 67634466-67634466 on Build GRCh38
rs888421826 Chr.11: 67634054-67634054 on Build GRCh38
rs750241409 Chr.11: 67634142-67634142 on Build GRCh38
rs753024376 Chr.11: 67634326-67634326 on Build GRCh38
rs200376968 Chr.11: 67634330-67634330 on Build GRCh38
rs775096048 Chr.11: 67634188-67634188 on Build GRCh38
rs755090677 Chr.11: 67634310-67634310 on Build GRCh38
rs779616885 Chr.11: 67634297-67634298 on Build GRCh38
rs751557584 Chr.11: 67634203-67634203 on Build GRCh38
rs1028235004 Chr.11: 67634075-67634075 on Build GRCh38
rs1011865322 Chr.11: 67634060-67634060 on Build GRCh38
rs755899386 Chr.11: 67634157-67634157 on Build GRCh38
rs748547632 Chr.11: 67634170-67634170 on Build GRCh38
rs975992253 Chr.11: 67634429-67634429 on Build GRCh38
rs755105995 Chr.11: 67634231-67634231 on Build GRCh38
rs12787511 Chr.11: 67634164-67634164 on Build GRCh38
rs116791834 Chr.11: 67633965-67633965 on Build GRCh38
rs778321159 Chr.11: 67634328-67634328 on Build GRCh38
rs952038618 Chr.11: 67633945-67633945 on Build GRCh38
rs114440123 Chr.11: 67634085-67634085 on Build GRCh38
rs569600737 Chr.11: 67634070-67634070 on Build GRCh38
rs533081637 Chr.11: 67634037-67634037 on Build GRCh38
rs747586701 Chr.11: 67634263-67634263 on Build GRCh38
rs750460059 Chr.11: 67634157-67634157 on Build GRCh38
rs1004720952 Chr.11: 67634055-67634055 on Build GRCh38
rs778409069 Chr.11: 67634257-67634257 on Build GRCh38
rs748984001 Chr.11: 67634355-67634355 on Build GRCh38
rs769289032 Chr.11: 67634281-67634281 on Build GRCh38
rs775125691 Chr.11: 67634282-67634282 on Build GRCh38
rs200089316 Chr.11: 67634251-67634251 on Build GRCh38
rs115181274 Chr.11: 67634159-67634159 on Build GRCh38
rs11227873 Chr.11: 67634258-67634258 on Build GRCh38
rs150470556 Chr.11: 67634360-67634360 on Build GRCh38
rs1000072502 Chr.11: 67634329-67634329 on Build GRCh38
rs767513849 Chr.11: 67634305-67634305 on Build GRCh38
rs773216675 Chr.11: 67634375-67634375 on Build GRCh38
rs953945815 Chr.11: 67634078-67634078 on Build GRCh38
rs142397323 Chr.11: 67634244-67634244 on Build GRCh38
rs7102811 Chr.11: 67633947-67633947 on Build GRCh38
rs753822108 Chr.11: 67634307-67634307 on Build GRCh38
rs765317926 Chr.11: 67634325-67634325 on Build GRCh38
rs971156477 Chr.11: 67634423-67634423 on Build GRCh38
rs776341108 Chr.11: 67634180-67634180 on Build GRCh38
rs770442625 Chr.11: 67634267-67634267 on Build GRCh38
rs1037555333 Chr.11: 67634252-67634252 on Build GRCh38
rs866905136 Chr.11: 67634277-67634277 on Build GRCh38
rs765054943 Chr.11: 67634182-67634182 on Build GRCh38
rs756202843 Chr.11: 67634158-67634158 on Build GRCh38
rs749543640 Chr.11: 67634163-67634163 on Build GRCh38
rs549356010 Chr.11: 67634074-67634074 on Build GRCh38
rs1034636324 Chr.11: 67634112-67634112 on Build GRCh38
rs926794665 Chr.11: 67634435-67634435 on Build GRCh38
rs778605304 Chr.11: 67633984-67633984 on Build GRCh38
rs1048651238 Chr.11: 67634238-67634238 on Build GRCh38
rs140602591 Chr.11: 67634348-67634348 on Build GRCh38
rs182068227 Chr.11: 67634181-67634181 on Build GRCh38
rs376354468 Chr.11: 67634374-67634374 on Build GRCh38
rs375008730 Chr.11: 67634172-67634172 on Build GRCh38
rs766413146 Chr.11: 67634206-67634206 on Build GRCh38
rs368290332 Chr.11: 67634407-67634407 on Build GRCh38
rs759518912 Chr.11: 67634406-67634406 on Build GRCh38
rs11227872 Chr.11: 67633957-67633957 on Build GRCh38
rs530850469 Chr.11: 67634071-67634071 on Build GRCh38
rs763964938 Chr.11: 67634194-67634194 on Build GRCh38
rs938124131 Chr.11: 67634445-67634445 on Build GRCh38
rs770622855 Chr.11: 67634177-67634177 on Build GRCh38
rs752645353 Chr.11: 67634139-67634139 on Build GRCh38
rs553618400 Chr.11: 67634154-67634154 on Build GRCh38
rs777707408 Chr.11: 67634156-67634156 on Build GRCh38
rs961976966 Chr.11: 67634428-67634428 on Build GRCh38
rs757652441 Chr.11: 67634337-67634337 on Build GRCh38
rs761700620 Chr.11: 67634365-67634365 on Build GRCh38
rs763946525 Chr.11: 67634296-67634296 on Build GRCh38
rs979817307 Chr.11: 67634433-67634433 on Build GRCh38
rs776051019 Chr.11: 67634275-67634275 on Build GRCh38
rs932043879 Chr.11: 67634016-67634016 on Build GRCh38
rs151295746 Chr.11: 67634327-67634327 on Build GRCh38

View Full Product Details