Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40929648-40930236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN1
Entrez Gene ID
1272
Gene Name
contactin 1
Gene Aliases
F3, GP135, MYPCN
UniGene
Hs.143434
Gene Chromosome Location
Chr.12: 40692439-41072412 on Build GRCh38
Transcripts
NM_001256063.1 NM_001256064.1 NM_001843.3 NM_175038.2 XM_005268651.2 XM_006719241.2 XM_011537926.2 XM_011537927.2 XM_017018821.1 XM_017018822.1 XM_017018823.1 XM_017018824.1 XM_017018825.1 XM_017018826.1 XM_017018827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986908 c.659A>G p.K220R Chr.12: 40929958-40929958 on Build GRCh38
6569545 c.700G>A p.E234K Chr.12: 40929999-40929999 on Build GRCh38
5557749 c.555A>G p.K185K Chr.12: 40929854-40929854 on Build GRCh38
4903480 c.641C>T p.S214F Chr.12: 40929940-40929940 on Build GRCh38
1361481 c.682C>A p.P228T Chr.12: 40929981-40929981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757382046 Chr.12: 40929747-40929747 on Build GRCh38
rs138193121 Chr.12: 40929673-40929673 on Build GRCh38
rs192664798 Chr.12: 40929823-40929823 on Build GRCh38
rs750578149 Chr.12: 40929885-40929885 on Build GRCh38
rs772846701 Chr.12: 40929792-40929792 on Build GRCh38
rs755054882 Chr.12: 40929912-40929912 on Build GRCh38
rs975167072 Chr.12: 40930149-40930149 on Build GRCh38
rs770683180 Chr.12: 40929810-40929810 on Build GRCh38
rs138639141 Chr.12: 40929942-40929942 on Build GRCh38
rs1031993188 Chr.12: 40930108-40930108 on Build GRCh38
rs545323042 Chr.12: 40930063-40930063 on Build GRCh38
rs999620339 Chr.12: 40930062-40930062 on Build GRCh38
rs71449788 Chr.12: 40929684-40929684 on Build GRCh38
rs775149768 Chr.12: 40929863-40929863 on Build GRCh38
rs375891152 Chr.12: 40929758-40929758 on Build GRCh38
rs370237035 Chr.12: 40929774-40929774 on Build GRCh38
rs368372193 Chr.12: 40930042-40930042 on Build GRCh38
rs1032482987 Chr.12: 40929780-40929780 on Build GRCh38
rs972742579 Chr.12: 40929993-40929993 on Build GRCh38
rs963764393 Chr.12: 40930134-40930134 on Build GRCh38
rs755987043 Chr.12: 40929992-40929992 on Build GRCh38
rs769861119 Chr.12: 40929763-40929763 on Build GRCh38
rs1019264144 Chr.12: 40930116-40930116 on Build GRCh38
rs1023481610 Chr.12: 40930195-40930195 on Build GRCh38
rs189031520 Chr.12: 40930198-40930198 on Build GRCh38
rs1007882148 Chr.12: 40930107-40930107 on Build GRCh38
rs777748960 Chr.12: 40929922-40929922 on Build GRCh38
rs4408370 Chr.12: 40929859-40929859 on Build GRCh38
rs767936676 Chr.12: 40929882-40929882 on Build GRCh38
rs369589652 Chr.12: 40930170-40930170 on Build GRCh38
rs559563371 Chr.12: 40930023-40930023 on Build GRCh38
rs868602241 Chr.12: 40929977-40929977 on Build GRCh38
rs201037986 Chr.12: 40929811-40929811 on Build GRCh38
rs968249698 Chr.12: 40930175-40930175 on Build GRCh38
rs886744446 Chr.12: 40930067-40930067 on Build GRCh38
rs757271845 Chr.12: 40930039-40930039 on Build GRCh38
rs898413005 Chr.12: 40929689-40929689 on Build GRCh38
rs759325815 Chr.12: 40929847-40929847 on Build GRCh38
rs775185622 Chr.12: 40930049-40930049 on Build GRCh38
rs745456807 Chr.12: 40929963-40929963 on Build GRCh38
rs1023466413 Chr.12: 40929884-40929884 on Build GRCh38
rs756344032 Chr.12: 40929910-40929910 on Build GRCh38
rs1056506124 Chr.12: 40929991-40929991 on Build GRCh38
rs182672697 Chr.12: 40930019-40930019 on Build GRCh38
rs762232466 Chr.12: 40930009-40930009 on Build GRCh38
rs746136701 Chr.12: 40929761-40929761 on Build GRCh38
rs769590342 Chr.12: 40929982-40929982 on Build GRCh38
rs970520312 Chr.12: 40930196-40930196 on Build GRCh38
rs575968419 Chr.12: 40929796-40929796 on Build GRCh38
rs1008430726 Chr.12: 40929719-40929719 on Build GRCh38
rs17622354 Chr.12: 40929678-40929678 on Build GRCh38
rs138266018 Chr.12: 40929891-40929891 on Build GRCh38
rs780114224 Chr.12: 40929769-40929769 on Build GRCh38
rs146088900 Chr.12: 40929911-40929911 on Build GRCh38
rs751975420 Chr.12: 40929746-40929746 on Build GRCh38
rs142799954 Chr.12: 40929779-40929779 on Build GRCh38
rs774907531 Chr.12: 40930002-40930002 on Build GRCh38
rs745687222 Chr.12: 40930046-40930046 on Build GRCh38
rs746981200 Chr.12: 40929932-40929932 on Build GRCh38
rs769749210 Chr.12: 40930048-40930048 on Build GRCh38
rs370940612 Chr.12: 40930024-40930024 on Build GRCh38
rs1019239823 Chr.12: 40929749-40929749 on Build GRCh38
rs995468192 Chr.12: 40929704-40929704 on Build GRCh38
rs763560467 Chr.12: 40929994-40929994 on Build GRCh38
rs761319223 Chr.12: 40930038-40930039 on Build GRCh38
rs752293954 Chr.12: 40929951-40929951 on Build GRCh38
rs757618456 Chr.12: 40929874-40929874 on Build GRCh38
rs371168812 Chr.12: 40930033-40930033 on Build GRCh38
rs1025576721 Chr.12: 40930064-40930064 on Build GRCh38
rs767811479 Chr.12: 40930016-40930016 on Build GRCh38
rs982045496 Chr.12: 40930208-40930208 on Build GRCh38
rs563673101 Chr.12: 40930123-40930123 on Build GRCh38
rs375395772 Chr.12: 40930034-40930034 on Build GRCh38
rs896552364 Chr.12: 40930018-40930018 on Build GRCh38
rs752046046 Chr.12: 40929872-40929872 on Build GRCh38
rs775098655 Chr.12: 40929984-40929984 on Build GRCh38
rs748951560 Chr.12: 40930051-40930051 on Build GRCh38

View Full Product Details