Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56781843-56782433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B6
Entrez Gene ID
8630
Gene Name
hydroxysteroid 17-beta dehydrogenase 6
Gene Aliases
HSE, RODH, SDR9C6
UniGene
Hs.524513
Gene Chromosome Location
Chr.12: 56752449-56787790 on Build GRCh38
Transcripts
NM_003725.3 XM_005269207.1 XM_005269208.1 XM_005269209.1 XM_006719672.1 XM_011538925.1 XM_011538926.1 XM_011538927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6682380 c.498G>A p.L166L Chr.12: 56782158-56782158 on Build GRCh38
5473413 c.362T>C p.L121S Chr.12: 56782022-56782022 on Build GRCh38
5532434 c.424A>C p.I142L Chr.12: 56782084-56782084 on Build GRCh38
401355 c.314-1G>T p.? Chr.12: 56781973-56781973 on Build GRCh38
1987940 c.388T>C p.S130P Chr.12: 56782048-56782048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550777376 Chr.12: 56782303-56782303 on Build GRCh38
rs759667910 Chr.12: 56782194-56782194 on Build GRCh38
rs752975801 Chr.12: 56782213-56782213 on Build GRCh38
rs779625816 Chr.12: 56782118-56782118 on Build GRCh38
rs961748369 Chr.12: 56781905-56781905 on Build GRCh38
rs760414708 Chr.12: 56781928-56781928 on Build GRCh38
rs566827869 Chr.12: 56782394-56782394 on Build GRCh38
rs781402793 Chr.12: 56782053-56782053 on Build GRCh38
rs910491908 Chr.12: 56782048-56782048 on Build GRCh38
rs148650187 Chr.12: 56782156-56782156 on Build GRCh38
rs767833118 Chr.12: 56782201-56782201 on Build GRCh38
rs928779752 Chr.12: 56782223-56782223 on Build GRCh38
rs71459389 Chr.12: 56782161-56782161 on Build GRCh38
rs3816616 Chr.12: 56782305-56782305 on Build GRCh38
rs776268123 Chr.12: 56782126-56782126 on Build GRCh38
rs201697172 Chr.12: 56782061-56782061 on Build GRCh38
rs10876921 Chr.12: 56782402-56782402 on Build GRCh38
rs998650366 Chr.12: 56782276-56782276 on Build GRCh38
rs997144394 Chr.12: 56782304-56782304 on Build GRCh38
rs972238825 Chr.12: 56782378-56782378 on Build GRCh38
rs181132444 Chr.12: 56782315-56782315 on Build GRCh38
rs890286252 Chr.12: 56782324-56782324 on Build GRCh38
rs566315767 Chr.12: 56782136-56782136 on Build GRCh38
rs560467329 Chr.12: 56782265-56782265 on Build GRCh38
rs529425917 Chr.12: 56782291-56782291 on Build GRCh38
rs768423788 Chr.12: 56782123-56782123 on Build GRCh38
rs376817921 Chr.12: 56782059-56782059 on Build GRCh38
rs767345332 Chr.12: 56781896-56781903 on Build GRCh38
rs781198023 Chr.12: 56782249-56782249 on Build GRCh38
rs987984178 Chr.12: 56781899-56781899 on Build GRCh38
rs778387732 Chr.12: 56782102-56782102 on Build GRCh38
rs756535253 Chr.12: 56782220-56782220 on Build GRCh38
rs757839484 Chr.12: 56782251-56782251 on Build GRCh38
rs751704162 Chr.12: 56782119-56782119 on Build GRCh38
rs953507085 Chr.12: 56781865-56781865 on Build GRCh38
rs771594241 Chr.12: 56782112-56782112 on Build GRCh38
rs954705904 Chr.12: 56781951-56781951 on Build GRCh38
rs113673994 Chr.12: 56782244-56782244 on Build GRCh38
rs1045922619 Chr.12: 56782255-56782255 on Build GRCh38
rs773213382 Chr.12: 56782280-56782280 on Build GRCh38
rs1032775460 Chr.12: 56782299-56782299 on Build GRCh38
rs989409932 Chr.12: 56782357-56782357 on Build GRCh38
rs757143511 Chr.12: 56782134-56782134 on Build GRCh38
rs766567845 Chr.12: 56782173-56782173 on Build GRCh38
rs747286653 Chr.12: 56781938-56781939 on Build GRCh38
rs373512375 Chr.12: 56782089-56782089 on Build GRCh38
rs761708785 Chr.12: 56782127-56782127 on Build GRCh38
rs766656110 Chr.12: 56782010-56782010 on Build GRCh38
rs577783084 Chr.12: 56782160-56782160 on Build GRCh38
rs149472960 Chr.12: 56781985-56781985 on Build GRCh38
rs1008893040 Chr.12: 56782341-56782341 on Build GRCh38
rs751841717 Chr.12: 56782020-56782020 on Build GRCh38
rs10876920 Chr.12: 56782391-56782391 on Build GRCh38
rs140840945 Chr.12: 56782273-56782273 on Build GRCh38
rs750497603 Chr.12: 56781990-56781990 on Build GRCh38
rs781121528 Chr.12: 56782163-56782163 on Build GRCh38
rs754357466 Chr.12: 56782246-56782246 on Build GRCh38
rs942307424 Chr.12: 56782287-56782287 on Build GRCh38
rs143952608 Chr.12: 56782096-56782096 on Build GRCh38
rs373290486 Chr.12: 56781955-56781955 on Build GRCh38
rs543955848 Chr.12: 56782031-56782031 on Build GRCh38
rs938883381 Chr.12: 56782264-56782264 on Build GRCh38
rs972186468 Chr.12: 56781956-56781956 on Build GRCh38
rs371026321 Chr.12: 56782075-56782075 on Build GRCh38
rs764537715 Chr.12: 56782240-56782240 on Build GRCh38
rs768754876 Chr.12: 56782110-56782110 on Build GRCh38
rs183689283 Chr.12: 56781906-56781906 on Build GRCh38
rs527535183 Chr.12: 56782267-56782267 on Build GRCh38
rs962086768 Chr.12: 56782354-56782354 on Build GRCh38
rs761641999 Chr.12: 56781976-56781976 on Build GRCh38
rs751751752 Chr.12: 56782188-56782188 on Build GRCh38
rs930853688 Chr.12: 56782076-56782076 on Build GRCh38
rs780928315 Chr.12: 56782268-56782268 on Build GRCh38
rs371459084 Chr.12: 56782277-56782277 on Build GRCh38
rs1007831938 Chr.12: 56782339-56782339 on Build GRCh38
rs745344854 Chr.12: 56782108-56782108 on Build GRCh38
rs776565720 Chr.12: 56781961-56781961 on Build GRCh38
rs376554459 Chr.12: 56781925-56781925 on Build GRCh38
rs545151902 Chr.12: 56782252-56782252 on Build GRCh38
rs189527846 Chr.12: 56781924-56781924 on Build GRCh38
rs1037302743 Chr.12: 56782295-56782295 on Build GRCh38
rs574635067 Chr.12: 56782323-56782323 on Build GRCh38
rs763066532 Chr.12: 56782162-56782162 on Build GRCh38
rs1019448827 Chr.12: 56782346-56782346 on Build GRCh38

View Full Product Details