Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40069561-40070132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAP1
Entrez Gene ID
10487
Gene Name
adenylate cyclase associated protein 1
Gene Aliases
CAP, CAP1-PEN
UniGene
Hs.370581
Gene Chromosome Location
Chr.1: 40040070-40072649 on Build GRCh38
Transcripts
NM_001105530.1 NM_006367.3 XM_005270368.1 XM_011540509.1 XM_011540510.1 XM_011540511.1 XM_011540512.1 XM_011540513.2 XM_011540514.1 XM_011540515.1 XM_017000067.1 XM_017000068.1 XM_017000069.1 XM_017000070.1 XM_017000071.1 XM_017000072.1 XM_017000073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909374 c.916C>A p.Q306K Chr.1: 40069797-40069797 on Build GRCh38
6888560 c.926C>T p.P309L Chr.1: 40069807-40069807 on Build GRCh38
7167199 c.829G>C p.D277H Chr.1: 40069710-40069710 on Build GRCh38
909376 c.975G>T p.E325D Chr.1: 40069856-40069856 on Build GRCh38
909375 c.938G>A p.R313Q Chr.1: 40069819-40069819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370362556 Chr.1: 40069818-40069819 on Build GRCh38
rs925063850 Chr.1: 40069631-40069631 on Build GRCh38
rs1048081226 Chr.1: 40070082-40070082 on Build GRCh38
rs755264509 Chr.1: 40069809-40069809 on Build GRCh38
rs367744502 Chr.1: 40069817-40069818 on Build GRCh38
rs529899984 Chr.1: 40069896-40069896 on Build GRCh38
rs765081992 Chr.1: 40069953-40069953 on Build GRCh38
rs187715440 Chr.1: 40069724-40069724 on Build GRCh38
rs776355240 Chr.1: 40069777-40069800 on Build GRCh38
rs751754089 Chr.1: 40069802-40069804 on Build GRCh38
rs774492587 Chr.1: 40069821-40069821 on Build GRCh38
rs373976739 Chr.1: 40069754-40069754 on Build GRCh38
rs757940618 Chr.1: 40069901-40069901 on Build GRCh38
rs767150422 Chr.1: 40069802-40069802 on Build GRCh38
rs767403475 Chr.1: 40069718-40069718 on Build GRCh38
rs762444407 Chr.1: 40069683-40069683 on Build GRCh38
rs201835889 Chr.1: 40069690-40069690 on Build GRCh38
rs866564567 Chr.1: 40070088-40070088 on Build GRCh38
rs761794847 Chr.1: 40069789-40069804 on Build GRCh38
rs375382486 Chr.1: 40069814-40069815 on Build GRCh38
rs543606897 Chr.1: 40069649-40069649 on Build GRCh38
rs774700059 Chr.1: 40069793-40069793 on Build GRCh38
rs61781910 Chr.1: 40069667-40069667 on Build GRCh38
rs781671919 Chr.1: 40069755-40069755 on Build GRCh38
rs369366604 Chr.1: 40069646-40069646 on Build GRCh38
rs773430808 Chr.1: 40069703-40069703 on Build GRCh38
rs766731752 Chr.1: 40069714-40069714 on Build GRCh38
rs763209951 Chr.1: 40069711-40069711 on Build GRCh38
rs985172754 Chr.1: 40069669-40069669 on Build GRCh38
rs375499575 Chr.1: 40069797-40069798 on Build GRCh38
rs555248399 Chr.1: 40070025-40070025 on Build GRCh38
rs781354043 Chr.1: 40069817-40069817 on Build GRCh38
rs746654749 Chr.1: 40069896-40069897 on Build GRCh38
rs764046970 Chr.1: 40069808-40069808 on Build GRCh38
rs140053828 Chr.1: 40070071-40070071 on Build GRCh38
rs772357523 Chr.1: 40069842-40069842 on Build GRCh38
rs768478870 Chr.1: 40069920-40069921 on Build GRCh38
rs1026261572 Chr.1: 40069766-40069766 on Build GRCh38
rs534707694 Chr.1: 40070013-40070013 on Build GRCh38
rs936129170 Chr.1: 40069986-40069986 on Build GRCh38
rs924907626 Chr.1: 40069982-40069982 on Build GRCh38
rs753810830 Chr.1: 40069809-40069809 on Build GRCh38
rs779751057 Chr.1: 40069827-40069829 on Build GRCh38
rs372682081 Chr.1: 40069810-40069810 on Build GRCh38
rs766465759 Chr.1: 40069802-40069809 on Build GRCh38
rs761808792 Chr.1: 40069894-40069894 on Build GRCh38
rs541465362 Chr.1: 40069763-40069763 on Build GRCh38
rs575025085 Chr.1: 40069636-40069636 on Build GRCh38
rs763928323 Chr.1: 40069889-40069889 on Build GRCh38
rs200574479 Chr.1: 40069691-40069691 on Build GRCh38
rs967500090 Chr.1: 40069606-40069606 on Build GRCh38
rs11556939 Chr.1: 40069759-40069759 on Build GRCh38
rs887701495 Chr.1: 40070083-40070083 on Build GRCh38
rs142100140 Chr.1: 40070002-40070002 on Build GRCh38
rs760164772 Chr.1: 40069643-40069643 on Build GRCh38
rs372384097 Chr.1: 40069936-40069936 on Build GRCh38
rs142186369 Chr.1: 40069995-40069995 on Build GRCh38
rs4660157 Chr.1: 40069973-40069973 on Build GRCh38
rs750043474 Chr.1: 40069814-40069814 on Build GRCh38
rs147610875 Chr.1: 40069949-40069949 on Build GRCh38
rs544011394 Chr.1: 40070095-40070095 on Build GRCh38
rs924755798 Chr.1: 40069619-40069619 on Build GRCh38
rs201677980 Chr.1: 40069786-40069786 on Build GRCh38
rs759615015 Chr.1: 40069825-40069825 on Build GRCh38
rs769108541 Chr.1: 40069660-40069660 on Build GRCh38
rs527275009 Chr.1: 40069736-40069736 on Build GRCh38
rs752571338 Chr.1: 40069720-40069720 on Build GRCh38
rs562185237 Chr.1: 40069630-40069630 on Build GRCh38
rs545043783 Chr.1: 40069717-40069717 on Build GRCh38
rs773379414 Chr.1: 40069767-40069767 on Build GRCh38
rs768539489 Chr.1: 40069773-40069773 on Build GRCh38
rs910758772 Chr.1: 40069997-40069997 on Build GRCh38
rs200369781 Chr.1: 40069818-40069818 on Build GRCh38
rs762977570 Chr.1: 40069801-40069804 on Build GRCh38
rs779788662 Chr.1: 40069904-40069904 on Build GRCh38
rs377554241 Chr.1: 40069819-40069819 on Build GRCh38
rs866623441 Chr.1: 40069586-40069586 on Build GRCh38
rs201057661 Chr.1: 40069890-40069890 on Build GRCh38
rs771182811 Chr.1: 40069788-40069788 on Build GRCh38
rs768792606 Chr.1: 40069920-40069920 on Build GRCh38
rs74068028 Chr.1: 40070003-40070003 on Build GRCh38
rs760428908 Chr.1: 40069884-40069884 on Build GRCh38
rs35749351 Chr.1: 40069762-40069762 on Build GRCh38
rs773202461 Chr.1: 40069800-40069801 on Build GRCh38
rs775104460 Chr.1: 40069848-40069848 on Build GRCh38
rs749964373 Chr.1: 40069900-40069900 on Build GRCh38
rs748537032 Chr.1: 40069761-40069761 on Build GRCh38
rs759869293 Chr.1: 40069796-40069796 on Build GRCh38
rs765023868 Chr.1: 40069798-40069799 on Build GRCh38
rs567443790 Chr.1: 40069800-40069800 on Build GRCh38
rs752547029 Chr.1: 40069804-40069804 on Build GRCh38
rs756078402 Chr.1: 40069722-40069722 on Build GRCh38
rs561415904 Chr.1: 40069587-40069587 on Build GRCh38
rs758727139 Chr.1: 40069651-40069651 on Build GRCh38

View Full Product Details