Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150625374-150625912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENSA
Entrez Gene ID
2029
Gene Name
endosulfine alpha
Gene Aliases
ARPP-19e
UniGene
Hs.632456
Gene Chromosome Location
Chr.1: 150622123-150629622 on Build GRCh38
Transcripts
NM_004436.2 NM_207042.1 NM_207043.1 NM_207044.1 NM_207045.1 NM_207046.1 NM_207047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442687 c.363G>C p.Q121H Chr.1: 150625677-150625677 on Build GRCh38
1638142 c.272A>G p.D91G Chr.1: 150625708-150625708 on Build GRCh38
895588 c.241G>A p.A81T Chr.1: 150625739-150625739 on Build GRCh38
5898092 c.265G>A p.A89T Chr.1: 150625775-150625775 on Build GRCh38
2182396 c.318C>T p.L106L Chr.1: 150625662-150625662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750303030 Chr.1: 150625721-150625721 on Build GRCh38
rs1044584958 Chr.1: 150625812-150625812 on Build GRCh38
rs759307017 Chr.1: 150625602-150625602 on Build GRCh38
rs1053732 Chr.1: 150625414-150625414 on Build GRCh38
rs760238315 Chr.1: 150625683-150625683 on Build GRCh38
rs758219128 Chr.1: 150625778-150625778 on Build GRCh38
rs757226308 Chr.1: 150625714-150625714 on Build GRCh38
rs191186947 Chr.1: 150625505-150625505 on Build GRCh38
rs902403692 Chr.1: 150625849-150625849 on Build GRCh38
rs200585444 Chr.1: 150625799-150625799 on Build GRCh38
rs377022254 Chr.1: 150625820-150625820 on Build GRCh38
rs749947307 Chr.1: 150625504-150625504 on Build GRCh38
rs17847054 Chr.1: 150625815-150625815 on Build GRCh38
rs773811012 Chr.1: 150625665-150625665 on Build GRCh38
rs775536016 Chr.1: 150625588-150625588 on Build GRCh38
rs779711387 Chr.1: 150625779-150625779 on Build GRCh38
rs1017605596 Chr.1: 150625582-150625582 on Build GRCh38
rs587668355 Chr.1: 150625842-150625842 on Build GRCh38
rs768741308 Chr.1: 150625816-150625816 on Build GRCh38
rs767225802 Chr.1: 150625604-150625604 on Build GRCh38
rs587715685 Chr.1: 150625474-150625474 on Build GRCh38
rs914838780 Chr.1: 150625687-150625687 on Build GRCh38
rs770062514 Chr.1: 150625662-150625662 on Build GRCh38
rs752396164 Chr.1: 150625676-150625676 on Build GRCh38
rs755659410 Chr.1: 150625617-150625617 on Build GRCh38
rs777078545 Chr.1: 150625618-150625618 on Build GRCh38
rs758637615 Chr.1: 150625678-150625679 on Build GRCh38
rs374404692 Chr.1: 150625661-150625661 on Build GRCh38
rs745693911 Chr.1: 150625632-150625632 on Build GRCh38
rs17847055 Chr.1: 150625691-150625691 on Build GRCh38
rs750798198 Chr.1: 150625601-150625601 on Build GRCh38
rs112156402 Chr.1: 150625640-150625640 on Build GRCh38
rs777283764 Chr.1: 150625597-150625597 on Build GRCh38
rs755074318 Chr.1: 150625695-150625695 on Build GRCh38
rs757808438 Chr.1: 150625451-150625451 on Build GRCh38
rs772314823 Chr.1: 150625585-150625585 on Build GRCh38
rs369716092 Chr.1: 150625644-150625644 on Build GRCh38
rs763394268 Chr.1: 150625666-150625666 on Build GRCh38
rs758668505 Chr.1: 150625408-150625408 on Build GRCh38
rs781464885 Chr.1: 150625837-150625837 on Build GRCh38
rs587695752 Chr.1: 150625878-150625878 on Build GRCh38
rs780577412 Chr.1: 150625820-150625823 on Build GRCh38
rs141017549 Chr.1: 150625698-150625698 on Build GRCh38
rs587662361 Chr.1: 150625537-150625537 on Build GRCh38
rs768563248 Chr.1: 150625592-150625592 on Build GRCh38
rs587619112 Chr.1: 150625599-150625599 on Build GRCh38
rs757171377 Chr.1: 150625630-150625630 on Build GRCh38
rs587741996 Chr.1: 150625598-150625598 on Build GRCh38
rs747437375 Chr.1: 150625442-150625442 on Build GRCh38
rs367689257 Chr.1: 150625631-150625631 on Build GRCh38
rs985945805 Chr.1: 150625546-150625546 on Build GRCh38
rs776840279 Chr.1: 150625645-150625645 on Build GRCh38
rs781072684 Chr.1: 150625473-150625473 on Build GRCh38
rs587638398 Chr.1: 150625468-150625468 on Build GRCh38
rs750966911 Chr.1: 150625625-150625625 on Build GRCh38
rs752239839 Chr.1: 150625607-150625607 on Build GRCh38
rs867278318 Chr.1: 150625444-150625444 on Build GRCh38
rs747899840 Chr.1: 150625422-150625422 on Build GRCh38
rs587613552 Chr.1: 150625876-150625876 on Build GRCh38
rs201883426 Chr.1: 150625657-150625657 on Build GRCh38
rs370302073 Chr.1: 150625847-150625847 on Build GRCh38
rs897733343 Chr.1: 150625439-150625439 on Build GRCh38
rs951853539 Chr.1: 150625520-150625520 on Build GRCh38
rs1053731 Chr.1: 150625464-150625464 on Build GRCh38
rs765707789 Chr.1: 150625616-150625616 on Build GRCh38
rs35031758 Chr.1: 150625486-150625486 on Build GRCh38
rs113697732 Chr.1: 150625641-150625641 on Build GRCh38
rs867580122 Chr.1: 150625775-150625775 on Build GRCh38
rs778844671 Chr.1: 150625716-150625716 on Build GRCh38
rs763650351 Chr.1: 150625686-150625686 on Build GRCh38
rs587706257 Chr.1: 150625413-150625413 on Build GRCh38
rs766591643 Chr.1: 150625674-150625674 on Build GRCh38
rs753837481 Chr.1: 150625627-150625627 on Build GRCh38

View Full Product Details