Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 52152142-52152723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
UniGene
Hs.473082
Gene Chromosome Location
Chr.20: 52084011-52191985 on Build GRCh38
Transcripts
NM_018197.2 NM_022088.4 NM_199426.1 NM_199427.2 XM_005260449.1 XM_011528900.1 XM_017027945.1 XM_017027946.1 XM_017027947.1 XM_017027948.1 XM_017027949.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099474 c.2018T>A p.L673H Chr.20: 52152174-52152174 on Build GRCh38
1665094 c.1698G>C p.Q566H Chr.20: 52152494-52152494 on Build GRCh38
1028083 c.1513G>T p.V505L Chr.20: 52152679-52152679 on Build GRCh38
4099478 c.1699C>T p.P567S Chr.20: 52152493-52152493 on Build GRCh38
6467750 c.1844A>T p.E615V Chr.20: 52152348-52152348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142733573 Chr.20: 52152260-52152260 on Build GRCh38
rs759475656 Chr.20: 52152202-52152202 on Build GRCh38
rs372513108 Chr.20: 52152682-52152682 on Build GRCh38
rs766205483 Chr.20: 52152309-52152309 on Build GRCh38
rs754067712 Chr.20: 52152617-52152646 on Build GRCh38
rs765768696 Chr.20: 52152553-52152553 on Build GRCh38
rs538759043 Chr.20: 52152628-52152628 on Build GRCh38
rs779357788 Chr.20: 52152417-52152417 on Build GRCh38
rs751963528 Chr.20: 52152681-52152681 on Build GRCh38
rs763019627 Chr.20: 52152458-52152458 on Build GRCh38
rs757241662 Chr.20: 52152212-52152212 on Build GRCh38
rs898657352 Chr.20: 52152515-52152515 on Build GRCh38
rs148156379 Chr.20: 52152623-52152623 on Build GRCh38
rs552566545 Chr.20: 52152598-52152598 on Build GRCh38
rs774690725 Chr.20: 52152195-52152195 on Build GRCh38
rs750446356 Chr.20: 52152297-52152297 on Build GRCh38
rs1028495656 Chr.20: 52152536-52152536 on Build GRCh38
rs773219331 Chr.20: 52152240-52152242 on Build GRCh38
rs1019602084 Chr.20: 52152542-52152542 on Build GRCh38
rs748291906 Chr.20: 52152527-52152527 on Build GRCh38
rs770251429 Chr.20: 52152253-52152253 on Build GRCh38
rs147640480 Chr.20: 52152477-52152477 on Build GRCh38
rs929817002 Chr.20: 52152519-52152519 on Build GRCh38
rs769770502 Chr.20: 52152599-52152599 on Build GRCh38
rs763344719 Chr.20: 52152175-52152175 on Build GRCh38
rs201143929 Chr.20: 52152509-52152509 on Build GRCh38
rs3746415 Chr.20: 52152644-52152644 on Build GRCh38
rs538149981 Chr.20: 52152402-52152402 on Build GRCh38
rs752877993 Chr.20: 52152685-52152685 on Build GRCh38
rs747184276 Chr.20: 52152584-52152584 on Build GRCh38
rs140355555 Chr.20: 52152451-52152451 on Build GRCh38
rs368408323 Chr.20: 52152691-52152691 on Build GRCh38
rs201509797 Chr.20: 52152246-52152246 on Build GRCh38
rs750897508 Chr.20: 52152556-52152556 on Build GRCh38
rs751137128 Chr.20: 52152353-52152353 on Build GRCh38
rs998570041 Chr.20: 52152573-52152573 on Build GRCh38
rs749638360 Chr.20: 52152261-52152261 on Build GRCh38
rs6096798 Chr.20: 52152686-52152686 on Build GRCh38
rs569471599 Chr.20: 52152390-52152390 on Build GRCh38
rs1011115375 Chr.20: 52152613-52152613 on Build GRCh38
rs780743874 Chr.20: 52152371-52152371 on Build GRCh38
rs768803312 Chr.20: 52152518-52152518 on Build GRCh38
rs954027266 Chr.20: 52152305-52152305 on Build GRCh38
rs373208828 Chr.20: 52152565-52152565 on Build GRCh38
rs774369370 Chr.20: 52152272-52152272 on Build GRCh38
rs757751291 Chr.20: 52152480-52152480 on Build GRCh38
rs1003932257 Chr.20: 52152255-52152255 on Build GRCh38
rs149755369 Chr.20: 52152301-52152301 on Build GRCh38
rs752353435 Chr.20: 52152470-52152470 on Build GRCh38
rs758754357 Chr.20: 52152505-52152505 on Build GRCh38
rs779336538 Chr.20: 52152481-52152481 on Build GRCh38
rs761318542 Chr.20: 52152656-52152656 on Build GRCh38
rs371012655 Chr.20: 52152173-52152173 on Build GRCh38
rs367952344 Chr.20: 52152673-52152673 on Build GRCh38
rs901189782 Chr.20: 52152544-52152544 on Build GRCh38
rs371444820 Chr.20: 52152436-52152436 on Build GRCh38
rs750213517 Chr.20: 52152214-52152214 on Build GRCh38
rs759312491 Chr.20: 52152466-52152466 on Build GRCh38
rs765315166 Chr.20: 52152213-52152213 on Build GRCh38
rs114019425 Chr.20: 52152538-52152538 on Build GRCh38
rs780974964 Chr.20: 52152614-52152616 on Build GRCh38
rs535504707 Chr.20: 52152537-52152537 on Build GRCh38
rs868792368 Chr.20: 52152428-52152428 on Build GRCh38
rs758336956 Chr.20: 52152224-52152224 on Build GRCh38
rs371253933 Chr.20: 52152502-52152502 on Build GRCh38
rs373844291 Chr.20: 52152254-52152254 on Build GRCh38
rs965870764 Chr.20: 52152380-52152380 on Build GRCh38
rs780585554 Chr.20: 52152577-52152577 on Build GRCh38
rs772308417 Chr.20: 52152434-52152434 on Build GRCh38
rs575442535 Chr.20: 52152684-52152684 on Build GRCh38
rs779843130 Chr.20: 52152229-52152229 on Build GRCh38
rs768872836 Chr.20: 52152443-52152443 on Build GRCh38
rs866264764 Chr.20: 52152429-52152429 on Build GRCh38
rs1044057328 Chr.20: 52152180-52152180 on Build GRCh38
rs753983765 Chr.20: 52152467-52152467 on Build GRCh38
rs6126479 Chr.20: 52152680-52152680 on Build GRCh38
rs771297618 Chr.20: 52152270-52152270 on Build GRCh38
rs759832692 Chr.20: 52152278-52152278 on Build GRCh38
rs755564775 Chr.20: 52152473-52152473 on Build GRCh38
rs750890950 Chr.20: 52152662-52152662 on Build GRCh38
rs143029301 Chr.20: 52152610-52152610 on Build GRCh38
rs1056138410 Chr.20: 52152230-52152230 on Build GRCh38
rs896206865 Chr.20: 52152251-52152251 on Build GRCh38
rs746267986 Chr.20: 52152424-52152424 on Build GRCh38
rs760917371 Chr.20: 52152536-52152537 on Build GRCh38
rs917571027 Chr.20: 52152701-52152701 on Build GRCh38
rs140887206 Chr.20: 52152280-52152280 on Build GRCh38
rs760879418 Chr.20: 52152284-52152284 on Build GRCh38
rs754633449 Chr.20: 52152370-52152370 on Build GRCh38
rs267605996 Chr.20: 52152237-52152237 on Build GRCh38
rs913736579 Chr.20: 52152169-52152169 on Build GRCh38
rs552476108 Chr.20: 52152377-52152377 on Build GRCh38
rs753323030 Chr.20: 52152539-52152539 on Build GRCh38
rs778192852 Chr.20: 52152394-52152394 on Build GRCh38
rs371710298 Chr.20: 52152596-52152596 on Build GRCh38
rs776878713 Chr.20: 52152525-52152525 on Build GRCh38
rs145330065 Chr.20: 52152465-52152465 on Build GRCh38
rs770892003 Chr.20: 52152528-52152528 on Build GRCh38
rs201261996 Chr.20: 52152293-52152293 on Build GRCh38
rs142815362 Chr.20: 52152191-52152191 on Build GRCh38
rs755233063 Chr.20: 52152588-52152588 on Build GRCh38
rs775235876 Chr.20: 52152624-52152624 on Build GRCh38
rs896174403 Chr.20: 52152491-52152491 on Build GRCh38
rs1018229701 Chr.20: 52152308-52152308 on Build GRCh38
rs746691994 Chr.20: 52152236-52152236 on Build GRCh38
rs777288666 Chr.20: 52152474-52152474 on Build GRCh38
rs201865916 Chr.20: 52152245-52152245 on Build GRCh38
rs771244712 Chr.20: 52152411-52152411 on Build GRCh38
rs1057386304 Chr.20: 52152194-52152194 on Build GRCh38
rs776331231 Chr.20: 52152647-52152647 on Build GRCh38
rs770091651 Chr.20: 52152166-52152166 on Build GRCh38
rs986762477 Chr.20: 52152363-52152363 on Build GRCh38
rs374626162 Chr.20: 52152659-52152659 on Build GRCh38
rs200007514 Chr.20: 52152242-52152242 on Build GRCh38
rs1019631807 Chr.20: 52152379-52152379 on Build GRCh38
rs747245391 Chr.20: 52152514-52152514 on Build GRCh38
rs767638637 Chr.20: 52152209-52152209 on Build GRCh38
rs767182744 Chr.20: 52152469-52152469 on Build GRCh38
rs758856624 Chr.20: 52152670-52152670 on Build GRCh38
rs913715787 Chr.20: 52152422-52152422 on Build GRCh38
rs753678487 Chr.20: 52152211-52152211 on Build GRCh38

View Full Product Details