Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14132438-14132842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
UniGene
Hs.517817
Gene Chromosome Location
Chr.3: 14124940-14143680 on Build GRCh38
Transcripts
NM_024334.2 XM_017007176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7402941 c.420G>C p.K140N Chr.3: 14132573-14132573 on Build GRCh38
4583997 c.429G>A p.T143T Chr.3: 14132582-14132582 on Build GRCh38
4114093 c.440A>G p.Y147C Chr.3: 14132593-14132593 on Build GRCh38
1039281 c.428C>T p.T143M Chr.3: 14132581-14132581 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200679654 Chr.3: 14132521-14132521 on Build GRCh38
rs140347235 Chr.3: 14132556-14132556 on Build GRCh38
rs906383978 Chr.3: 14132616-14132616 on Build GRCh38
rs967530900 Chr.3: 14132754-14132754 on Build GRCh38
rs1009682494 Chr.3: 14132772-14132772 on Build GRCh38
rs748969808 Chr.3: 14132528-14132528 on Build GRCh38
rs370539390 Chr.3: 14132820-14132820 on Build GRCh38
rs544554435 Chr.3: 14132581-14132581 on Build GRCh38
rs1053499435 Chr.3: 14132590-14132590 on Build GRCh38
rs142500068 Chr.3: 14132819-14132819 on Build GRCh38
rs765773804 Chr.3: 14132514-14132514 on Build GRCh38
rs766312977 Chr.3: 14132593-14132593 on Build GRCh38
rs573490765 Chr.3: 14132502-14132502 on Build GRCh38
rs373590238 Chr.3: 14132602-14132602 on Build GRCh38
rs373911134 Chr.3: 14132506-14132506 on Build GRCh38
rs779719982 Chr.3: 14132517-14132517 on Build GRCh38
rs368256924 Chr.3: 14132817-14132817 on Build GRCh38
rs936302875 Chr.3: 14132774-14132774 on Build GRCh38
rs956785376 Chr.3: 14132524-14132524 on Build GRCh38
rs763593970 Chr.3: 14132486-14132486 on Build GRCh38
rs141389147 Chr.3: 14132634-14132634 on Build GRCh38
rs794729180 Chr.3: 14132565-14132565 on Build GRCh38
rs917618583 Chr.3: 14132618-14132618 on Build GRCh38
rs776611413 Chr.3: 14132573-14132573 on Build GRCh38
rs745443053 Chr.3: 14132632-14132632 on Build GRCh38
rs1019702462 Chr.3: 14132800-14132800 on Build GRCh38
rs4685075 Chr.3: 14132646-14132646 on Build GRCh38
rs949055931 Chr.3: 14132631-14132631 on Build GRCh38
rs751945696 Chr.3: 14132522-14132522 on Build GRCh38
rs770609078 Chr.3: 14132538-14132538 on Build GRCh38
rs888415742 Chr.3: 14132653-14132653 on Build GRCh38
rs771621691 Chr.3: 14132638-14132638 on Build GRCh38
rs73022892 Chr.3: 14132497-14132497 on Build GRCh38
rs779379048 Chr.3: 14132640-14132640 on Build GRCh38
rs755350435 Chr.3: 14132525-14132525 on Build GRCh38
rs976880930 Chr.3: 14132548-14132548 on Build GRCh38
rs763100204 Chr.3: 14132586-14132586 on Build GRCh38
rs145619906 Chr.3: 14132577-14132577 on Build GRCh38
rs984187981 Chr.3: 14132517-14132517 on Build GRCh38
rs964082657 Chr.3: 14132685-14132685 on Build GRCh38
rs965409708 Chr.3: 14132809-14132809 on Build GRCh38
rs377207870 Chr.3: 14132610-14132610 on Build GRCh38
rs201217046 Chr.3: 14132582-14132582 on Build GRCh38
rs952837460 Chr.3: 14132511-14132511 on Build GRCh38
rs768069110 Chr.3: 14132823-14132823 on Build GRCh38
rs926198782 Chr.3: 14132773-14132773 on Build GRCh38
rs746639451 Chr.3: 14132570-14132572 on Build GRCh38
rs148412226 Chr.3: 14132670-14132670 on Build GRCh38
rs778405877 Chr.3: 14132554-14132554 on Build GRCh38
rs1019649607 Chr.3: 14132551-14132551 on Build GRCh38
rs1029760464 Chr.3: 14132734-14132734 on Build GRCh38
rs370422391 Chr.3: 14132555-14132555 on Build GRCh38
rs553502856 Chr.3: 14132469-14132469 on Build GRCh38
rs202103307 Chr.3: 14132615-14132615 on Build GRCh38
rs762090341 Chr.3: 14132508-14132508 on Build GRCh38
rs976030066 Chr.3: 14132561-14132561 on Build GRCh38
rs781640632 Chr.3: 14132608-14132608 on Build GRCh38
rs369973984 Chr.3: 14132505-14132505 on Build GRCh38
rs760075635 Chr.3: 14132478-14132478 on Build GRCh38
rs876657589 Chr.3: 14132552-14132552 on Build GRCh38
rs376194839 Chr.3: 14132501-14132501 on Build GRCh38
rs781487997 Chr.3: 14132526-14132526 on Build GRCh38
rs530183718 Chr.3: 14132607-14132607 on Build GRCh38
rs397517384 Chr.3: 14132566-14132566 on Build GRCh38
rs560249672 Chr.3: 14132647-14132647 on Build GRCh38
rs756941756 Chr.3: 14132761-14132761 on Build GRCh38
rs760628106 Chr.3: 14132563-14132563 on Build GRCh38
rs765528284 Chr.3: 14132509-14132509 on Build GRCh38
rs559271953 Chr.3: 14132559-14132559 on Build GRCh38
rs4685074 Chr.3: 14132463-14132463 on Build GRCh38
rs564825681 Chr.3: 14132767-14132767 on Build GRCh38
rs1005963205 Chr.3: 14132661-14132661 on Build GRCh38
rs976827571 Chr.3: 14132708-14132708 on Build GRCh38
rs551211598 Chr.3: 14132733-14132733 on Build GRCh38
rs768508021 Chr.3: 14132613-14132614 on Build GRCh38
rs370566174 Chr.3: 14132627-14132627 on Build GRCh38
rs41284009 Chr.3: 14132818-14132818 on Build GRCh38
rs892621309 Chr.3: 14132679-14132679 on Build GRCh38
rs896138468 Chr.3: 14132462-14132462 on Build GRCh38

View Full Product Details