Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24148922-24149518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF31
Entrez Gene ID
55072
Gene Name
ring finger protein 31
Gene Aliases
HOIP, ZIBRA
UniGene
Hs.375217
Gene Chromosome Location
Chr.14: 24146875-24160661 on Build GRCh38
Transcripts
NM_001310332.1 NM_017999.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5664915 c.721G>T p.D241Y Chr.14: 24149495-24149495 on Build GRCh38
4506890 c.732C>T p.F244F Chr.14: 24149506-24149506 on Build GRCh38
3495352 c.648C>T p.P216P Chr.14: 24149422-24149422 on Build GRCh38
1161108 c.692C>T p.S231F Chr.14: 24149466-24149466 on Build GRCh38
3706222 c.646C>T p.P216S Chr.14: 24149420-24149420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894111157 Chr.14: 24148975-24148975 on Build GRCh38
rs765711403 Chr.14: 24149444-24149444 on Build GRCh38
rs755276406 Chr.14: 24149466-24149466 on Build GRCh38
rs1040026611 Chr.14: 24149305-24149305 on Build GRCh38
rs750943498 Chr.14: 24149456-24149456 on Build GRCh38
rs879249416 Chr.14: 24149120-24149120 on Build GRCh38
rs1042780855 Chr.14: 24149010-24149010 on Build GRCh38
rs199527364 Chr.14: 24149390-24149390 on Build GRCh38
rs535613393 Chr.14: 24149458-24149458 on Build GRCh38
rs959819195 Chr.14: 24149283-24149283 on Build GRCh38
rs890631210 Chr.14: 24149333-24149333 on Build GRCh38
rs904265865 Chr.14: 24149020-24149020 on Build GRCh38
rs1052764634 Chr.14: 24149325-24149325 on Build GRCh38
rs567393911 Chr.14: 24149420-24149420 on Build GRCh38
rs548968073 Chr.14: 24149152-24149152 on Build GRCh38
rs964702691 Chr.14: 24149051-24149051 on Build GRCh38
rs762222518 Chr.14: 24149429-24149429 on Build GRCh38
rs748848206 Chr.14: 24149114-24149114 on Build GRCh38
rs934430520 Chr.14: 24149318-24149318 on Build GRCh38
rs770595748 Chr.14: 24149217-24149217 on Build GRCh38
rs536010125 Chr.14: 24149027-24149027 on Build GRCh38
rs112309739 Chr.14: 24149033-24149033 on Build GRCh38
rs920723776 Chr.14: 24149059-24149059 on Build GRCh38
rs76506991 Chr.14: 24149294-24149294 on Build GRCh38
rs920085194 Chr.14: 24149221-24149221 on Build GRCh38
rs1002753537 Chr.14: 24149428-24149428 on Build GRCh38
rs772018625 Chr.14: 24149380-24149380 on Build GRCh38
rs372729032 Chr.14: 24148967-24148970 on Build GRCh38
rs138293863 Chr.14: 24149125-24149125 on Build GRCh38
rs775351085 Chr.14: 24149417-24149417 on Build GRCh38
rs925581673 Chr.14: 24149309-24149309 on Build GRCh38
rs991988611 Chr.14: 24149182-24149182 on Build GRCh38
rs892978560 Chr.14: 24149077-24149077 on Build GRCh38
rs1004971588 Chr.14: 24149483-24149483 on Build GRCh38
rs944909546 Chr.14: 24149383-24149383 on Build GRCh38
rs996334244 Chr.14: 24149045-24149045 on Build GRCh38
rs559325863 Chr.14: 24149194-24149194 on Build GRCh38
rs148955427 Chr.14: 24149113-24149113 on Build GRCh38
rs954987782 Chr.14: 24149040-24149040 on Build GRCh38
rs28373297 Chr.14: 24149041-24149041 on Build GRCh38
rs938560578 Chr.14: 24148955-24148955 on Build GRCh38
rs569705519 Chr.14: 24148962-24148965 on Build GRCh38
rs759335598 Chr.14: 24149357-24149357 on Build GRCh38
rs372191060 Chr.14: 24149394-24149394 on Build GRCh38
rs747529822 Chr.14: 24149043-24149043 on Build GRCh38
rs887062454 Chr.14: 24148944-24148944 on Build GRCh38
rs886413630 Chr.14: 24149480-24149480 on Build GRCh38
rs950858997 Chr.14: 24149066-24149066 on Build GRCh38
rs981447671 Chr.14: 24149130-24149130 on Build GRCh38
rs35706103 Chr.14: 24149009-24149009 on Build GRCh38
rs1035618227 Chr.14: 24149449-24149449 on Build GRCh38
rs551251178 Chr.14: 24149056-24149056 on Build GRCh38
rs569834236 Chr.14: 24148986-24148986 on Build GRCh38
rs11621170 Chr.14: 24149034-24149034 on Build GRCh38
rs1056846126 Chr.14: 24148963-24148963 on Build GRCh38
rs1050612660 Chr.14: 24149347-24149347 on Build GRCh38
rs577681084 Chr.14: 24149112-24149112 on Build GRCh38
rs143864107 Chr.14: 24149163-24149163 on Build GRCh38
rs1057046871 Chr.14: 24149109-24149109 on Build GRCh38
rs780425900 Chr.14: 24149462-24149462 on Build GRCh38
rs528373764 Chr.14: 24149227-24149227 on Build GRCh38
rs978481051 Chr.14: 24149303-24149303 on Build GRCh38
rs1028786830 Chr.14: 24149058-24149058 on Build GRCh38
rs557761265 Chr.14: 24149067-24149067 on Build GRCh38
rs773937995 Chr.14: 24149270-24149270 on Build GRCh38
rs549776100 Chr.14: 24149296-24149296 on Build GRCh38
rs983505120 Chr.14: 24149259-24149259 on Build GRCh38
rs377644189 Chr.14: 24149255-24149255 on Build GRCh38
rs753446597 Chr.14: 24149398-24149398 on Build GRCh38
rs386775670 Chr.14: 24149294-24149295 on Build GRCh38
rs908391180 Chr.14: 24149262-24149262 on Build GRCh38
rs760448261 Chr.14: 24149387-24149387 on Build GRCh38
rs775245836 Chr.14: 24149386-24149386 on Build GRCh38
rs376778540 Chr.14: 24149467-24149467 on Build GRCh38
rs111853946 Chr.14: 24149119-24149119 on Build GRCh38
rs769764453 Chr.14: 24149481-24149481 on Build GRCh38
rs79491227 Chr.14: 24149295-24149295 on Build GRCh38
rs908360159 Chr.14: 24149088-24149088 on Build GRCh38
rs181239130 Chr.14: 24149258-24149258 on Build GRCh38
rs901508605 Chr.14: 24149316-24149316 on Build GRCh38
rs751749904 Chr.14: 24149464-24149464 on Build GRCh38
rs556213738 Chr.14: 24148960-24148960 on Build GRCh38
rs991705787 Chr.14: 24149014-24149014 on Build GRCh38
rs1053230994 Chr.14: 24149135-24149135 on Build GRCh38
rs938591546 Chr.14: 24149089-24149089 on Build GRCh38
rs1017821375 Chr.14: 24149188-24149188 on Build GRCh38
rs369458996 Chr.14: 24149479-24149479 on Build GRCh38

View Full Product Details