Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114325810-114326407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAMP1
Entrez Gene ID
283489
Gene Name
chromosome alignment maintaining phosphoprotein 1
Gene Aliases
C13orf8, CAMP, CHAMP, MRD40, ZNF828
UniGene
Hs.7542
Gene Chromosome Location
Chr.13: 114314490-114327328 on Build GRCh38
Transcripts
NM_001164144.1 NM_001164145.1 NM_032436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362097 c.2086G>C p.E696Q Chr.13: 114325928-114325928 on Build GRCh38
6139001 c.2305C>T p.R769C Chr.13: 114326147-114326147 on Build GRCh38
6074506 c.2306G>T p.R769L Chr.13: 114326148-114326148 on Build GRCh38
7250835 c.2039G>A p.R680K Chr.13: 114325881-114325881 on Build GRCh38
3813502 c.2120G>C p.G707A Chr.13: 114325962-114325962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571190481 Chr.13: 114326188-114326188 on Build GRCh38
rs782385979 Chr.13: 114325833-114325833 on Build GRCh38
rs373039957 Chr.13: 114326122-114326122 on Build GRCh38
rs782121908 Chr.13: 114326173-114326173 on Build GRCh38
rs782652236 Chr.13: 114326291-114326291 on Build GRCh38
rs782738856 Chr.13: 114326217-114326217 on Build GRCh38
rs782706179 Chr.13: 114326035-114326035 on Build GRCh38
rs541199861 Chr.13: 114326340-114326340 on Build GRCh38
rs782007597 Chr.13: 114325864-114325864 on Build GRCh38
rs781891231 Chr.13: 114326256-114326256 on Build GRCh38
rs146400058 Chr.13: 114325873-114325873 on Build GRCh38
rs782326298 Chr.13: 114326306-114326306 on Build GRCh38
rs530106558 Chr.13: 114326110-114326110 on Build GRCh38
rs915085355 Chr.13: 114326017-114326017 on Build GRCh38
rs956667133 Chr.13: 114325956-114325956 on Build GRCh38
rs782472338 Chr.13: 114326263-114326263 on Build GRCh38
rs863225073 Chr.13: 114325871-114325871 on Build GRCh38
rs549128164 Chr.13: 114325909-114325909 on Build GRCh38
rs9525332 Chr.13: 114325924-114325924 on Build GRCh38
rs778495008 Chr.13: 114326372-114326372 on Build GRCh38
rs376247811 Chr.13: 114326148-114326148 on Build GRCh38
rs545355043 Chr.13: 114326359-114326359 on Build GRCh38
rs782180635 Chr.13: 114325845-114325845 on Build GRCh38
rs200252544 Chr.13: 114326237-114326237 on Build GRCh38
rs782504028 Chr.13: 114326288-114326288 on Build GRCh38
rs553407055 Chr.13: 114326347-114326347 on Build GRCh38
rs781807614 Chr.13: 114326258-114326258 on Build GRCh38
rs994435488 Chr.13: 114326266-114326266 on Build GRCh38
rs782498070 Chr.13: 114325877-114325877 on Build GRCh38
rs782548115 Chr.13: 114326257-114326257 on Build GRCh38
rs782812169 Chr.13: 114325870-114325870 on Build GRCh38
rs782717140 Chr.13: 114325939-114325939 on Build GRCh38
rs782241760 Chr.13: 114326281-114326281 on Build GRCh38
rs782772066 Chr.13: 114326254-114326254 on Build GRCh38
rs546667566 Chr.13: 114325930-114325930 on Build GRCh38
rs573063891 Chr.13: 114326334-114326334 on Build GRCh38
rs782218818 Chr.13: 114325900-114325900 on Build GRCh38
rs782293650 Chr.13: 114326119-114326119 on Build GRCh38
rs781925044 Chr.13: 114326209-114326209 on Build GRCh38
rs201877258 Chr.13: 114326241-114326241 on Build GRCh38
rs782300134 Chr.13: 114325891-114325891 on Build GRCh38
rs138469382 Chr.13: 114325912-114325912 on Build GRCh38
rs782145955 Chr.13: 114326015-114326015 on Build GRCh38
rs531028864 Chr.13: 114325881-114325881 on Build GRCh38
rs782000882 Chr.13: 114326326-114326326 on Build GRCh38
rs112961928 Chr.13: 114325993-114325993 on Build GRCh38
rs1036000661 Chr.13: 114326179-114326179 on Build GRCh38
rs116051126 Chr.13: 114326308-114326308 on Build GRCh38
rs956885076 Chr.13: 114326355-114326355 on Build GRCh38
rs374749899 Chr.13: 114326305-114326305 on Build GRCh38
rs548006337 Chr.13: 114325858-114325858 on Build GRCh38
rs782661164 Chr.13: 114326118-114326118 on Build GRCh38
rs570263813 Chr.13: 114326341-114326341 on Build GRCh38
rs923906622 Chr.13: 114325925-114325925 on Build GRCh38
rs782582908 Chr.13: 114325897-114325897 on Build GRCh38
rs577974765 Chr.13: 114326357-114326357 on Build GRCh38
rs144440153 Chr.13: 114326039-114326039 on Build GRCh38
rs782616919 Chr.13: 114326267-114326267 on Build GRCh38
rs375209771 Chr.13: 114326333-114326333 on Build GRCh38
rs782476517 Chr.13: 114326042-114326042 on Build GRCh38
rs35812683 Chr.13: 114325855-114325855 on Build GRCh38
rs782747799 Chr.13: 114325972-114325972 on Build GRCh38
rs782215496 Chr.13: 114326126-114326126 on Build GRCh38
rs369844864 Chr.13: 114325914-114325914 on Build GRCh38
rs927742550 Chr.13: 114326124-114326124 on Build GRCh38
rs782042437 Chr.13: 114325839-114325839 on Build GRCh38
rs138697692 Chr.13: 114325874-114325874 on Build GRCh38
rs533342855 Chr.13: 114325859-114325859 on Build GRCh38
rs781866640 Chr.13: 114325978-114325978 on Build GRCh38
rs782259506 Chr.13: 114325913-114325913 on Build GRCh38
rs782346249 Chr.13: 114326128-114326128 on Build GRCh38
rs782532236 Chr.13: 114326116-114326116 on Build GRCh38
rs149344812 Chr.13: 114326251-114326251 on Build GRCh38
rs1027175377 Chr.13: 114325848-114325848 on Build GRCh38
rs782108456 Chr.13: 114326314-114326314 on Build GRCh38
rs782633216 Chr.13: 114326063-114326063 on Build GRCh38
rs782442773 Chr.13: 114325895-114325895 on Build GRCh38
rs782584547 Chr.13: 114326125-114326125 on Build GRCh38
rs782333083 Chr.13: 114325853-114325853 on Build GRCh38
rs186679023 Chr.13: 114325867-114325867 on Build GRCh38
rs782736727 Chr.13: 114325875-114325875 on Build GRCh38
rs781950538 Chr.13: 114325933-114325933 on Build GRCh38
rs781958255 Chr.13: 114325854-114325854 on Build GRCh38
rs782382113 Chr.13: 114326318-114326318 on Build GRCh38
rs782073505 Chr.13: 114326211-114326211 on Build GRCh38
rs752555069 Chr.13: 114326218-114326218 on Build GRCh38
rs939091454 Chr.13: 114326149-114326149 on Build GRCh38

View Full Product Details