Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72086022-72086580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXNL4B
Entrez Gene ID
54957
Gene Name
thioredoxin like 4B
Gene Aliases
DLP, Dim2
UniGene
Hs.134406
Gene Chromosome Location
Chr.16: 72044261-72094316 on Build GRCh38
Transcripts
NM_001142317.1 NM_001142318.1 NM_001324354.1 NM_001324355.1 NM_017853.2 XM_017023377.1

SNP Detail

SNP ID Chromosome Location
rs1040755789 Chr.16: 72086511-72086511 on Build GRCh38
rs796224047 Chr.16: 72086189-72086192 on Build GRCh38
rs553606579 Chr.16: 72086148-72086148 on Build GRCh38
rs1022106905 Chr.16: 72086367-72086370 on Build GRCh38
rs756880604 Chr.16: 72086537-72086537 on Build GRCh38
rs148582661 Chr.16: 72086189-72086190 on Build GRCh38
rs574025325 Chr.16: 72086188-72086189 on Build GRCh38
rs1010176181 Chr.16: 72086212-72086212 on Build GRCh38
rs571895769 Chr.16: 72086395-72086395 on Build GRCh38
rs900416705 Chr.16: 72086335-72086335 on Build GRCh38
rs948858256 Chr.16: 72086130-72086130 on Build GRCh38
rs535849388 Chr.16: 72086188-72086188 on Build GRCh38
rs930812257 Chr.16: 72086505-72086505 on Build GRCh38
rs574612987 Chr.16: 72086365-72086365 on Build GRCh38
rs1000251444 Chr.16: 72086374-72086374 on Build GRCh38
rs924620376 Chr.16: 72086077-72086077 on Build GRCh38
rs190285787 Chr.16: 72086186-72086186 on Build GRCh38
rs377071693 Chr.16: 72086142-72086142 on Build GRCh38
rs1047924673 Chr.16: 72086512-72086512 on Build GRCh38
rs895953746 Chr.16: 72086116-72086116 on Build GRCh38
rs55953809 Chr.16: 72086163-72086163 on Build GRCh38
rs199528394 Chr.16: 72086558-72086558 on Build GRCh38
rs934877035 Chr.16: 72086094-72086097 on Build GRCh38
rs752686461 Chr.16: 72086293-72086293 on Build GRCh38
rs956033391 Chr.16: 72086189-72086194 on Build GRCh38
rs747998561 Chr.16: 72086190-72086190 on Build GRCh38
rs920784665 Chr.16: 72086181-72086181 on Build GRCh38
rs930850911 Chr.16: 72086434-72086434 on Build GRCh38
rs898896008 Chr.16: 72086147-72086147 on Build GRCh38
rs879077221 Chr.16: 72086529-72086531 on Build GRCh38
rs749335758 Chr.16: 72086276-72086276 on Build GRCh38
rs551592367 Chr.16: 72086053-72086053 on Build GRCh38
rs538699326 Chr.16: 72086140-72086140 on Build GRCh38
rs560368088 Chr.16: 72086454-72086454 on Build GRCh38
rs948972062 Chr.16: 72086192-72086192 on Build GRCh38
rs930713464 Chr.16: 72086182-72086182 on Build GRCh38
rs920733333 Chr.16: 72086499-72086499 on Build GRCh38
rs189616602 Chr.16: 72086392-72086392 on Build GRCh38
rs543816358 Chr.16: 72086260-72086260 on Build GRCh38
rs539922480 Chr.16: 72086210-72086210 on Build GRCh38
rs1057395588 Chr.16: 72086098-72086098 on Build GRCh38
rs181155460 Chr.16: 72086208-72086208 on Build GRCh38
rs1036464542 Chr.16: 72086179-72086179 on Build GRCh38
rs978774117 Chr.16: 72086064-72086064 on Build GRCh38
rs765322931 Chr.16: 72086131-72086131 on Build GRCh38
rs940964999 Chr.16: 72086178-72086178 on Build GRCh38
rs772690540 Chr.16: 72086516-72086516 on Build GRCh38
rs904355489 Chr.16: 72086552-72086552 on Build GRCh38
rs571239138 Chr.16: 72086083-72086083 on Build GRCh38
rs916939516 Chr.16: 72086341-72086341 on Build GRCh38
rs992809143 Chr.16: 72086447-72086450 on Build GRCh38
rs777548101 Chr.16: 72086346-72086346 on Build GRCh38
rs562515040 Chr.16: 72086278-72086278 on Build GRCh38
rs533915050 Chr.16: 72086125-72086126 on Build GRCh38
rs185197310 Chr.16: 72086369-72086369 on Build GRCh38
rs186356634 Chr.16: 72086171-72086171 on Build GRCh38
rs944823941 Chr.16: 72086509-72086509 on Build GRCh38
rs968569562 Chr.16: 72086258-72086258 on Build GRCh38
rs558269099 Chr.16: 72086211-72086211 on Build GRCh38
rs770942855 Chr.16: 72086302-72086302 on Build GRCh38
rs907613679 Chr.16: 72086540-72086540 on Build GRCh38
rs527517967 Chr.16: 72086513-72086513 on Build GRCh38
rs766291500 Chr.16: 72086114-72086114 on Build GRCh38
rs944490563 Chr.16: 72086543-72086544 on Build GRCh38
rs968434291 Chr.16: 72086051-72086051 on Build GRCh38
rs1020112276 Chr.16: 72086217-72086217 on Build GRCh38
rs1032165840 Chr.16: 72086385-72086385 on Build GRCh38
rs978532510 Chr.16: 72086084-72086084 on Build GRCh38
rs571653725 Chr.16: 72086115-72086115 on Build GRCh38
rs1008517243 Chr.16: 72086283-72086283 on Build GRCh38
rs576768589 Chr.16: 72086256-72086256 on Build GRCh38
rs536968281 Chr.16: 72086172-72086172 on Build GRCh38
rs754737750 Chr.16: 72086164-72086164 on Build GRCh38
rs917502695 Chr.16: 72086191-72086191 on Build GRCh38
rs980779120 Chr.16: 72086497-72086497 on Build GRCh38
rs1022578645 Chr.16: 72086185-72086185 on Build GRCh38
rs920752485 Chr.16: 72086409-72086409 on Build GRCh38
rs778014670 Chr.16: 72086267-72086267 on Build GRCh38
rs1047936235 Chr.16: 72086249-72086249 on Build GRCh38
rs956147691 Chr.16: 72086426-72086426 on Build GRCh38
rs746372319 Chr.16: 72086462-72086462 on Build GRCh38

View Full Product Details