Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133348821-133349350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL7A
Entrez Gene ID
6130
Gene Name
ribosomal protein L7a
Gene Aliases
L7A, SURF3, TRUP
UniGene
Hs.499839
Gene Chromosome Location
Chr.9: 133348214-133351425 on Build GRCh38
Transcripts
NM_000972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5801446 c.205+7_205+19delAACAAACGGCAGA p.? Chr.9: 133349049-133349061 on Build GRCh38
3664198 c.86C>G p.P29R Chr.9: 133348923-133348923 on Build GRCh38
5998603 c.4-2A>G p.? Chr.9: 133348920-133348920 on Build GRCh38
5801445 c.124+7_124+19delAACAAACGGCAGA p.? Chr.9: 133349049-133349061 on Build GRCh38
4816534 c.111G>C p.K37N Chr.9: 133348948-133348948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782505453 Chr.9: 133348874-133348874 on Build GRCh38
rs1002764919 Chr.9: 133349121-133349121 on Build GRCh38
rs587768272 Chr.9: 133349229-133349229 on Build GRCh38
rs587718297 Chr.9: 133349110-133349110 on Build GRCh38
rs866117788 Chr.9: 133349032-133349032 on Build GRCh38
rs782086601 Chr.9: 133349077-133349078 on Build GRCh38
rs781844776 Chr.9: 133349040-133349040 on Build GRCh38
rs782559003 Chr.9: 133348934-133348934 on Build GRCh38
rs911351579 Chr.9: 133349169-133349169 on Build GRCh38
rs201848146 Chr.9: 133349055-133349055 on Build GRCh38
rs782329924 Chr.9: 133348952-133348952 on Build GRCh38
rs946727549 Chr.9: 133349230-133349230 on Build GRCh38
rs782663301 Chr.9: 133349060-133349060 on Build GRCh38
rs140158783 Chr.9: 133348956-133348956 on Build GRCh38
rs587597213 Chr.9: 133349117-133349117 on Build GRCh38
rs782376974 Chr.9: 133349331-133349331 on Build GRCh38
rs898554834 Chr.9: 133349290-133349290 on Build GRCh38
rs951318889 Chr.9: 133349195-133349195 on Build GRCh38
rs782780896 Chr.9: 133349075-133349075 on Build GRCh38
rs1020199601 Chr.9: 133348851-133348851 on Build GRCh38
rs149247952 Chr.9: 133348989-133348989 on Build GRCh38
rs781803737 Chr.9: 133348865-133348865 on Build GRCh38
rs782749128 Chr.9: 133348904-133348904 on Build GRCh38
rs981750078 Chr.9: 133349205-133349205 on Build GRCh38
rs782022469 Chr.9: 133348914-133348914 on Build GRCh38
rs200213490 Chr.9: 133348870-133348870 on Build GRCh38
rs782163703 Chr.9: 133348957-133348957 on Build GRCh38
rs1055525456 Chr.9: 133349123-133349123 on Build GRCh38
rs946696403 Chr.9: 133349113-133349113 on Build GRCh38
rs782006364 Chr.9: 133348894-133348894 on Build GRCh38
rs782316950 Chr.9: 133349248-133349248 on Build GRCh38
rs1051768660 Chr.9: 133349320-133349320 on Build GRCh38
rs782403624 Chr.9: 133348960-133348960 on Build GRCh38
rs782340207 Chr.9: 133348882-133348882 on Build GRCh38
rs781975723 Chr.9: 133349074-133349075 on Build GRCh38
rs73660477 Chr.9: 133349157-133349157 on Build GRCh38
rs112569069 Chr.9: 133348895-133348895 on Build GRCh38
rs144425262 Chr.9: 133348993-133348993 on Build GRCh38
rs61733976 Chr.9: 133348959-133348959 on Build GRCh38
rs781805579 Chr.9: 133348924-133348924 on Build GRCh38
rs748567836 Chr.9: 133349167-133349167 on Build GRCh38
rs782034205 Chr.9: 133349135-133349135 on Build GRCh38
rs782627550 Chr.9: 133348875-133348875 on Build GRCh38
rs964239413 Chr.9: 133349221-133349221 on Build GRCh38
rs587639875 Chr.9: 133349259-133349259 on Build GRCh38
rs781989638 Chr.9: 133349078-133349078 on Build GRCh38
rs938568364 Chr.9: 133349250-133349250 on Build GRCh38
rs782227461 Chr.9: 133348879-133348879 on Build GRCh38
rs1025667823 Chr.9: 133349150-133349150 on Build GRCh38
rs963148687 Chr.9: 133349136-133349136 on Build GRCh38
rs959425457 Chr.9: 133348915-133348915 on Build GRCh38
rs782206028 Chr.9: 133349191-133349191 on Build GRCh38
rs182444745 Chr.9: 133349188-133349188 on Build GRCh38
rs1029869490 Chr.9: 133349140-133349140 on Build GRCh38
rs370331576 Chr.9: 133348889-133348889 on Build GRCh38
rs782703472 Chr.9: 133349052-133349052 on Build GRCh38
rs781901619 Chr.9: 133348910-133348910 on Build GRCh38
rs782522949 Chr.9: 133349038-133349038 on Build GRCh38
rs782070749 Chr.9: 133349049-133349052 on Build GRCh38
rs966868775 Chr.9: 133348986-133348986 on Build GRCh38
rs782643638 Chr.9: 133349109-133349109 on Build GRCh38
rs61733975 Chr.9: 133348942-133348942 on Build GRCh38
rs782447706 Chr.9: 133348866-133348866 on Build GRCh38
rs781970326 Chr.9: 133349065-133349065 on Build GRCh38
rs782596337 Chr.9: 133348923-133348923 on Build GRCh38
rs962387899 Chr.9: 133349165-133349165 on Build GRCh38
rs1055640985 Chr.9: 133349264-133349264 on Build GRCh38
rs782377187 Chr.9: 133348939-133348939 on Build GRCh38
rs200867852 Chr.9: 133348871-133348871 on Build GRCh38
rs202094353 Chr.9: 133348876-133348876 on Build GRCh38
rs147366255 Chr.9: 133349300-133349300 on Build GRCh38
rs781925389 Chr.9: 133348884-133348884 on Build GRCh38
rs115808246 Chr.9: 133349047-133349047 on Build GRCh38
rs587772127 Chr.9: 133349272-133349272 on Build GRCh38
rs782672194 Chr.9: 133348935-133348935 on Build GRCh38
rs1018410883 Chr.9: 133349162-133349162 on Build GRCh38
rs587721459 Chr.9: 133349260-133349260 on Build GRCh38
rs782273980 Chr.9: 133348867-133348867 on Build GRCh38
rs376472313 Chr.9: 133348909-133348909 on Build GRCh38
rs138377720 Chr.9: 133349222-133349222 on Build GRCh38
rs782209853 Chr.9: 133348951-133348951 on Build GRCh38
rs782734893 Chr.9: 133348854-133348854 on Build GRCh38
rs1012707981 Chr.9: 133349133-133349133 on Build GRCh38
rs111889641 Chr.9: 133348868-133348868 on Build GRCh38
rs782489093 Chr.9: 133348853-133348853 on Build GRCh38
rs994162788 Chr.9: 133349146-133349146 on Build GRCh38
rs782071464 Chr.9: 133348905-133348905 on Build GRCh38
rs782816752 Chr.9: 133349011-133349011 on Build GRCh38
rs77053072 Chr.9: 133349073-133349073 on Build GRCh38
rs1025615260 Chr.9: 133349329-133349329 on Build GRCh38
rs1037962067 Chr.9: 133349273-133349273 on Build GRCh38
rs781890308 Chr.9: 133349030-133349030 on Build GRCh38
rs782474203 Chr.9: 133349101-133349101 on Build GRCh38
rs782113166 Chr.9: 133348979-133348979 on Build GRCh38
rs782142365 Chr.9: 133348918-133348918 on Build GRCh38
rs894437862 Chr.9: 133349265-133349265 on Build GRCh38
rs1041361850 Chr.9: 133349239-133349239 on Build GRCh38
rs368775843 Chr.9: 133349069-133349069 on Build GRCh38
rs782791948 Chr.9: 133348919-133348919 on Build GRCh38
rs781992976 Chr.9: 133348972-133348972 on Build GRCh38
rs782255884 Chr.9: 133349063-133349063 on Build GRCh38
rs782627034 Chr.9: 133348947-133348947 on Build GRCh38
rs187084133 Chr.9: 133349317-133349317 on Build GRCh38
rs782415138 Chr.9: 133349056-133349056 on Build GRCh38
rs781839445 Chr.9: 133349080-133349080 on Build GRCh38
rs375396197 Chr.9: 133348888-133348888 on Build GRCh38
rs1006039846 Chr.9: 133349266-133349266 on Build GRCh38
rs782367984 Chr.9: 133349064-133349064 on Build GRCh38
rs782491634 Chr.9: 133348922-133348922 on Build GRCh38
rs781939250 Chr.9: 133349053-133349053 on Build GRCh38
rs781860070 Chr.9: 133348843-133348843 on Build GRCh38
rs918832856 Chr.9: 133349023-133349023 on Build GRCh38
rs973178593 Chr.9: 133348999-133348999 on Build GRCh38
rs75296864 Chr.9: 133348938-133348938 on Build GRCh38
rs149274136 Chr.9: 133349225-133349225 on Build GRCh38
rs945727295 Chr.9: 133349079-133349079 on Build GRCh38
rs894320373 Chr.9: 133349128-133349128 on Build GRCh38
rs370344235 Chr.9: 133349070-133349070 on Build GRCh38
rs782065861 Chr.9: 133348996-133348996 on Build GRCh38
rs782549999 Chr.9: 133349130-133349130 on Build GRCh38

View Full Product Details