Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15826009-15826467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S2
Entrez Gene ID
8905
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Gene Aliases
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
UniGene
Hs.121592
Gene Chromosome Location
Chr.X: 15825806-15855014 on Build GRCh38
Transcripts
NM_001272071.1 NM_003916.4

SNP Detail

SNP ID Chromosome Location
rs761779751 Chr.X: 15826044-15826044 on Build GRCh38
rs750241601 Chr.X: 15826106-15826106 on Build GRCh38
rs766027688 Chr.X: 15826358-15826358 on Build GRCh38
rs899393151 Chr.X: 15826180-15826180 on Build GRCh38
rs755237953 Chr.X: 15826373-15826373 on Build GRCh38
rs1012605479 Chr.X: 15826339-15826339 on Build GRCh38
rs940663426 Chr.X: 15826136-15826136 on Build GRCh38
rs894104991 Chr.X: 15826267-15826267 on Build GRCh38
rs1056986018 Chr.X: 15826164-15826164 on Build GRCh38
rs767277961 Chr.X: 15826103-15826103 on Build GRCh38
rs145357287 Chr.X: 15826206-15826207 on Build GRCh38
rs1036577479 Chr.X: 15826153-15826153 on Build GRCh38
rs201057676 Chr.X: 15826077-15826077 on Build GRCh38
rs11559236 Chr.X: 15826244-15826244 on Build GRCh38
rs940128588 Chr.X: 15826049-15826049 on Build GRCh38
rs1050533459 Chr.X: 15826346-15826347 on Build GRCh38
rs760281928 Chr.X: 15826212-15826212 on Build GRCh38
rs938563240 Chr.X: 15826125-15826125 on Build GRCh38
rs4830549 Chr.X: 15826406-15826406 on Build GRCh38
rs930841026 Chr.X: 15826230-15826230 on Build GRCh38

View Full Product Details