Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70287442-70287919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD11
Entrez Gene ID
51248
Gene Name
PDZ domain containing 11
Gene Aliases
AIPP1, PDZK11, PISP
UniGene
Hs.11042
Gene Chromosome Location
Chr.X: 70286361-70290040 on Build GRCh38
Transcripts
NM_016484.4 XM_011530973.2 XM_017029568.1

SNP Detail

SNP ID Chromosome Location
rs200559348 Chr.X: 70287492-70287493 on Build GRCh38
rs943252804 Chr.X: 70287676-70287676 on Build GRCh38
rs915005275 Chr.X: 70287490-70287490 on Build GRCh38
rs774730581 Chr.X: 70287764-70287764 on Build GRCh38
rs763678771 Chr.X: 70287855-70287855 on Build GRCh38
rs369155616 Chr.X: 70287729-70287729 on Build GRCh38
rs758634672 Chr.X: 70287493-70287496 on Build GRCh38
rs893734188 Chr.X: 70287828-70287828 on Build GRCh38
rs369599592 Chr.X: 70287872-70287872 on Build GRCh38
rs376362055 Chr.X: 70287833-70287833 on Build GRCh38
rs959614010 Chr.X: 70287534-70287534 on Build GRCh38
rs779689084 Chr.X: 70287662-70287662 on Build GRCh38
rs752269794 Chr.X: 70287846-70287846 on Build GRCh38
rs973322613 Chr.X: 70287581-70287581 on Build GRCh38
rs750860718 Chr.X: 70287813-70287813 on Build GRCh38
rs372097765 Chr.X: 70287802-70287802 on Build GRCh38
rs757906923 Chr.X: 70287848-70287848 on Build GRCh38
rs906693324 Chr.X: 70287567-70287567 on Build GRCh38
rs755956243 Chr.X: 70287654-70287654 on Build GRCh38
rs756972395 Chr.X: 70287866-70287866 on Build GRCh38
rs751234370 Chr.X: 70287865-70287865 on Build GRCh38
rs763267723 Chr.X: 70287723-70287723 on Build GRCh38
rs142893388 Chr.X: 70287807-70287807 on Build GRCh38
rs1007586723 Chr.X: 70287876-70287876 on Build GRCh38
rs766897504 Chr.X: 70287496-70287497 on Build GRCh38
rs775771527 Chr.X: 70287716-70287716 on Build GRCh38
rs767562458 Chr.X: 70287535-70287535 on Build GRCh38
rs745681788 Chr.X: 70287544-70287544 on Build GRCh38
rs987131544 Chr.X: 70287613-70287613 on Build GRCh38
rs1017499386 Chr.X: 70287561-70287561 on Build GRCh38
rs376659417 Chr.X: 70287694-70287694 on Build GRCh38
rs781089787 Chr.X: 70287867-70287867 on Build GRCh38
rs768936239 Chr.X: 70287762-70287762 on Build GRCh38
rs929042301 Chr.X: 70287605-70287605 on Build GRCh38
rs1004243191 Chr.X: 70287574-70287577 on Build GRCh38
rs962885871 Chr.X: 70287580-70287580 on Build GRCh38
rs190805228 Chr.X: 70287590-70287590 on Build GRCh38
rs1053827476 Chr.X: 70287481-70287484 on Build GRCh38
rs764637335 Chr.X: 70287842-70287842 on Build GRCh38

View Full Product Details