Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143197616-143198197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP38
Entrez Gene ID
84640
Gene Name
ubiquitin specific peptidase 38
Gene Aliases
HP43.8KD
UniGene
Hs.480848
Gene Chromosome Location
Chr.4: 143184917-143223874 on Build GRCh38
Transcripts
NM_001290325.1 NM_001290326.1 NM_032557.6 XM_011532360.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1427299 c.1030T>C p.S344P Chr.4: 143197904-143197904 on Build GRCh38
6099216 c.971C>T p.P324L Chr.4: 143197845-143197845 on Build GRCh38
1757104 c.1016T>C p.L339P Chr.4: 143197890-143197890 on Build GRCh38
4879899 c.955A>C p.N319H Chr.4: 143197829-143197829 on Build GRCh38
6818681 c.960A>G p.R320R Chr.4: 143197834-143197834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768670603 Chr.4: 143197823-143197824 on Build GRCh38
rs1027331034 Chr.4: 143197689-143197689 on Build GRCh38
rs756621775 Chr.4: 143198030-143198030 on Build GRCh38
rs565938934 Chr.4: 143197967-143197967 on Build GRCh38
rs112817305 Chr.4: 143198038-143198038 on Build GRCh38
rs377766871 Chr.4: 143197847-143197847 on Build GRCh38
rs750998283 Chr.4: 143197961-143197961 on Build GRCh38
rs932443644 Chr.4: 143198163-143198163 on Build GRCh38
rs1006636728 Chr.4: 143197706-143197706 on Build GRCh38
rs370336302 Chr.4: 143197777-143197777 on Build GRCh38
rs981546625 Chr.4: 143197770-143197770 on Build GRCh38
rs750316766 Chr.4: 143197846-143197846 on Build GRCh38
rs996612896 Chr.4: 143197653-143197653 on Build GRCh38
rs574029506 Chr.4: 143197801-143197801 on Build GRCh38
rs751547360 Chr.4: 143197953-143197953 on Build GRCh38
rs976354620 Chr.4: 143198129-143198129 on Build GRCh38
rs1016729920 Chr.4: 143197871-143197871 on Build GRCh38
rs761018001 Chr.4: 143197937-143197937 on Build GRCh38
rs898357053 Chr.4: 143198157-143198157 on Build GRCh38
rs531818717 Chr.4: 143197751-143197751 on Build GRCh38
rs368529972 Chr.4: 143197948-143197948 on Build GRCh38
rs201470099 Chr.4: 143197799-143197799 on Build GRCh38
rs888337422 Chr.4: 143197693-143197693 on Build GRCh38
rs1035752596 Chr.4: 143197773-143197773 on Build GRCh38
rs775025171 Chr.4: 143197912-143197912 on Build GRCh38
rs924836005 Chr.4: 143198106-143198106 on Build GRCh38
rs766233315 Chr.4: 143197860-143197860 on Build GRCh38
rs768776146 Chr.4: 143197943-143197943 on Build GRCh38
rs1053396826 Chr.4: 143198049-143198049 on Build GRCh38
rs145787864 Chr.4: 143197982-143197982 on Build GRCh38
rs147637039 Chr.4: 143197723-143197723 on Build GRCh38
rs371195237 Chr.4: 143197934-143197934 on Build GRCh38
rs761484807 Chr.4: 143197807-143197807 on Build GRCh38
rs765150924 Chr.4: 143197838-143197838 on Build GRCh38
rs1006627657 Chr.4: 143198125-143198125 on Build GRCh38
rs962509268 Chr.4: 143197878-143197878 on Build GRCh38
rs543107635 Chr.4: 143198022-143198022 on Build GRCh38
rs993256890 Chr.4: 143197995-143197996 on Build GRCh38
rs183161476 Chr.4: 143198020-143198020 on Build GRCh38
rs752276868 Chr.4: 143197975-143197975 on Build GRCh38
rs754506809 Chr.4: 143197870-143197870 on Build GRCh38
rs147219891 Chr.4: 143197864-143197864 on Build GRCh38
rs754230751 Chr.4: 143198048-143198048 on Build GRCh38
rs749625606 Chr.4: 143197907-143197907 on Build GRCh38
rs969276247 Chr.4: 143198077-143198077 on Build GRCh38
rs141985651 Chr.4: 143197915-143197915 on Build GRCh38
rs759691257 Chr.4: 143197796-143197796 on Build GRCh38
rs972663728 Chr.4: 143197922-143197922 on Build GRCh38
rs148184087 Chr.4: 143198141-143198141 on Build GRCh38
rs991619828 Chr.4: 143197795-143197795 on Build GRCh38
rs778257147 Chr.4: 143197906-143197906 on Build GRCh38
rs761852268 Chr.4: 143197914-143197914 on Build GRCh38
rs766255426 Chr.4: 143197950-143197950 on Build GRCh38
rs1035784612 Chr.4: 143198027-143198027 on Build GRCh38
rs751628017 Chr.4: 143197908-143197908 on Build GRCh38
rs187890872 Chr.4: 143198171-143198171 on Build GRCh38
rs868062038 Chr.4: 143198021-143198021 on Build GRCh38
rs145318222 Chr.4: 143197652-143197652 on Build GRCh38
rs767081429 Chr.4: 143197973-143197973 on Build GRCh38
rs747838635 Chr.4: 143197874-143197874 on Build GRCh38
rs978067535 Chr.4: 143197815-143197815 on Build GRCh38
rs889423943 Chr.4: 143198110-143198110 on Build GRCh38
rs756429025 Chr.4: 143197891-143197891 on Build GRCh38
rs578116161 Chr.4: 143198122-143198122 on Build GRCh38
rs1048611186 Chr.4: 143198166-143198166 on Build GRCh38
rs761933370 Chr.4: 143197937-143197938 on Build GRCh38
rs776664815 Chr.4: 143197937-143197937 on Build GRCh38
rs140205066 Chr.4: 143198169-143198169 on Build GRCh38
rs550047150 Chr.4: 143197756-143197756 on Build GRCh38
rs571348392 Chr.4: 143197782-143197782 on Build GRCh38
rs774511914 Chr.4: 143197793-143197793 on Build GRCh38
rs771399241 Chr.4: 143197909-143197909 on Build GRCh38
rs977726942 Chr.4: 143198100-143198100 on Build GRCh38
rs772321535 Chr.4: 143197932-143197932 on Build GRCh38
rs1021827464 Chr.4: 143198047-143198047 on Build GRCh38
rs934234877 Chr.4: 143198060-143198060 on Build GRCh38
rs375796770 Chr.4: 143197942-143197942 on Build GRCh38
rs755732818 Chr.4: 143197976-143197976 on Build GRCh38
rs766678736 Chr.4: 143198046-143198046 on Build GRCh38
rs958736727 Chr.4: 143198042-143198042 on Build GRCh38
rs913489848 Chr.4: 143198053-143198053 on Build GRCh38
rs764255036 Chr.4: 143197848-143197848 on Build GRCh38
rs924332105 Chr.4: 143197833-143197833 on Build GRCh38
rs554126637 Chr.4: 143197986-143197986 on Build GRCh38
rs556756535 Chr.4: 143198078-143198078 on Build GRCh38

View Full Product Details