Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83720513-83721111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN1
Entrez Gene ID
1404
Gene Name
hyaluronan and proteoglycan link protein 1
Gene Aliases
CRT1, CRTL1
UniGene
Hs.2799
Gene Chromosome Location
Chr.5: 83638198-83721210 on Build GRCh38
Transcripts
NM_001884.3 XM_017009051.1 XM_017009053.1

SNP Detail

SNP ID Chromosome Location
rs752959938 Chr.5: 83721078-83721078 on Build GRCh38
rs1048362362 Chr.5: 83720826-83720826 on Build GRCh38
rs1045190979 Chr.5: 83720686-83720686 on Build GRCh38
rs908465368 Chr.5: 83720576-83720576 on Build GRCh38
rs984364104 Chr.5: 83720850-83720850 on Build GRCh38
rs1055422911 Chr.5: 83721064-83721064 on Build GRCh38
rs775177246 Chr.5: 83720681-83720681 on Build GRCh38
rs777638758 Chr.5: 83721080-83721080 on Build GRCh38
rs892703927 Chr.5: 83720952-83720952 on Build GRCh38
rs1022262452 Chr.5: 83720917-83720924 on Build GRCh38
rs531896601 Chr.5: 83720765-83720765 on Build GRCh38
rs543743236 Chr.5: 83720992-83720992 on Build GRCh38
rs3734092 Chr.5: 83720924-83720924 on Build GRCh38
rs6862559 Chr.5: 83720908-83720908 on Build GRCh38
rs73135931 Chr.5: 83720568-83720568 on Build GRCh38
rs113444306 Chr.5: 83720935-83720935 on Build GRCh38
rs1011317985 Chr.5: 83720915-83720924 on Build GRCh38
rs917899293 Chr.5: 83720910-83720910 on Build GRCh38
rs983170157 Chr.5: 83721041-83721041 on Build GRCh38
rs769484362 Chr.5: 83720612-83720612 on Build GRCh38
rs572117504 Chr.5: 83720863-83720863 on Build GRCh38
rs369348897 Chr.5: 83720563-83720563 on Build GRCh38
rs940663856 Chr.5: 83720580-83720580 on Build GRCh38
rs996478944 Chr.5: 83721015-83721015 on Build GRCh38
rs1015508545 Chr.5: 83720877-83720877 on Build GRCh38
rs144938331 Chr.5: 83720729-83720729 on Build GRCh38
rs568641419 Chr.5: 83721079-83721079 on Build GRCh38
rs920232454 Chr.5: 83721036-83721036 on Build GRCh38
rs562126869 Chr.5: 83720727-83720727 on Build GRCh38
rs961320109 Chr.5: 83720898-83720898 on Build GRCh38
rs936842749 Chr.5: 83720705-83720705 on Build GRCh38
rs926702395 Chr.5: 83720930-83720930 on Build GRCh38
rs761782763 Chr.5: 83721040-83721040 on Build GRCh38
rs954534629 Chr.5: 83720954-83720954 on Build GRCh38
rs765559626 Chr.5: 83721063-83721063 on Build GRCh38
rs754083270 Chr.5: 83720921-83720924 on Build GRCh38
rs954647113 Chr.5: 83720692-83720692 on Build GRCh38
rs565255810 Chr.5: 83721083-83721083 on Build GRCh38
rs1041301396 Chr.5: 83721002-83721002 on Build GRCh38
rs888233425 Chr.5: 83721055-83721055 on Build GRCh38
rs149938174 Chr.5: 83720562-83720562 on Build GRCh38
rs148249697 Chr.5: 83720908-83720909 on Build GRCh38
rs758419784 Chr.5: 83721035-83721036 on Build GRCh38
rs188723145 Chr.5: 83720552-83720552 on Build GRCh38
rs538653305 Chr.5: 83720766-83720766 on Build GRCh38
rs770198782 Chr.5: 83720923-83720924 on Build GRCh38
rs765421422 Chr.5: 83721057-83721057 on Build GRCh38
rs900819713 Chr.5: 83721013-83721013 on Build GRCh38
rs936850412 Chr.5: 83720933-83720933 on Build GRCh38
rs761266343 Chr.5: 83720811-83720811 on Build GRCh38
rs768004946 Chr.5: 83720909-83720912 on Build GRCh38
rs1019317654 Chr.5: 83720547-83720547 on Build GRCh38
rs1031428062 Chr.5: 83720672-83720676 on Build GRCh38
rs945654069 Chr.5: 83720983-83720983 on Build GRCh38
rs917999675 Chr.5: 83720628-83720628 on Build GRCh38
rs1017474637 Chr.5: 83721001-83721001 on Build GRCh38
rs972098252 Chr.5: 83720608-83720608 on Build GRCh38
rs796776299 Chr.5: 83720923-83720924 on Build GRCh38
rs567437650 Chr.5: 83720675-83720675 on Build GRCh38
rs981218935 Chr.5: 83721089-83721089 on Build GRCh38
rs996531241 Chr.5: 83720782-83720782 on Build GRCh38
rs1055371025 Chr.5: 83720868-83720868 on Build GRCh38
rs538184302 Chr.5: 83720703-83720703 on Build GRCh38
rs900911220 Chr.5: 83720768-83720768 on Build GRCh38
rs746761717 Chr.5: 83720911-83720912 on Build GRCh38
rs866998320 Chr.5: 83720872-83720872 on Build GRCh38
rs980817410 Chr.5: 83720928-83720928 on Build GRCh38
rs759669191 Chr.5: 83721019-83721019 on Build GRCh38
rs939480523 Chr.5: 83721062-83721062 on Build GRCh38
rs745332284 Chr.5: 83720572-83720572 on Build GRCh38
rs72772959 Chr.5: 83720842-83720842 on Build GRCh38
rs181114718 Chr.5: 83720556-83720556 on Build GRCh38
rs977666995 Chr.5: 83720630-83720630 on Build GRCh38
rs1044135613 Chr.5: 83721085-83721085 on Build GRCh38
rs551782194 Chr.5: 83720549-83720549 on Build GRCh38
rs1015873731 Chr.5: 83721076-83721076 on Build GRCh38
rs949478305 Chr.5: 83720668-83720668 on Build GRCh38
rs879306719 Chr.5: 83720893-83720893 on Build GRCh38
rs952545218 Chr.5: 83720845-83720845 on Build GRCh38
rs549168014 Chr.5: 83720661-83720661 on Build GRCh38
rs759026908 Chr.5: 83720988-83720990 on Build GRCh38
rs756453119 Chr.5: 83720919-83720924 on Build GRCh38
rs184679814 Chr.5: 83720736-83720736 on Build GRCh38

View Full Product Details