Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133973587-133974179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VDAC1
Entrez Gene ID
7416
Gene Name
voltage dependent anion channel 1
Gene Aliases
PORIN, VDAC-1
UniGene
Hs.519320
Gene Chromosome Location
Chr.5: 133971875-134070987 on Build GRCh38
Transcripts
NM_003374.2 NR_036624.1 NR_036625.1 XM_005272075.3 XM_017009821.1 XM_017009822.1 XM_017009823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665252 c.760+1G>A p.? Chr.5: 133973790-133973790 on Build GRCh38
6390275 c.725T>C p.L242P Chr.5: 133973826-133973826 on Build GRCh38
4639534 c.752T>C p.L251P Chr.5: 133973799-133973799 on Build GRCh38
6819752 c.738A>G p.G246G Chr.5: 133973813-133973813 on Build GRCh38
6299047 c.727A>T p.I243L Chr.5: 133973824-133973824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575399164 Chr.5: 133973949-133973949 on Build GRCh38
rs567179814 Chr.5: 133973652-133973652 on Build GRCh38
rs897935289 Chr.5: 133973934-133973934 on Build GRCh38
rs1033945622 Chr.5: 133974001-133974001 on Build GRCh38
rs775224948 Chr.5: 133973847-133973847 on Build GRCh38
rs77732824 Chr.5: 133974045-133974045 on Build GRCh38
rs761661572 Chr.5: 133973900-133973900 on Build GRCh38
rs771250383 Chr.5: 133973794-133973794 on Build GRCh38
rs375859645 Chr.5: 133973831-133973831 on Build GRCh38
rs1053964117 Chr.5: 133973756-133973756 on Build GRCh38
rs781023382 Chr.5: 133973668-133973668 on Build GRCh38
rs751322449 Chr.5: 133973890-133973890 on Build GRCh38
rs776981869 Chr.5: 133973808-133973808 on Build GRCh38
rs1041559905 Chr.5: 133973758-133973758 on Build GRCh38
rs935807169 Chr.5: 133973791-133973791 on Build GRCh38
rs914182208 Chr.5: 133974150-133974150 on Build GRCh38
rs977157827 Chr.5: 133974085-133974085 on Build GRCh38
rs370413816 Chr.5: 133973685-133973685 on Build GRCh38
rs945685338 Chr.5: 133974080-133974083 on Build GRCh38
rs935706413 Chr.5: 133974136-133974136 on Build GRCh38
rs916897059 Chr.5: 133974068-133974068 on Build GRCh38
rs201989551 Chr.5: 133973865-133973865 on Build GRCh38
rs143911563 Chr.5: 133973998-133973998 on Build GRCh38
rs371614785 Chr.5: 133973793-133973793 on Build GRCh38
rs190644738 Chr.5: 133974018-133974018 on Build GRCh38
rs556631319 Chr.5: 133973726-133973726 on Build GRCh38
rs1008932474 Chr.5: 133973893-133973893 on Build GRCh38
rs952265367 Chr.5: 133973982-133973982 on Build GRCh38
rs1015560010 Chr.5: 133973980-133973980 on Build GRCh38
rs1056693679 Chr.5: 133973677-133973680 on Build GRCh38
rs772522232 Chr.5: 133973766-133973766 on Build GRCh38
rs769931323 Chr.5: 133973837-133973837 on Build GRCh38
rs925652136 Chr.5: 133974122-133974122 on Build GRCh38
rs925622625 Chr.5: 133973674-133973674 on Build GRCh38
rs962814917 Chr.5: 133973631-133973631 on Build GRCh38
rs534550326 Chr.5: 133973664-133973664 on Build GRCh38
rs997360154 Chr.5: 133973929-133973929 on Build GRCh38
rs561763556 Chr.5: 133974123-133974123 on Build GRCh38
rs925600126 Chr.5: 133974124-133974124 on Build GRCh38
rs540207071 Chr.5: 133974089-133974089 on Build GRCh38
rs961192698 Chr.5: 133973983-133973983 on Build GRCh38
rs768115827 Chr.5: 133973745-133973746 on Build GRCh38
rs866567997 Chr.5: 133973675-133973675 on Build GRCh38
rs1041514238 Chr.5: 133973825-133973825 on Build GRCh38
rs113954293 Chr.5: 133973917-133973917 on Build GRCh38
rs200670358 Chr.5: 133973777-133973777 on Build GRCh38
rs746182745 Chr.5: 133973738-133973740 on Build GRCh38
rs1024576576 Chr.5: 133973999-133973999 on Build GRCh38
rs557094528 Chr.5: 133973942-133973942 on Build GRCh38
rs1005434057 Chr.5: 133973955-133973955 on Build GRCh38
rs1018953531 Chr.5: 133973920-133973920 on Build GRCh38
rs763829989 Chr.5: 133973879-133973879 on Build GRCh38
rs755706327 Chr.5: 133973740-133973740 on Build GRCh38
rs199569040 Chr.5: 133973765-133973765 on Build GRCh38
rs529374243 Chr.5: 133974149-133974149 on Build GRCh38
rs926844768 Chr.5: 133973672-133973672 on Build GRCh38
rs993569713 Chr.5: 133973937-133973937 on Build GRCh38
rs773999034 Chr.5: 133973758-133973761 on Build GRCh38
rs13360512 Chr.5: 133973624-133973624 on Build GRCh38

View Full Product Details