Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25243296-25243870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK5
Entrez Gene ID
80005
Gene Name
dedicator of cytokinesis 5
Gene Aliases
-
UniGene
Hs.195403
Gene Chromosome Location
Chr.8: 25184723-25415716 on Build GRCh38
Transcripts
NM_001322810.1 NM_024940.7 XM_011544661.1 XM_011544662.1 XM_011544663.2 XM_017013865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098419 c.71A>G p.D24G Chr.8: 25243701-25243701 on Build GRCh38
6113252 c.126_127GG>AT p.G43C Chr.8: 25243756-25243757 on Build GRCh38
7296137 c.44-6T>A p.? Chr.8: 25243668-25243668 on Build GRCh38
6328533 c.75G>A p.V25V Chr.8: 25243705-25243705 on Build GRCh38
2155374 c.112C>A p.L38M Chr.8: 25243742-25243742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76256507 Chr.8: 25243349-25243349 on Build GRCh38
rs779242604 Chr.8: 25243787-25243787 on Build GRCh38
rs762577734 Chr.8: 25243703-25243703 on Build GRCh38
rs749488331 Chr.8: 25243665-25243665 on Build GRCh38
rs137877997 Chr.8: 25243689-25243689 on Build GRCh38
rs951155746 Chr.8: 25243392-25243392 on Build GRCh38
rs771121025 Chr.8: 25243674-25243674 on Build GRCh38
rs1019426585 Chr.8: 25243744-25243744 on Build GRCh38
rs761191943 Chr.8: 25243693-25243693 on Build GRCh38
rs111674029 Chr.8: 25243438-25243440 on Build GRCh38
rs553802115 Chr.8: 25243565-25243565 on Build GRCh38
rs561725983 Chr.8: 25243687-25243687 on Build GRCh38
rs75525567 Chr.8: 25243334-25243334 on Build GRCh38
rs752655929 Chr.8: 25243750-25243750 on Build GRCh38
rs140538988 Chr.8: 25243676-25243676 on Build GRCh38
rs184284715 Chr.8: 25243619-25243619 on Build GRCh38
rs975991946 Chr.8: 25243472-25243472 on Build GRCh38
rs1025491154 Chr.8: 25243391-25243391 on Build GRCh38
rs191735564 Chr.8: 25243424-25243424 on Build GRCh38
rs1054934141 Chr.8: 25243610-25243610 on Build GRCh38
rs755894705 Chr.8: 25243627-25243627 on Build GRCh38
rs936433048 Chr.8: 25243495-25243495 on Build GRCh38
rs545756029 Chr.8: 25243609-25243609 on Build GRCh38
rs899131550 Chr.8: 25243702-25243702 on Build GRCh38
rs765234181 Chr.8: 25243800-25243800 on Build GRCh38
rs931825024 Chr.8: 25243539-25243539 on Build GRCh38
rs936472411 Chr.8: 25243825-25243825 on Build GRCh38
rs572228434 Chr.8: 25243588-25243588 on Build GRCh38
rs777577591 Chr.8: 25243634-25243634 on Build GRCh38
rs1050256565 Chr.8: 25243548-25243548 on Build GRCh38
rs75920458 Chr.8: 25243774-25243774 on Build GRCh38
rs753667084 Chr.8: 25243772-25243772 on Build GRCh38
rs759016364 Chr.8: 25243732-25243732 on Build GRCh38
rs938252495 Chr.8: 25243680-25243680 on Build GRCh38
rs187573021 Chr.8: 25243380-25243380 on Build GRCh38
rs370221374 Chr.8: 25243700-25243700 on Build GRCh38
rs749973395 Chr.8: 25243802-25243803 on Build GRCh38
rs975509003 Chr.8: 25243481-25243481 on Build GRCh38
rs568678016 Chr.8: 25243487-25243487 on Build GRCh38
rs10574448 Chr.8: 25243440-25243442 on Build GRCh38
rs777436666 Chr.8: 25243754-25243754 on Build GRCh38
rs768913050 Chr.8: 25243694-25243694 on Build GRCh38
rs1032424067 Chr.8: 25243796-25243796 on Build GRCh38
rs1046535203 Chr.8: 25243670-25243670 on Build GRCh38
rs753089758 Chr.8: 25243455-25243455 on Build GRCh38
rs971572845 Chr.8: 25243354-25243354 on Build GRCh38
rs73558459 Chr.8: 25243329-25243329 on Build GRCh38
rs766001492 Chr.8: 25243711-25243711 on Build GRCh38
rs558165580 Chr.8: 25243618-25243618 on Build GRCh38
rs1010217102 Chr.8: 25243658-25243658 on Build GRCh38
rs766975337 Chr.8: 25243742-25243742 on Build GRCh38
rs9692988 Chr.8: 25243381-25243381 on Build GRCh38
rs1008094428 Chr.8: 25243739-25243739 on Build GRCh38
rs539379147 Chr.8: 25243333-25243333 on Build GRCh38
rs370687065 Chr.8: 25243707-25243707 on Build GRCh38
rs556459476 Chr.8: 25243441-25243441 on Build GRCh38
rs148369439 Chr.8: 25243372-25243372 on Build GRCh38
rs751118171 Chr.8: 25243723-25243723 on Build GRCh38
rs1002644339 Chr.8: 25243330-25243330 on Build GRCh38
rs146170766 Chr.8: 25243637-25243637 on Build GRCh38
rs868620404 Chr.8: 25243608-25243608 on Build GRCh38
rs535662725 Chr.8: 25243493-25243493 on Build GRCh38
rs993551791 Chr.8: 25243382-25243382 on Build GRCh38
rs376980871 Chr.8: 25243798-25243798 on Build GRCh38
rs757301238 Chr.8: 25243738-25243738 on Build GRCh38
rs1024533162 Chr.8: 25243338-25243338 on Build GRCh38
rs963982550 Chr.8: 25243475-25243475 on Build GRCh38
rs756023582 Chr.8: 25243753-25243753 on Build GRCh38
rs907563815 Chr.8: 25243400-25243400 on Build GRCh38
rs961740162 Chr.8: 25243724-25243724 on Build GRCh38
rs73558460 Chr.8: 25243817-25243817 on Build GRCh38
rs1029643722 Chr.8: 25243332-25243332 on Build GRCh38
rs746044466 Chr.8: 25243794-25243794 on Build GRCh38
rs986630165 Chr.8: 25243423-25243423 on Build GRCh38

View Full Product Details