Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12088547-12089133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHTKD1
Entrez Gene ID
55526
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Gene Aliases
AMOXAD, CMT2Q
UniGene
Hs.104980
Gene Chromosome Location
Chr.10: 12068917-12123228 on Build GRCh38
Transcripts
NM_018706.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4392648 c.857A>G p.N286S Chr.10: 12089125-12089125 on Build GRCh38
5772419 c.808G>T p.D270Y Chr.10: 12089076-12089076 on Build GRCh38
3934947 c.769G>A p.A257T Chr.10: 12089037-12089037 on Build GRCh38
6496499 c.790C>T p.H264Y Chr.10: 12089058-12089058 on Build GRCh38
5002470 c.728G>A p.R243H Chr.10: 12088996-12088996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917079059 Chr.10: 12088869-12088869 on Build GRCh38
rs749394389 Chr.10: 12089093-12089093 on Build GRCh38
rs531103791 Chr.10: 12088695-12088695 on Build GRCh38
rs974847822 Chr.10: 12088778-12088778 on Build GRCh38
rs749218142 Chr.10: 12088829-12088829 on Build GRCh38
rs755438188 Chr.10: 12088786-12088786 on Build GRCh38
rs953269786 Chr.10: 12088631-12088631 on Build GRCh38
rs143069945 Chr.10: 12089048-12089048 on Build GRCh38
rs374760148 Chr.10: 12089092-12089092 on Build GRCh38
rs370385309 Chr.10: 12089012-12089012 on Build GRCh38
rs764869852 Chr.10: 12089068-12089068 on Build GRCh38
rs761602486 Chr.10: 12088960-12088960 on Build GRCh38
rs973216642 Chr.10: 12088581-12088581 on Build GRCh38
rs949814479 Chr.10: 12088694-12088694 on Build GRCh38
rs750129976 Chr.10: 12088615-12088615 on Build GRCh38
rs768767270 Chr.10: 12089097-12089097 on Build GRCh38
rs755879860 Chr.10: 12088669-12088669 on Build GRCh38
rs891984020 Chr.10: 12088724-12088724 on Build GRCh38
rs144889711 Chr.10: 12088652-12088652 on Build GRCh38
rs1045941816 Chr.10: 12088773-12088773 on Build GRCh38
rs756701886 Chr.10: 12089091-12089091 on Build GRCh38
rs1000860597 Chr.10: 12088714-12088714 on Build GRCh38
rs1028829925 Chr.10: 12088843-12088843 on Build GRCh38
rs781779430 Chr.10: 12088996-12088996 on Build GRCh38
rs377037358 Chr.10: 12089049-12089049 on Build GRCh38
rs765841515 Chr.10: 12088973-12088973 on Build GRCh38
rs754612294 Chr.10: 12088987-12088987 on Build GRCh38
rs113395386 Chr.10: 12088833-12088833 on Build GRCh38
rs748366299 Chr.10: 12089107-12089107 on Build GRCh38
rs1055875577 Chr.10: 12088716-12088716 on Build GRCh38
rs895577077 Chr.10: 12088693-12088693 on Build GRCh38
rs768873512 Chr.10: 12088941-12088941 on Build GRCh38
rs753304961 Chr.10: 12089059-12089059 on Build GRCh38
rs553049198 Chr.10: 12089086-12089086 on Build GRCh38
rs542507859 Chr.10: 12088645-12088645 on Build GRCh38
rs200951603 Chr.10: 12088962-12088962 on Build GRCh38
rs761570353 Chr.10: 12089029-12089029 on Build GRCh38
rs946035749 Chr.10: 12088677-12088677 on Build GRCh38
rs781272434 Chr.10: 12088873-12088873 on Build GRCh38
rs774627644 Chr.10: 12089020-12089020 on Build GRCh38
rs747318060 Chr.10: 12089065-12089065 on Build GRCh38
rs566710328 Chr.10: 12088825-12088825 on Build GRCh38
rs113564561 Chr.10: 12088804-12088804 on Build GRCh38
rs996671991 Chr.10: 12089038-12089038 on Build GRCh38
rs533308379 Chr.10: 12088805-12088805 on Build GRCh38
rs1010386076 Chr.10: 12088725-12088725 on Build GRCh38
rs374421488 Chr.10: 12088981-12088981 on Build GRCh38
rs199630748 Chr.10: 12089114-12089114 on Build GRCh38
rs950297429 Chr.10: 12088751-12088751 on Build GRCh38
rs752385586 Chr.10: 12089072-12089072 on Build GRCh38
rs796635554 Chr.10: 12088933-12088933 on Build GRCh38
rs149066296 Chr.10: 12088989-12088989 on Build GRCh38
rs952045468 Chr.10: 12088823-12088823 on Build GRCh38
rs894765108 Chr.10: 12088738-12088738 on Build GRCh38
rs775906377 Chr.10: 12088935-12088935 on Build GRCh38
rs183528207 Chr.10: 12088832-12088832 on Build GRCh38
rs547072345 Chr.10: 12088807-12088807 on Build GRCh38
rs779049645 Chr.10: 12089099-12089099 on Build GRCh38
rs367981696 Chr.10: 12089005-12089005 on Build GRCh38
rs774028181 Chr.10: 12088943-12088943 on Build GRCh38
rs769073128 Chr.10: 12088994-12088994 on Build GRCh38
rs759022908 Chr.10: 12089060-12089060 on Build GRCh38
rs780665944 Chr.10: 12088584-12088584 on Build GRCh38
rs906197265 Chr.10: 12088795-12088795 on Build GRCh38
rs939920900 Chr.10: 12088850-12088850 on Build GRCh38
rs926037630 Chr.10: 12088707-12088707 on Build GRCh38
rs940381260 Chr.10: 12089040-12089040 on Build GRCh38
rs957443938 Chr.10: 12088972-12088972 on Build GRCh38
rs544852919 Chr.10: 12088704-12088704 on Build GRCh38
rs3740015 Chr.10: 12089082-12089082 on Build GRCh38
rs373335151 Chr.10: 12088940-12088940 on Build GRCh38
rs972568728 Chr.10: 12088856-12088856 on Build GRCh38
rs905526574 Chr.10: 12088709-12088709 on Build GRCh38
rs772236981 Chr.10: 12088982-12088982 on Build GRCh38
rs772239939 Chr.10: 12089108-12089108 on Build GRCh38
rs548060401 Chr.10: 12088906-12088906 on Build GRCh38
rs966556170 Chr.10: 12088759-12088759 on Build GRCh38
rs757746095 Chr.10: 12089079-12089079 on Build GRCh38
rs564595206 Chr.10: 12088783-12088783 on Build GRCh38
rs766253070 Chr.10: 12089051-12089051 on Build GRCh38
rs750343717 Chr.10: 12088964-12088964 on Build GRCh38
rs771829043 Chr.10: 12089033-12089033 on Build GRCh38
rs898903715 Chr.10: 12088831-12088831 on Build GRCh38
rs949876367 Chr.10: 12088904-12088904 on Build GRCh38
rs780313786 Chr.10: 12089004-12089004 on Build GRCh38
rs186713033 Chr.10: 12088965-12088965 on Build GRCh38
rs779881734 Chr.10: 12088721-12088721 on Build GRCh38
rs757640799 Chr.10: 12088995-12088995 on Build GRCh38
rs375938580 Chr.10: 12088715-12088715 on Build GRCh38

View Full Product Details