Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44275223-44275822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244880 c.471A>C p.K157N Chr.11: 44275654-44275654 on Build GRCh38
4145711 c.621A>G p.S207S Chr.11: 44275504-44275504 on Build GRCh38
1979325 c.646C>T p.R216W Chr.11: 44275479-44275479 on Build GRCh38
4032839 c.613G>A p.A205T Chr.11: 44275512-44275512 on Build GRCh38
4032837 c.681G>T p.E227D Chr.11: 44275444-44275444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773780472 Chr.11: 44275489-44275489 on Build GRCh38
rs774556630 Chr.11: 44275564-44275564 on Build GRCh38
rs1032983890 Chr.11: 44275712-44275712 on Build GRCh38
rs11037929 Chr.11: 44275703-44275703 on Build GRCh38
rs775809878 Chr.11: 44275687-44275687 on Build GRCh38
rs961458408 Chr.11: 44275435-44275435 on Build GRCh38
rs376402455 Chr.11: 44275446-44275446 on Build GRCh38
rs1014376517 Chr.11: 44275670-44275670 on Build GRCh38
rs11037930 Chr.11: 44275793-44275793 on Build GRCh38
rs774844132 Chr.11: 44275679-44275679 on Build GRCh38
rs180962051 Chr.11: 44275507-44275507 on Build GRCh38
rs150424138 Chr.11: 44275520-44275520 on Build GRCh38
rs61737295 Chr.11: 44275531-44275531 on Build GRCh38
rs755698536 Chr.11: 44275445-44275445 on Build GRCh38
rs550698378 Chr.11: 44275477-44275477 on Build GRCh38
rs749570490 Chr.11: 44275667-44275667 on Build GRCh38
rs145166164 Chr.11: 44275397-44275397 on Build GRCh38
rs941623068 Chr.11: 44275787-44275787 on Build GRCh38
rs1014714152 Chr.11: 44275478-44275478 on Build GRCh38
rs754154108 Chr.11: 44275303-44275303 on Build GRCh38
rs587776614 Chr.11: 44275621-44275621 on Build GRCh38
rs371740812 Chr.11: 44275638-44275638 on Build GRCh38
rs750924687 Chr.11: 44275319-44275319 on Build GRCh38
rs964859140 Chr.11: 44275297-44275297 on Build GRCh38
rs986694906 Chr.11: 44275376-44275376 on Build GRCh38
rs876657391 Chr.11: 44275622-44275622 on Build GRCh38
rs778173696 Chr.11: 44275663-44275663 on Build GRCh38
rs913641127 Chr.11: 44275272-44275272 on Build GRCh38
rs146633975 Chr.11: 44275354-44275354 on Build GRCh38
rs996306671 Chr.11: 44275572-44275572 on Build GRCh38
rs909511266 Chr.11: 44275786-44275786 on Build GRCh38
rs530779077 Chr.11: 44275324-44275324 on Build GRCh38
rs750173751 Chr.11: 44275623-44275623 on Build GRCh38
rs780830177 Chr.11: 44275345-44275345 on Build GRCh38
rs985107700 Chr.11: 44275778-44275778 on Build GRCh38
rs866675365 Chr.11: 44275511-44275511 on Build GRCh38
rs773510510 Chr.11: 44275548-44275548 on Build GRCh38
rs775067051 Chr.11: 44275587-44275587 on Build GRCh38
rs369517160 Chr.11: 44275318-44275318 on Build GRCh38
rs143599866 Chr.11: 44275704-44275704 on Build GRCh38
rs143620051 Chr.11: 44275556-44275556 on Build GRCh38
rs957960219 Chr.11: 44275715-44275715 on Build GRCh38
rs774447144 Chr.11: 44275603-44275603 on Build GRCh38
rs766990322 Chr.11: 44275405-44275405 on Build GRCh38
rs769556306 Chr.11: 44275601-44275601 on Build GRCh38
rs758249273 Chr.11: 44275529-44275529 on Build GRCh38
rs568863681 Chr.11: 44275498-44275498 on Build GRCh38
rs201303900 Chr.11: 44275547-44275547 on Build GRCh38
rs138094422 Chr.11: 44275473-44275473 on Build GRCh38
rs757563132 Chr.11: 44275701-44275701 on Build GRCh38
rs553689111 Chr.11: 44275651-44275651 on Build GRCh38
rs1024810014 Chr.11: 44275696-44275696 on Build GRCh38
rs587777700 Chr.11: 44275479-44275479 on Build GRCh38
rs574004169 Chr.11: 44275253-44275253 on Build GRCh38
rs140457891 Chr.11: 44275509-44275509 on Build GRCh38
rs926512059 Chr.11: 44275518-44275518 on Build GRCh38
rs948519420 Chr.11: 44275468-44275468 on Build GRCh38
rs751119502 Chr.11: 44275411-44275411 on Build GRCh38
rs760676423 Chr.11: 44275379-44275379 on Build GRCh38
rs954730060 Chr.11: 44275620-44275620 on Build GRCh38
rs932290500 Chr.11: 44275594-44275594 on Build GRCh38
rs932389281 Chr.11: 44275775-44275775 on Build GRCh38
rs768770098 Chr.11: 44275384-44275384 on Build GRCh38
rs777337755 Chr.11: 44275510-44275510 on Build GRCh38
rs914437943 Chr.11: 44275741-44275741 on Build GRCh38
rs104894193 Chr.11: 44275472-44275472 on Build GRCh38
rs201889959 Chr.11: 44275443-44275443 on Build GRCh38
rs200419726 Chr.11: 44275355-44275355 on Build GRCh38
rs772643460 Chr.11: 44275683-44275683 on Build GRCh38
rs770858153 Chr.11: 44275462-44275462 on Build GRCh38
rs945078994 Chr.11: 44275273-44275273 on Build GRCh38
rs560768905 Chr.11: 44275757-44275757 on Build GRCh38
rs104894197 Chr.11: 44275505-44275505 on Build GRCh38
rs374136767 Chr.11: 44275680-44275680 on Build GRCh38
rs756993500 Chr.11: 44275516-44275516 on Build GRCh38
rs11037927 Chr.11: 44275278-44275278 on Build GRCh38
rs587777701 Chr.11: 44275452-44275452 on Build GRCh38
rs989357708 Chr.11: 44275729-44275729 on Build GRCh38
rs753642057 Chr.11: 44275512-44275512 on Build GRCh38
rs78795983 Chr.11: 44275294-44275294 on Build GRCh38
rs745862299 Chr.11: 44275524-44275524 on Build GRCh38
rs776867365 Chr.11: 44275386-44275386 on Build GRCh38
rs370195126 Chr.11: 44275481-44275481 on Build GRCh38
rs199675093 Chr.11: 44275329-44275329 on Build GRCh38
rs1037162334 Chr.11: 44275650-44275650 on Build GRCh38
rs754454206 Chr.11: 44275424-44275424 on Build GRCh38
rs765070227 Chr.11: 44275607-44275607 on Build GRCh38
rs542676508 Chr.11: 44275754-44275754 on Build GRCh38
rs575657627 Chr.11: 44275747-44275747 on Build GRCh38
rs888638764 Chr.11: 44275375-44275375 on Build GRCh38
rs763754428 Chr.11: 44275591-44275591 on Build GRCh38
rs746505736 Chr.11: 44275335-44275335 on Build GRCh38
rs749075893 Chr.11: 44275449-44275449 on Build GRCh38
rs758432138 Chr.11: 44275628-44275628 on Build GRCh38
rs563651473 Chr.11: 44275742-44275742 on Build GRCh38
rs368396709 Chr.11: 44275578-44275578 on Build GRCh38
rs760043881 Chr.11: 44275588-44275588 on Build GRCh38
rs752434708 Chr.11: 44275441-44275441 on Build GRCh38
rs757581336 Chr.11: 44275317-44275317 on Build GRCh38
rs147204958 Chr.11: 44275636-44275636 on Build GRCh38
rs560707944 Chr.11: 44275336-44275336 on Build GRCh38
rs770130333 Chr.11: 44275482-44275482 on Build GRCh38
rs747909089 Chr.11: 44275353-44275353 on Build GRCh38
rs376287400 Chr.11: 44275597-44275597 on Build GRCh38
rs760145254 Chr.11: 44275576-44275576 on Build GRCh38
rs748173197 Chr.11: 44275544-44275544 on Build GRCh38
rs780739319 Chr.11: 44275440-44275440 on Build GRCh38
rs281865154 Chr.11: 44275494-44275494 on Build GRCh38
rs559809572 Chr.11: 44275264-44275264 on Build GRCh38
rs779790993 Chr.11: 44275631-44275631 on Build GRCh38
rs945149652 Chr.11: 44275550-44275550 on Build GRCh38
rs528383817 Chr.11: 44275356-44275356 on Build GRCh38
rs10769028 Chr.11: 44275504-44275504 on Build GRCh38
rs775610689 Chr.11: 44275369-44275369 on Build GRCh38
rs104894192 Chr.11: 44275389-44275389 on Build GRCh38
rs373163780 Chr.11: 44275320-44275320 on Build GRCh38
rs756876669 Chr.11: 44275598-44275598 on Build GRCh38
rs11037928 Chr.11: 44275396-44275396 on Build GRCh38
rs750798110 Chr.11: 44275404-44275404 on Build GRCh38
rs771264729 Chr.11: 44275674-44275674 on Build GRCh38
rs763344358 Chr.11: 44275495-44275495 on Build GRCh38

View Full Product Details